Assay Details
| Gene Symbol: ST14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021978.3 | NP_068813.1 | 5 - 6 | 794 | 66 | |
| GenBank mRNA | AB030036.1 | - | 5 - 6 | 616 | 66 | |
| AF057145.1 | - | 5 - 6 | 623 | 66 | ||
| AF118224.2 | - | 5 - 6 | 636 | 66 | ||
| AF133086.1 | - | 5 - 6 | 637 | 66 | ||
| AK314173.1 | - | 5 - 6 | 777 | 66 | ||
| BC030532.1 | - | 5 - 6 | 718 | 5213189 | 66 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC11001195.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR11015038.hg.1 | ENST00000278742.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11015038.hg.1 | ENST00000525779.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11015038.hg.1 | NM_021978.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11015038.hg.1 | TC11001195.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009524.hg.1 | ENST00000278742.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009524.hg.1 | ENST00000278742.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009524.hg.1 | ENST00000278742.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009524.hg.1 | ENST00000278742.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009524.hg.1 | ENST00000278742.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009524.hg.1 | ENST00000278742.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009524.hg.1 | ENST00000278742.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009524.hg.1 | ENST00000278742.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009524.hg.1 | ENST00000278742.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009524.hg.1 | ENST00000278742.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009524.hg.1 | ENST00000278742.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009524.hg.1 | ENST00000278742.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009524.hg.1 | ENST00000278742.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009524.hg.1 | ENST00000278742.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009524.hg.1 | ENST00000278742.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009524.hg.1 | ENST00000278742.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009524.hg.1 | ENST00000278742.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009524.hg.1 | ENST00000278742.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009524.hg.1 | ENST00000278742.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009524.hg.1 | ENST00000278742.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009524.hg.1 | ENST00000278742.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009524.hg.1 | ENST00000278742.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009524.hg.1 | ENST00000278742.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009524.hg.1 | ENST00000278742.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009524.hg.1 | ENST00000524718.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009524.hg.1 | ENST00000524718.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009524.hg.1 | ENST00000524718.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009524.hg.1 | ENST00000524718.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009524.hg.1 | ENST00000524718.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009524.hg.1 | ENST00000524718.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009524.hg.1 | ENST00000524718.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009524.hg.1 | ENST00000524718.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009524.hg.1 | ENST00000524718.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009524.hg.1 | ENST00000530376.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009524.hg.1 | ENST00000530376.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009524.hg.1 | ENST00000530376.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009524.hg.1 | ENST00000530376.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009524.hg.1 | ENST00000530376.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009524.hg.1 | ENST00000530376.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009524.hg.1 | ENST00000530532.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009524.hg.1 | ENST00000530532.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009524.hg.1 | ENST00000530532.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009524.hg.1 | ENST00000530532.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009524.hg.1 | ENST00000530532.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009524.hg.1 | ENST00000530532.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009524.hg.1 | ENST00000530532.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009524.hg.1 | NM_021978.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009524.hg.1 | NM_021978.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009524.hg.1 | NM_021978.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009524.hg.1 | NM_021978.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009524.hg.1 | NM_021978.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009524.hg.1 | NM_021978.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009524.hg.1 | NM_021978.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009524.hg.1 | NM_021978.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009524.hg.1 | NM_021978.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009524.hg.1 | NM_021978.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009524.hg.1 | NM_021978.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009524.hg.1 | NM_021978.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009524.hg.1 | NM_021978.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009524.hg.1 | NM_021978.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009524.hg.1 | NM_021978.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009524.hg.1 | NM_021978.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009524.hg.1 | NM_021978.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009524.hg.1 | NM_021978.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009524.hg.1 | NM_021978.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009524.hg.1 | NM_021978.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009524.hg.1 | NM_021978.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009524.hg.1 | NM_021978.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100009524.hg.1 | NM_021978.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100171612.hg.1 | ENST00000278742.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100171612.hg.1 | ENST00000524718.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100171612.hg.1 | ENST00000530532.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100171612.hg.1 | NM_021978.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100171612.hg.1 | ENST00000278742.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100171612.hg.1 | ENST00000524718.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100171612.hg.1 | ENST00000530532.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100171612.hg.1 | NM_021978.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100171612.hg.1 | TC1100009524.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1100009524.hg.1 | ENST00000278742.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009524.hg.1 | ENST00000278742.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009524.hg.1 | ENST00000278742.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009524.hg.1 | ENST00000278742.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009524.hg.1 | ENST00000278742.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009524.hg.1 | ENST00000278742.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009524.hg.1 | ENST00000278742.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009524.hg.1 | ENST00000278742.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009524.hg.1 | ENST00000278742.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009524.hg.1 | ENST00000278742.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009524.hg.1 | ENST00000278742.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009524.hg.1 | ENST00000278742.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009524.hg.1 | ENST00000278742.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009524.hg.1 | ENST00000278742.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009524.hg.1 | ENST00000278742.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009524.hg.1 | ENST00000278742.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009524.hg.1 | ENST00000278742.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009524.hg.1 | ENST00000278742.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009524.hg.1 | ENST00000278742.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009524.hg.1 | ENST00000278742.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009524.hg.1 | ENST00000278742.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009524.hg.1 | ENST00000278742.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009524.hg.1 | ENST00000278742.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009524.hg.1 | ENST00000278742.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009524.hg.1 | ENST00000524718.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009524.hg.1 | ENST00000524718.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009524.hg.1 | ENST00000524718.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009524.hg.1 | ENST00000524718.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009524.hg.1 | ENST00000524718.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009524.hg.1 | ENST00000524718.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009524.hg.1 | ENST00000524718.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009524.hg.1 | ENST00000524718.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009524.hg.1 | ENST00000524718.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009524.hg.1 | ENST00000530376.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009524.hg.1 | ENST00000530376.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009524.hg.1 | ENST00000530376.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009524.hg.1 | ENST00000530376.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009524.hg.1 | ENST00000530376.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009524.hg.1 | ENST00000530376.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009524.hg.1 | ENST00000530532.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009524.hg.1 | ENST00000530532.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009524.hg.1 | ENST00000530532.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009524.hg.1 | ENST00000530532.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009524.hg.1 | ENST00000530532.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009524.hg.1 | ENST00000530532.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009524.hg.1 | ENST00000530532.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100009524.hg.1 | NM_021978.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100009524.hg.1 | NM_021978.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100009524.hg.1 | NM_021978.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100009524.hg.1 | NM_021978.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100009524.hg.1 | NM_021978.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100009524.hg.1 | NM_021978.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100009524.hg.1 | NM_021978.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100009524.hg.1 | NM_021978.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100009524.hg.1 | NM_021978.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100009524.hg.1 | NM_021978.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100009524.hg.1 | NM_021978.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100009524.hg.1 | NM_021978.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100009524.hg.1 | NM_021978.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100009524.hg.1 | NM_021978.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100009524.hg.1 | NM_021978.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100009524.hg.1 | NM_021978.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100009524.hg.1 | NM_021978.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100009524.hg.1 | NM_021978.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100009524.hg.1 | NM_021978.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100009524.hg.1 | NM_021978.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100009524.hg.1 | NM_021978.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100009524.hg.1 | NM_021978.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100009524.hg.1 | NM_021978.3 | Refseq |

ST14
Genomic Map