Assay Details
Gene Symbol: PRKAB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_006253.4 | NP_006244.2 | 5 - 6 | 952 | 70 | |
XM_005253909.1 | XP_005253966.1 | 6 - 7 | 854 | 70 | ||
GenBank mRNA | AF022116.1 | - | 5 - 6 | 928 | 70 | |
AJ224515.1 | - | 5 - 6 | 663 | 70 | ||
AK301165.1 | - | 6 - 7 | 801 | 70 | ||
AK312495.1 | - | 6 - 7 | 766 | 70 | ||
BC001007.2 | - | 5 - 6 | 929 | 2961281 | 70 | |
BC001056.2 | - | 5 - 6 | 929 | 2961281 | 70 | |
BC001823.2 | - | 5 - 6 | 929 | 2961281 | 70 | |
BC017671.1 | - | 5 - 6 | 938 | 3884583 | 70 | |
BC018818.2 | - | 5 - 6 | 929 | 70 | ||
U83994.1 | - | 5 - 6 | 908 | 70 | ||
Y12556.1 | - | 5 - 6 | 744 | 70 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC12000936.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
protein phosphorylation
fatty acid biosynthetic process
cell cycle arrest
signal transduction
positive regulation of gene expression
macroautophagy
regulation of protein kinase activity
protein heterooligomerization
regulation of signal transduction by p53 class mediator
protein kinase activity
AMP-activated protein kinase activity
protein binding
protein kinase binding
protein phosphorylation
fatty acid biosynthetic process
cell cycle arrest
signal transduction
positive regulation of gene expression
macroautophagy
regulation of protein kinase activity
protein heterooligomerization
regulation of signal transduction by p53 class mediator
protein kinase activity
AMP-activated protein kinase activity
protein binding
protein kinase binding
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1200009103.hg.1 | ENST00000229328.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009103.hg.1 | ENST00000229328.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009103.hg.1 | ENST00000229328.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009103.hg.1 | ENST00000229328.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009103.hg.1 | ENST00000229328.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009103.hg.1 | ENST00000229328.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009103.hg.1 | ENST00000229328.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009103.hg.1 | ENST00000229328.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009103.hg.1 | ENST00000229328.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009103.hg.1 | ENST00000229328.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009103.hg.1 | ENST00000229328.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009103.hg.1 | ENST00000229328.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009103.hg.1 | ENST00000229328.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009103.hg.1 | ENST00000229328.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009103.hg.1 | ENST00000229328.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009103.hg.1 | ENST00000229328.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009103.hg.1 | ENST00000537057.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009103.hg.1 | ENST00000537057.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009103.hg.1 | ENST00000537057.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009103.hg.1 | ENST00000537057.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009103.hg.1 | ENST00000537400.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009103.hg.1 | ENST00000537400.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009103.hg.1 | ENST00000537400.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009103.hg.1 | ENST00000537400.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009103.hg.1 | ENST00000538271.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009103.hg.1 | ENST00000538271.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009103.hg.1 | ENST00000540121.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009103.hg.1 | ENST00000540121.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009103.hg.1 | ENST00000540121.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009103.hg.1 | ENST00000540121.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009103.hg.1 | ENST00000540121.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009103.hg.1 | ENST00000540121.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009103.hg.1 | ENST00000540121.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009103.hg.1 | ENST00000540121.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009103.hg.1 | ENST00000540121.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009103.hg.1 | ENST00000541640.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009103.hg.1 | ENST00000541640.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009103.hg.1 | ENST00000541640.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009103.hg.1 | ENST00000541640.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009103.hg.1 | ENST00000541640.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009103.hg.1 | ENST00000541640.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009103.hg.1 | ENST00000541640.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009103.hg.1 | ENST00000541640.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009103.hg.1 | ENST00000541640.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009103.hg.1 | ENST00000541640.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009103.hg.1 | ENST00000541640.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009103.hg.1 | ENST00000541640.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009103.hg.1 | ENST00000541640.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009103.hg.1 | ENST00000542698.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009103.hg.1 | ENST00000542698.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009103.hg.1 | ENST00000542698.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009103.hg.1 | ENST00000542698.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009103.hg.1 | ENST00000542698.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009103.hg.1 | ENST00000542698.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009103.hg.1 | ENST00000542698.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009103.hg.1 | ENST00000542698.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009103.hg.1 | ENST00000545223.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009103.hg.1 | ENST00000545223.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009103.hg.1 | ENST00000545223.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009103.hg.1 | ENST00000545223.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009103.hg.1 | ENST00000545223.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009103.hg.1 | ENST00000545223.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009103.hg.1 | ENST00000545870.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009103.hg.1 | ENST00000545870.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009103.hg.1 | ENST00000545870.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009103.hg.1 | ENST00000545870.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009103.hg.1 | ENST00000545870.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009103.hg.1 | ENST00000630317.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009103.hg.1 | ENST00000630317.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009103.hg.1 | NM_006253.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009103.hg.1 | NM_006253.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009103.hg.1 | NM_006253.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009103.hg.1 | NM_006253.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009103.hg.1 | NM_006253.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009103.hg.1 | NM_006253.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009103.hg.1 | NM_006253.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009103.hg.1 | NM_006253.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009103.hg.1 | NM_006253.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009103.hg.1 | NM_006253.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009103.hg.1 | NM_006253.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009103.hg.1 | NM_006253.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009103.hg.1 | NM_006253.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009103.hg.1 | NM_006253.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR12012139.hg.1 | ENST00000229328.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR12012139.hg.1 | ENST00000539596.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR12012139.hg.1 | ENST00000540121.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR12012139.hg.1 | ENST00000541640.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR12012139.hg.1 | NM_006253.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR12012139.hg.1 | TC12000936.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | PSR1200167272.hg.1 | ENST00000229328.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200167272.hg.1 | ENST00000538271.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200167272.hg.1 | ENST00000540121.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200167272.hg.1 | ENST00000541640.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200167272.hg.1 | ENST00000542698.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200167272.hg.1 | ENST00000545870.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200167272.hg.1 | NM_006253.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200167272.hg.1 | ENST00000229328.9 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200167272.hg.1 | ENST00000538271.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200167272.hg.1 | ENST00000540121.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200167272.hg.1 | ENST00000541640.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200167272.hg.1 | ENST00000542698.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200167272.hg.1 | ENST00000545870.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200167272.hg.1 | NM_006253.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200167272.hg.1 | TC1200009103.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC1200009103.hg.1 | ENST00000229328.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009103.hg.1 | ENST00000229328.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009103.hg.1 | ENST00000229328.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009103.hg.1 | ENST00000229328.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009103.hg.1 | ENST00000229328.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009103.hg.1 | ENST00000229328.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009103.hg.1 | ENST00000229328.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009103.hg.1 | ENST00000229328.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009103.hg.1 | ENST00000229328.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009103.hg.1 | ENST00000229328.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009103.hg.1 | ENST00000229328.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009103.hg.1 | ENST00000229328.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009103.hg.1 | ENST00000229328.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009103.hg.1 | ENST00000229328.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009103.hg.1 | ENST00000229328.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009103.hg.1 | ENST00000229328.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009103.hg.1 | ENST00000537057.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009103.hg.1 | ENST00000537057.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009103.hg.1 | ENST00000537057.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009103.hg.1 | ENST00000537057.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009103.hg.1 | ENST00000537400.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009103.hg.1 | ENST00000537400.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009103.hg.1 | ENST00000537400.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009103.hg.1 | ENST00000537400.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009103.hg.1 | ENST00000538271.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009103.hg.1 | ENST00000538271.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009103.hg.1 | ENST00000540121.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009103.hg.1 | ENST00000540121.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009103.hg.1 | ENST00000540121.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009103.hg.1 | ENST00000540121.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009103.hg.1 | ENST00000540121.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009103.hg.1 | ENST00000540121.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009103.hg.1 | ENST00000540121.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009103.hg.1 | ENST00000540121.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009103.hg.1 | ENST00000540121.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009103.hg.1 | ENST00000541640.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009103.hg.1 | ENST00000541640.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009103.hg.1 | ENST00000541640.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009103.hg.1 | ENST00000541640.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009103.hg.1 | ENST00000541640.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009103.hg.1 | ENST00000541640.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009103.hg.1 | ENST00000541640.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009103.hg.1 | ENST00000541640.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009103.hg.1 | ENST00000541640.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009103.hg.1 | ENST00000541640.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009103.hg.1 | ENST00000541640.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009103.hg.1 | ENST00000541640.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009103.hg.1 | ENST00000541640.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009103.hg.1 | ENST00000542698.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009103.hg.1 | ENST00000542698.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009103.hg.1 | ENST00000542698.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009103.hg.1 | ENST00000542698.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009103.hg.1 | ENST00000542698.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009103.hg.1 | ENST00000542698.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009103.hg.1 | ENST00000542698.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009103.hg.1 | ENST00000542698.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009103.hg.1 | ENST00000545223.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009103.hg.1 | ENST00000545223.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009103.hg.1 | ENST00000545223.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009103.hg.1 | ENST00000545223.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009103.hg.1 | ENST00000545223.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009103.hg.1 | ENST00000545223.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009103.hg.1 | ENST00000545870.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009103.hg.1 | ENST00000545870.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009103.hg.1 | ENST00000545870.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009103.hg.1 | ENST00000545870.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009103.hg.1 | ENST00000545870.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009103.hg.1 | ENST00000630317.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009103.hg.1 | ENST00000630317.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009103.hg.1 | NM_006253.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009103.hg.1 | NM_006253.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009103.hg.1 | NM_006253.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009103.hg.1 | NM_006253.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009103.hg.1 | NM_006253.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009103.hg.1 | NM_006253.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009103.hg.1 | NM_006253.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009103.hg.1 | NM_006253.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009103.hg.1 | NM_006253.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009103.hg.1 | NM_006253.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009103.hg.1 | NM_006253.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009103.hg.1 | NM_006253.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009103.hg.1 | NM_006253.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009103.hg.1 | NM_006253.4 | Refseq |
