Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011332.hg.1 |
ENST00000460282.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12013070.hg.1 |
ENST00000262225.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12013070.hg.1 |
ENST00000536875.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12013070.hg.1 |
ENST00000541504.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12013070.hg.1 |
NM_006815.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12013070.hg.1 |
TC12000988.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009247.hg.1 |
ENST00000262225.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009247.hg.1 |
ENST00000262225.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009247.hg.1 |
ENST00000262225.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009247.hg.1 |
ENST00000262225.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009247.hg.1 |
ENST00000262225.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009247.hg.1 |
ENST00000262225.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009247.hg.1 |
ENST00000262225.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009247.hg.1 |
ENST00000262225.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009247.hg.1 |
ENST00000262225.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009247.hg.1 |
ENST00000438031.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009247.hg.1 |
ENST00000438031.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009247.hg.1 |
ENST00000438031.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009247.hg.1 |
ENST00000438031.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009247.hg.1 |
ENST00000509052.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009247.hg.1 |
ENST00000509052.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009247.hg.1 |
ENST00000509052.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009247.hg.1 |
ENST00000509052.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009247.hg.1 |
ENST00000509052.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009247.hg.1 |
ENST00000543425.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009247.hg.1 |
ENST00000543425.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009247.hg.1 |
ENST00000543425.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009247.hg.1 |
ENST00000543425.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009247.hg.1 |
ENST00000543425.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009247.hg.1 |
ENST00000543425.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009247.hg.1 |
ENST00000543425.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009247.hg.1 |
ENST00000543425.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009247.hg.1 |
ENST00000544188.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009247.hg.1 |
ENST00000544188.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009247.hg.1 |
NM_006815.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009247.hg.1 |
NM_006815.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009247.hg.1 |
NM_006815.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009247.hg.1 |
NM_006815.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009247.hg.1 |
NM_006815.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009247.hg.1 |
NM_006815.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009247.hg.1 |
NM_006815.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009247.hg.1 |
NM_006815.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12013069.hg.1 |
ENST00000262225.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12013069.hg.1 |
ENST00000432022.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12013069.hg.1 |
ENST00000536875.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12013069.hg.1 |
ENST00000541504.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12013069.hg.1 |
NM_006815.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12013069.hg.1 |
TC12000988.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR12013067.hg.1 |
ENST00000262225.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12013067.hg.1 |
ENST00000432022.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12013067.hg.1 |
ENST00000541504.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12013067.hg.1 |
NM_006815.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12013067.hg.1 |
TC12000988.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR12013068.hg.1 |
ENST00000262225.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12013068.hg.1 |
ENST00000432022.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12013068.hg.1 |
ENST00000541504.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12013068.hg.1 |
NM_006815.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12013068.hg.1 |
TC12000988.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR12013074.hg.1 |
ENST00000262225.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12013074.hg.1 |
ENST00000432022.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12013074.hg.1 |
ENST00000509052.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12013074.hg.1 |
ENST00000536875.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12013074.hg.1 |
NM_006815.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12013074.hg.1 |
TC12000988.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1200168828.hg.1 |
ENST00000262225.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200168828.hg.1 |
ENST00000438031.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200168828.hg.1 |
ENST00000509052.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200168828.hg.1 |
ENST00000543425.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200168828.hg.1 |
ENST00000544188.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200168828.hg.1 |
NM_006815.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200168828.hg.1 |
ENST00000262225.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200168828.hg.1 |
ENST00000438031.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200168828.hg.1 |
ENST00000509052.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200168828.hg.1 |
ENST00000543425.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200168828.hg.1 |
ENST00000544188.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200168828.hg.1 |
NM_006815.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200168828.hg.1 |
TC1200009247.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1200168826.hg.1 |
ENST00000262225.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200168826.hg.1 |
ENST00000438031.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200168826.hg.1 |
ENST00000543425.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200168826.hg.1 |
ENST00000544188.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200168826.hg.1 |
NM_006815.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200168826.hg.1 |
ENST00000262225.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200168826.hg.1 |
ENST00000438031.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200168826.hg.1 |
ENST00000543425.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200168826.hg.1 |
ENST00000544188.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200168826.hg.1 |
NM_006815.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200168826.hg.1 |
TC1200009247.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0300011332.hg.1 |
ENST00000460282.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009247.hg.1 |
ENST00000262225.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009247.hg.1 |
ENST00000262225.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009247.hg.1 |
ENST00000262225.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009247.hg.1 |
ENST00000262225.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009247.hg.1 |
ENST00000262225.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009247.hg.1 |
ENST00000262225.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009247.hg.1 |
ENST00000262225.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009247.hg.1 |
ENST00000262225.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009247.hg.1 |
ENST00000262225.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009247.hg.1 |
ENST00000438031.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009247.hg.1 |
ENST00000438031.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009247.hg.1 |
ENST00000438031.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009247.hg.1 |
ENST00000438031.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009247.hg.1 |
ENST00000509052.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009247.hg.1 |
ENST00000509052.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009247.hg.1 |
ENST00000509052.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009247.hg.1 |
ENST00000509052.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009247.hg.1 |
ENST00000509052.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009247.hg.1 |
ENST00000543425.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009247.hg.1 |
ENST00000543425.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009247.hg.1 |
ENST00000543425.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009247.hg.1 |
ENST00000543425.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009247.hg.1 |
ENST00000543425.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009247.hg.1 |
ENST00000543425.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009247.hg.1 |
ENST00000543425.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009247.hg.1 |
ENST00000543425.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009247.hg.1 |
ENST00000544188.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009247.hg.1 |
ENST00000544188.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009247.hg.1 |
NM_006815.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009247.hg.1 |
NM_006815.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009247.hg.1 |
NM_006815.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009247.hg.1 |
NM_006815.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009247.hg.1 |
NM_006815.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009247.hg.1 |
NM_006815.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009247.hg.1 |
NM_006815.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009247.hg.1 |
NM_006815.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200168823.hg.1 |
ENST00000262225.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200168823.hg.1 |
ENST00000438031.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200168823.hg.1 |
ENST00000543425.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200168823.hg.1 |
NM_006815.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200168823.hg.1 |
ENST00000262225.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200168823.hg.1 |
ENST00000438031.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200168823.hg.1 |
ENST00000543425.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200168823.hg.1 |
NM_006815.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200168823.hg.1 |
TC1200009247.hg.1 |
Clariom D TC |