Assay Details
| Gene Symbol: CD79B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | S79249.1 | - | 2 - 3 | 124 | 63 | |
| X83539.1 | - | 2 - 3 | 124 | 63 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CD79B| Gene Symbol: CD79B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | S79249.1 | - | 2 - 3 | 124 | 63 | |
| X83539.1 | - | 2 - 3 | 124 | 63 | ||
Set Membership: |
|