Assay Details
| Gene Symbol: TMEM267 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005248335.3 | XP_005248392.1 | 2 - 3 | 220 | 111 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005248335.3 transcripts.
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0500010640.hg.1 | ENST00000397080.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010640.hg.1 | ENST00000397080.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010640.hg.1 | ENST00000397080.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010640.hg.1 | ENST00000397080.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010640.hg.1 | ENST00000397080.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010640.hg.1 | ENST00000397080.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010640.hg.1 | ENST00000397080.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010640.hg.1 | ENST00000397080.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010640.hg.1 | ENST00000397080.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010640.hg.1 | ENST00000397080.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010640.hg.1 | ENST00000397080.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010640.hg.1 | ENST00000397080.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010640.hg.1 | ENST00000500337.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010640.hg.1 | ENST00000500337.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010640.hg.1 | ENST00000500337.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010640.hg.1 | ENST00000500337.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010640.hg.1 | ENST00000500337.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010640.hg.1 | ENST00000500337.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010640.hg.1 | ENST00000500337.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010640.hg.1 | ENST00000500337.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010640.hg.1 | ENST00000500337.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010640.hg.1 | ENST00000500337.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010640.hg.1 | ENST00000500337.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010640.hg.1 | ENST00000500337.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010640.hg.1 | ENST00000500337.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010640.hg.1 | ENST00000500337.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010640.hg.1 | ENST00000500337.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010640.hg.1 | ENST00000506860.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010640.hg.1 | ENST00000506860.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010640.hg.1 | ENST00000506860.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010640.hg.1 | ENST00000506860.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010640.hg.1 | ENST00000506860.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010640.hg.1 | ENST00000506860.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010640.hg.1 | ENST00000510130.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010640.hg.1 | ENST00000510130.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010640.hg.1 | ENST00000510130.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010640.hg.1 | ENST00000510130.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010640.hg.1 | ENST00000510130.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010640.hg.1 | ENST00000510130.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010640.hg.1 | ENST00000511525.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010640.hg.1 | ENST00000511525.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010640.hg.1 | ENST00000511525.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010640.hg.1 | ENST00000511525.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010640.hg.1 | ENST00000512085.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010640.hg.1 | ENST00000512085.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010640.hg.1 | ENST00000512085.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010640.hg.1 | ENST00000512085.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010640.hg.1 | ENST00000512085.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010640.hg.1 | ENST00000512085.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010640.hg.1 | ENST00000512085.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010640.hg.1 | ENST00000512085.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010640.hg.1 | ENST00000512085.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010640.hg.1 | ENST00000512085.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010640.hg.1 | ENST00000512085.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010640.hg.1 | ENST00000512085.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010640.hg.1 | NM_022483.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010640.hg.1 | NM_022483.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010640.hg.1 | NM_022483.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010640.hg.1 | NM_022483.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010640.hg.1 | NM_022483.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010640.hg.1 | NM_022483.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010640.hg.1 | NM_022483.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010640.hg.1 | NM_022483.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010640.hg.1 | NM_022483.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010640.hg.1 | NM_022483.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010640.hg.1 | NM_022483.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010640.hg.1 | NM_022483.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010640.hg.1 | NM_022483.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010640.hg.1 | ENST00000397080.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010640.hg.1 | ENST00000397080.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010640.hg.1 | ENST00000397080.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010640.hg.1 | ENST00000397080.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010640.hg.1 | ENST00000397080.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010640.hg.1 | ENST00000397080.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010640.hg.1 | ENST00000397080.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010640.hg.1 | ENST00000397080.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010640.hg.1 | ENST00000397080.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010640.hg.1 | ENST00000397080.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010640.hg.1 | ENST00000397080.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010640.hg.1 | ENST00000397080.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010640.hg.1 | ENST00000500337.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010640.hg.1 | ENST00000500337.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010640.hg.1 | ENST00000500337.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010640.hg.1 | ENST00000500337.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010640.hg.1 | ENST00000500337.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010640.hg.1 | ENST00000500337.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010640.hg.1 | ENST00000500337.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010640.hg.1 | ENST00000500337.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010640.hg.1 | ENST00000500337.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010640.hg.1 | ENST00000500337.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010640.hg.1 | ENST00000500337.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010640.hg.1 | ENST00000500337.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010640.hg.1 | ENST00000500337.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010640.hg.1 | ENST00000500337.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010640.hg.1 | ENST00000500337.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010640.hg.1 | ENST00000506860.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010640.hg.1 | ENST00000506860.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010640.hg.1 | ENST00000506860.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010640.hg.1 | ENST00000506860.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010640.hg.1 | ENST00000506860.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010640.hg.1 | ENST00000506860.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010640.hg.1 | ENST00000510130.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010640.hg.1 | ENST00000510130.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010640.hg.1 | ENST00000510130.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010640.hg.1 | ENST00000510130.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010640.hg.1 | ENST00000510130.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010640.hg.1 | ENST00000510130.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010640.hg.1 | ENST00000511525.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010640.hg.1 | ENST00000511525.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010640.hg.1 | ENST00000511525.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010640.hg.1 | ENST00000511525.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010640.hg.1 | ENST00000512085.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010640.hg.1 | ENST00000512085.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010640.hg.1 | ENST00000512085.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010640.hg.1 | ENST00000512085.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010640.hg.1 | ENST00000512085.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010640.hg.1 | ENST00000512085.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010640.hg.1 | ENST00000512085.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010640.hg.1 | ENST00000512085.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010640.hg.1 | ENST00000512085.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010640.hg.1 | ENST00000512085.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010640.hg.1 | ENST00000512085.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010640.hg.1 | ENST00000512085.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010640.hg.1 | NM_022483.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010640.hg.1 | NM_022483.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010640.hg.1 | NM_022483.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010640.hg.1 | NM_022483.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010640.hg.1 | NM_022483.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010640.hg.1 | NM_022483.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010640.hg.1 | NM_022483.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010640.hg.1 | NM_022483.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010640.hg.1 | NM_022483.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010640.hg.1 | NM_022483.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010640.hg.1 | NM_022483.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010640.hg.1 | NM_022483.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010640.hg.1 | NM_022483.4 | Refseq |

TMEM267
Genomic Map