Assay Details
| Gene Symbol: LRP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_002332.2 | NP_002323.2 | 2 - 3 | 658 | 82 | |
| XM_017019303.1 | XP_016874792.1 | 2 - 3 | 756 | 82 | ||
| GenBank mRNA | AB209498.1 | - | 1 - 2 | 58 | 82 | |
| BC021204.2 | - | 2 - 3 | 247 | 82 | ||
| BC045107.1 | - | 2 - 3 | 441 | 82 | ||
| BC052593.1 | - | 2 - 3 | 664 | 82 | ||
| BC072015.1 | - | 2 - 3 | 664 | 82 | ||
| X13916.1 | - | 2 - 3 | 658 | 82 | ||
More Information
Set Membership: |
|

LRP1
Genomic Map