Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR08009587.hg.1 |
TC08000709.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR08009587.hg.1 |
ENST00000314393.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08009587.hg.1 |
NM_014943.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR08009591.hg.1 |
TC08000709.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR08009591.hg.1 |
ENST00000314393.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08009591.hg.1 |
NM_014943.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008715.hg.1 |
ENST00000314393.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008715.hg.1 |
ENST00000314393.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008715.hg.1 |
ENST00000314393.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008715.hg.1 |
ENST00000314393.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008715.hg.1 |
ENST00000314393.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008715.hg.1 |
ENST00000314393.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008715.hg.1 |
ENST00000314393.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008715.hg.1 |
ENST00000314393.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008715.hg.1 |
ENST00000534247.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008715.hg.1 |
ENST00000534247.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008715.hg.1 |
ENST00000534247.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008715.hg.1 |
ENST00000534247.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008715.hg.1 |
NM_014943.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008715.hg.1 |
NM_014943.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008715.hg.1 |
NM_014943.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008715.hg.1 |
NM_014943.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008715.hg.1 |
NM_014943.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008715.hg.1 |
NM_014943.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0800008715.hg.1 |
NM_014943.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR08009589.hg.1 |
TC08000709.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR08009589.hg.1 |
ENST00000314393.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08009589.hg.1 |
NM_014943.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR08009588.hg.1 |
TC08000709.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR08009588.hg.1 |
ENST00000314393.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08009588.hg.1 |
NM_014943.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR08009590.hg.1 |
TC08000709.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR08009590.hg.1 |
ENST00000314393.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR08009590.hg.1 |
NM_014943.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800159709.hg.1 |
ENST00000314393.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800159709.hg.1 |
ENST00000534247.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800159709.hg.1 |
NM_014943.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800159709.hg.1 |
ENST00000314393.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800159709.hg.1 |
ENST00000534247.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800159709.hg.1 |
NM_014943.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800159709.hg.1 |
TC0800008715.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0800008715.hg.1 |
ENST00000314393.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008715.hg.1 |
ENST00000314393.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008715.hg.1 |
ENST00000314393.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008715.hg.1 |
ENST00000314393.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008715.hg.1 |
ENST00000314393.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008715.hg.1 |
ENST00000314393.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008715.hg.1 |
ENST00000314393.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008715.hg.1 |
ENST00000314393.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008715.hg.1 |
ENST00000534247.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008715.hg.1 |
ENST00000534247.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008715.hg.1 |
ENST00000534247.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008715.hg.1 |
ENST00000534247.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800008715.hg.1 |
NM_014943.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800008715.hg.1 |
NM_014943.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800008715.hg.1 |
NM_014943.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800008715.hg.1 |
NM_014943.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800008715.hg.1 |
NM_014943.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800008715.hg.1 |
NM_014943.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800008715.hg.1 |
NM_014943.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800159708.hg.1 |
ENST00000314393.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800159708.hg.1 |
ENST00000534247.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800159708.hg.1 |
NM_014943.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800159708.hg.1 |
ENST00000314393.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800159708.hg.1 |
ENST00000534247.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800159708.hg.1 |
NM_014943.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800159708.hg.1 |
TC0800008715.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0800159704.hg.1 |
ENST00000314393.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800159704.hg.1 |
NM_014943.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800159704.hg.1 |
ENST00000314393.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800159704.hg.1 |
NM_014943.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800159704.hg.1 |
TC0800008715.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0800159705.hg.1 |
ENST00000314393.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800159705.hg.1 |
NM_014943.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800159705.hg.1 |
ENST00000314393.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800159705.hg.1 |
NM_014943.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800159705.hg.1 |
TC0800008715.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0800159707.hg.1 |
ENST00000314393.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800159707.hg.1 |
ENST00000534247.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800159707.hg.1 |
NM_014943.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800159707.hg.1 |
ENST00000314393.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800159707.hg.1 |
ENST00000534247.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800159707.hg.1 |
NM_014943.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800159707.hg.1 |
TC0800008715.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0800159710.hg.1 |
ENST00000314393.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800159710.hg.1 |
NM_014943.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800159710.hg.1 |
ENST00000314393.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0800159710.hg.1 |
NM_014943.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0800159710.hg.1 |
TC0800008715.hg.1 |
Clariom D TC |