Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008975.hg.1 |
ENST00000289448.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008975.hg.1 |
ENST00000289448.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008975.hg.1 |
ENST00000289448.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008975.hg.1 |
ENST00000289448.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008975.hg.1 |
ENST00000289448.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008975.hg.1 |
ENST00000289448.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008975.hg.1 |
NM_021182.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008975.hg.1 |
NM_021182.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008975.hg.1 |
NM_021182.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008975.hg.1 |
NM_021182.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008975.hg.1 |
NM_021182.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500008975.hg.1 |
NM_021182.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR05011208.hg.1 |
TC05000789.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR05011208.hg.1 |
ENST00000289448.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05011208.hg.1 |
NM_021182.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR05011206.hg.1 |
TC05000789.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR05011206.hg.1 |
ENST00000289448.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05011206.hg.1 |
NM_021182.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR05011207.hg.1 |
TC05000789.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR05011207.hg.1 |
ENST00000289448.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05011207.hg.1 |
NM_021182.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR05011205.hg.1 |
TC05000789.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR05011205.hg.1 |
ENST00000289448.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05011205.hg.1 |
NM_021182.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500162476.hg.1 |
ENST00000289448.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500162476.hg.1 |
NM_021182.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500162476.hg.1 |
ENST00000289448.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500162476.hg.1 |
NM_021182.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500162476.hg.1 |
TC0500008975.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0500008975.hg.1 |
ENST00000289448.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008975.hg.1 |
ENST00000289448.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008975.hg.1 |
ENST00000289448.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008975.hg.1 |
ENST00000289448.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008975.hg.1 |
ENST00000289448.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008975.hg.1 |
ENST00000289448.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500008975.hg.1 |
NM_021182.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500008975.hg.1 |
NM_021182.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500008975.hg.1 |
NM_021182.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500008975.hg.1 |
NM_021182.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500008975.hg.1 |
NM_021182.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500008975.hg.1 |
NM_021182.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500162470.hg.1 |
ENST00000289448.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500162470.hg.1 |
NM_021182.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500162470.hg.1 |
ENST00000289448.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500162470.hg.1 |
NM_021182.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500162470.hg.1 |
TC0500008975.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0500162473.hg.1 |
ENST00000289448.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500162473.hg.1 |
NM_021182.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500162473.hg.1 |
ENST00000289448.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500162473.hg.1 |
NM_021182.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500162473.hg.1 |
TC0500008975.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0500162475.hg.1 |
ENST00000289448.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500162475.hg.1 |
NM_021182.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500162475.hg.1 |
ENST00000289448.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500162475.hg.1 |
NM_021182.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500162475.hg.1 |
TC0500008975.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0500162472.hg.1 |
ENST00000289448.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500162472.hg.1 |
NM_021182.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500162472.hg.1 |
ENST00000289448.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500162472.hg.1 |
NM_021182.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500162472.hg.1 |
TC0500008975.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0500162478.hg.1 |
ENST00000289448.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500162478.hg.1 |
NM_021182.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500162478.hg.1 |
ENST00000289448.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500162478.hg.1 |
NM_021182.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500162478.hg.1 |
TC0500008975.hg.1 |
Clariom D TC |