Assay Details
| Gene Symbol: AGO4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_017629.3 | NP_060099.2 | 2 - 3 | 371 | 136 | |
| XM_005270578.3 | XP_005270635.1 | 2 - 3 | 256 | 136 | ||
| XM_017000529.1 | XP_016856018.1 | 2 - 3 | 268 | 136 | ||
| GenBank mRNA | AB046787.1 | - | 2 - 3 | 371 | 136 | |
| AL831908.1 | - | 1 - 2 | 158 | 136 | ||
| BC152450.1 | - | 2 - 3 | 371 | 8862582 | 136 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01000462.hg.1
Additional Information:
This assay is in the 5'UTR region of the XM_005270578.3 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
mRNA catabolic process
Wnt signaling pathway, calcium modulating pathway
RNA secondary structure unwinding
pre-miRNA processing
posttranscriptional gene silencing by RNA
production of miRNAs involved in gene silencing by miRNA
miRNA mediated inhibition of translation
mRNA cleavage involved in gene silencing by miRNA
miRNA loading onto RISC involved in gene silencing by miRNA
phosphatidylinositol-mediated signaling
mRNA cleavage involved in gene silencing by siRNA
double-stranded RNA binding
single-stranded RNA binding
protein binding
miRNA binding
mRNA catabolic process
Wnt signaling pathway, calcium modulating pathway
RNA secondary structure unwinding
pre-miRNA processing
posttranscriptional gene silencing by RNA
production of miRNAs involved in gene silencing by miRNA
miRNA mediated inhibition of translation
mRNA cleavage involved in gene silencing by miRNA
miRNA loading onto RISC involved in gene silencing by miRNA
phosphatidylinositol-mediated signaling
mRNA cleavage involved in gene silencing by siRNA
double-stranded RNA binding
single-stranded RNA binding
protein binding
miRNA binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR01007104.hg.1 | TC01000462.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01007104.hg.1 | ENST00000373210.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01007104.hg.1 | NM_017629.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007784.hg.1 | ENST00000373210.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007784.hg.1 | ENST00000373210.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007784.hg.1 | ENST00000373210.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007784.hg.1 | ENST00000373210.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007784.hg.1 | ENST00000373210.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007784.hg.1 | ENST00000373210.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007784.hg.1 | ENST00000373210.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007784.hg.1 | ENST00000373210.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007784.hg.1 | ENST00000373210.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007784.hg.1 | ENST00000373210.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007784.hg.1 | ENST00000373210.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007784.hg.1 | ENST00000373210.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007784.hg.1 | ENST00000373210.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007784.hg.1 | ENST00000373210.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007784.hg.1 | ENST00000373210.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007784.hg.1 | ENST00000373210.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007784.hg.1 | ENST00000373210.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007784.hg.1 | ENST00000373210.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007784.hg.1 | ENST00000373210.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007784.hg.1 | ENST00000373210.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007784.hg.1 | ENST00000373210.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007784.hg.1 | ENST00000373210.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007784.hg.1 | ENST00000373210.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007784.hg.1 | ENST00000373210.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007784.hg.1 | ENST00000373210.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007784.hg.1 | ENST00000373210.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007784.hg.1 | ENST00000373210.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007784.hg.1 | ENST00000488778.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007784.hg.1 | ENST00000488778.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007784.hg.1 | ENST00000488778.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007784.hg.1 | ENST00000488778.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007784.hg.1 | ENST00000496257.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007784.hg.1 | ENST00000496257.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007784.hg.1 | ENST00000496257.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007784.hg.1 | NM_017629.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007784.hg.1 | NM_017629.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007784.hg.1 | NM_017629.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007784.hg.1 | NM_017629.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007784.hg.1 | NM_017629.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007784.hg.1 | NM_017629.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007784.hg.1 | NM_017629.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007784.hg.1 | NM_017629.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007784.hg.1 | NM_017629.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007784.hg.1 | NM_017629.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007784.hg.1 | NM_017629.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007784.hg.1 | NM_017629.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007784.hg.1 | NM_017629.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007784.hg.1 | NM_017629.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007784.hg.1 | NM_017629.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007784.hg.1 | NM_017629.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007784.hg.1 | NM_017629.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007784.hg.1 | NM_017629.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007784.hg.1 | NM_017629.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007784.hg.1 | NM_017629.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007784.hg.1 | NM_017629.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007784.hg.1 | NM_017629.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007784.hg.1 | NM_017629.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007784.hg.1 | NM_017629.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007784.hg.1 | NM_017629.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007784.hg.1 | NM_017629.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01007101.hg.1 | TC01000462.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01007101.hg.1 | ENST00000373210.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01007101.hg.1 | NM_017629.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01007103.hg.1 | TC01000462.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01007103.hg.1 | ENST00000373210.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01007103.hg.1 | NM_017629.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007784.hg.1 | ENST00000373210.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007784.hg.1 | ENST00000373210.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007784.hg.1 | ENST00000373210.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007784.hg.1 | ENST00000373210.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007784.hg.1 | ENST00000373210.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007784.hg.1 | ENST00000373210.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007784.hg.1 | ENST00000373210.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007784.hg.1 | ENST00000373210.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007784.hg.1 | ENST00000373210.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007784.hg.1 | ENST00000373210.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007784.hg.1 | ENST00000373210.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007784.hg.1 | ENST00000373210.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007784.hg.1 | ENST00000373210.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007784.hg.1 | ENST00000373210.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007784.hg.1 | ENST00000373210.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007784.hg.1 | ENST00000373210.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007784.hg.1 | ENST00000373210.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007784.hg.1 | ENST00000373210.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007784.hg.1 | ENST00000373210.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007784.hg.1 | ENST00000373210.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007784.hg.1 | ENST00000373210.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007784.hg.1 | ENST00000373210.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007784.hg.1 | ENST00000373210.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007784.hg.1 | ENST00000373210.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007784.hg.1 | ENST00000373210.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007784.hg.1 | ENST00000373210.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007784.hg.1 | ENST00000373210.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007784.hg.1 | ENST00000488778.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007784.hg.1 | ENST00000488778.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007784.hg.1 | ENST00000488778.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007784.hg.1 | ENST00000488778.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007784.hg.1 | ENST00000496257.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007784.hg.1 | ENST00000496257.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007784.hg.1 | ENST00000496257.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007784.hg.1 | NM_017629.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007784.hg.1 | NM_017629.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007784.hg.1 | NM_017629.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007784.hg.1 | NM_017629.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007784.hg.1 | NM_017629.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007784.hg.1 | NM_017629.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007784.hg.1 | NM_017629.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007784.hg.1 | NM_017629.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007784.hg.1 | NM_017629.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007784.hg.1 | NM_017629.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007784.hg.1 | NM_017629.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007784.hg.1 | NM_017629.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007784.hg.1 | NM_017629.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007784.hg.1 | NM_017629.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007784.hg.1 | NM_017629.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007784.hg.1 | NM_017629.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007784.hg.1 | NM_017629.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007784.hg.1 | NM_017629.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007784.hg.1 | NM_017629.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007784.hg.1 | NM_017629.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007784.hg.1 | NM_017629.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007784.hg.1 | NM_017629.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007784.hg.1 | NM_017629.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007784.hg.1 | NM_017629.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007784.hg.1 | NM_017629.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007784.hg.1 | NM_017629.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100154767.hg.1 | ENST00000373210.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100154767.hg.1 | NM_017629.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100154767.hg.1 | ENST00000373210.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100154767.hg.1 | NM_017629.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100154767.hg.1 | TC0100007784.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100154764.hg.1 | ENST00000373210.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100154764.hg.1 | NM_017629.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100154764.hg.1 | ENST00000373210.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100154764.hg.1 | NM_017629.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100154764.hg.1 | TC0100007784.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100154768.hg.1 | ENST00000373210.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100154768.hg.1 | NM_017629.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100154768.hg.1 | ENST00000373210.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100154768.hg.1 | NM_017629.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100154768.hg.1 | TC0100007784.hg.1 | Clariom D TC |

AGO4
Genomic Map