Assay Details
Gene Symbol: TAS2R13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_023920.2 | NP_076409.1 | 1 | 842 | 153 | |
GenBank mRNA | BC069376.1 | - | 1 | 578 | 7262240 | 153 |
BC095518.1 | - | 1 | 842 | 30706594 | 153 | |
BC115402.1 | - | 1 | 571 | 153 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC12001234.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Human Whole Genome ChIP Tiling Array | A_17_P28622208 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P28622239 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P08190306 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P08190288 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P28622238 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P28622237 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P28622246 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P08190296 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P28622222 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P08190307 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P28622221 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P28622248 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P28622219 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P28622212 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P08190328 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P28622233 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P28622207 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P08190324 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P28622213 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P28622215 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P08190311 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P28622250 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P08190312 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P28622220 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P28622234 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P08190309 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P08190337 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P28622210 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P28622206 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P08190308 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P08190334 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P28622243 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P28622242 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P28622218 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P28622216 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P28622245 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P08190290 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_23_P99080 | - | - |
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_44k_23_P99080 | - | - |
Agilent Whole Human Genome Oligo Microarray 4x44K | 28309 | - | - |
Agilent Human Genome CGH Microarray 44B | A_14_P113737 | - | - |
Agilent Human Genome CGH Microarray 44B | A_14_P200339 | - | - |
Agilent Human Genome CGH Microarray 44A | A_14_P113737 | - | - |
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 8527 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_23_P99080 | - | - |
SurePrint G3 Human GE Microarray G4851A | A_23_P99080 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08190304 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28622216 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28622220 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28622229 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08190312 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28622210 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08190306 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08190315 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28622225 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28622218 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08190311 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28622239 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28622207 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08190324 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28622221 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28622231 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28622233 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28622208 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28622234 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08190308 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28622206 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28622243 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08190309 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28622213 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08190307 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28622242 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08190290 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28622215 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28622237 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28622212 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08190303 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08190308 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28622237 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28622206 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28622210 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28622229 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08190303 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28622225 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28622242 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28622239 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28622218 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28622213 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28622231 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28622233 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08190309 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08190315 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28622234 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28622220 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28622212 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28622207 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28622215 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08190290 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08190311 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08190312 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28622216 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08190304 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08190307 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08190306 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P08190324 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28622243 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28622208 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28622221 | - | - |
Whole Human Genome Oligo Microarray G4112A | A_23_P99080 | - | - |
Agilent Human Genome CGH Microarray 105A (G4412A) | A_14_P200339 | - | - |
Agilent Human Genome CGH Microarray 105A (G4412A) | A_14_P113737 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P08190292 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P08190320 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P08190312 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P08190315 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P08190286 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P08190308 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P08190306 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P08190298 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P08190311 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P08190323 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P08190295 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P08190325 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P08190310 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P08190304 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P08190316 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P08190319 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P08190314 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P08190282 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P08190324 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P08190303 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P08190289 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P08190300 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P08190293 | - | - |
Agilent Human Genome 244K CGH Microarray (Alpha Test) | A_14_P200339 | - | - |
Agilent Human Genome 244K CGH Microarray (Alpha Test) | A_14_P113737 | - | - |
Agilent Whole Human Genome Microarray 4x44K G4112F | A_23_P99080 | - | - |
Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 | 22451 | - | - |
Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 | 2217 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P08190300 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P08190324 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P08190332 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P08190307 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P08190325 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P08190293 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P08190299 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P08190295 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P08190339 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P08190319 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P08190286 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P08190310 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P08190311 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P08190331 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P08190288 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P08190292 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P08190303 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P08190314 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P08190336 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P08190328 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P08190298 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P08190323 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P08190316 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P08190289 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P08190306 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Gene 1.0 ST Array | NM_023920_at | - | - |
Affymetrix Human Gene 1.0 ST Array | 7961277 | - | - |
Affymetrix Human Gene 1.0 ST Array | 7961276 | - | - |
Affymetrix Human Gene 1.0 ST Array | 7961278 | - | - |
Affymetrix Human Gene 1.1 ST Array | 7961275 | - | - |
Affymetrix HT HG-U133+ PM Array Plate | 221395_PM_at | - | - |
Affymetrix Human Genome U219 Array | 11732780_at | - | - |
Affymetrix Human Genome U219 Array | 11732780_PM_at | - | - |
Affymetrix Human Transcriptome Array 2.0 | TC1200009921.hg.1 | ENST00000390677.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009921.hg.1 | ENST00000390677.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009921.hg.1 | ENST00000390677.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200009921.hg.1 | NM_023920.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009921.hg.1 | NM_023920.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200009921.hg.1 | NM_023920.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR12016264.hg.1 | ENST00000390677.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR12016264.hg.1 | NM_023920.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR12016264.hg.1 | TC12001234.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | TC1200009921.hg.1 | ENST00000390677.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009921.hg.1 | ENST00000390677.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009921.hg.1 | ENST00000390677.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200009921.hg.1 | NM_023920.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009921.hg.1 | NM_023920.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1200009921.hg.1 | NM_023920.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200175012.hg.1 | ENST00000390677.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200175012.hg.1 | NM_023920.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200175012.hg.1 | ENST00000390677.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200175012.hg.1 | NM_023920.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200175012.hg.1 | TC1200009921.hg.1 | Clariom D TC |
Human Genome U133 Plus 2.0 Array | 221395_PM_at | - | - |
Human Genome U133 Plus 2.0 Array | 221395_at | - | - |
Human Genome U133A 2.0 Array | 221395_PM_at | - | - |
Human Genome U133A 2.0 Array | 221395_at | - | - |
Affymetrix GeneChip Human Genome U133 Plus 2.0 Array based on a custom CDF (REFSEQ_7) | NM_023920_at | - | - |
AffymetrixPathwork Diagnostics Pathchip | pw510204 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 221395_at11 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 221395_at10 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 221395_at3 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 221395_at2 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 221395_at1 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 221395_at8 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 221395_at7 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 221395_at9 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 221395_at4 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 221395_at5 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 221395_at6 | - | - |
Human Genome U133 Set | 221395_PM_at | - | - |
Human Genome U133 Set | 221395_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A047325 | - | - |
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A047325 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Human Genome Survey Microarray Version 2.0 | 107242 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
HumanHT-12 v4 Expression BeadChip | ILMN_1696528 | NM_023920.2 | human_transcriptome |
HumanHT-12 v4 Expression BeadChip | ILMN_1696528 | BC095518.1 | human_transcriptome |
Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1696528 | NM_023920.2 | human_transcriptome |
Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1696528 | BC095518.1 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_1696528 | NM_023920.2 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_1696528 | BC095518.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1696528 | NM_023920.2 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1696528 | BC095518.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1696528 | NM_023920.2 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1696528 | BC095518.1 | human_transcriptome |
Sentrix Human-6 Expression BeadChip | GI_12965177-S | BC115403.1 | human_transcriptome |
Sentrix Human-6 Expression BeadChip | GI_12965177-S | NM_023920.2 | human_transcriptome |
Sentrix Human-6 Expression BeadChip | GI_12965177-S | BC069376.1 | human_transcriptome |
Sentrix Human-6 Expression BeadChip | GI_12965177-S | BC095518.1 | human_transcriptome |
Sentrix Human-6 Expression BeadChip | GI_12965177-S | BC115402.1 | human_transcriptome |
Illumina HumanRef-8 v2 Expression BeadChip | GI_21919-S | - | - |
Illumina human-6 v1.0 expression beadchip | 0004760017 | - | - |
Illumina humanRef-8 v1.0 expression beadchip | 0004760017 | - | - |
Illumina human-6 v2.0 expression beadchip | ILMN_1696528 | NM_023920.2 | human_transcriptome |
Illumina human-6 v2.0 expression beadchip | ILMN_1696528 | BC095518.1 | human_transcriptome |
Illumina humanRef-8 v2.0 expression beadchip | ILMN_1696528 | NM_023920.2 | human_transcriptome |
Illumina humanRef-8 v2.0 expression beadchip | ILMN_1696528 | BC095518.1 | human_transcriptome |
Illumina Sentrix Human-6 v2 Expression BeadChip | 1450100 | - | - |
Illumina HumanRef-6 v2.0 expression beadchip | ILMN_21919 | - | - |
Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1696528 | NM_023920.2 | human_transcriptome |
Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1696528 | BC095518.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1696528 | NM_023920.2 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1696528 | BC095518.1 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1696528 | NM_023920.2 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1696528 | BC095518.1 | human_transcriptome |
Illumina HumanWG-6_V2_0_R2 | ILMN_1696528 | NM_023920.2 | human_transcriptome |
Illumina HumanWG-6_V2_0_R2 | ILMN_1696528 | BC095518.1 | human_transcriptome |
Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1696528 | NM_023920.2 | human_transcriptome |
Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1696528 | BC095518.1 | human_transcriptome |
