Assay Details
Gene Symbol: CFAP100 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_182628.2 | NP_872434.2 | 14 - 15 | 1521 | 64 | |
XM_006713623.2 | XP_006713686.1 | 15 - 16 | 1672 | 64 | ||
GenBank mRNA | AK097402.1 | - | 14 - 15 | 1521 | 64 | |
BC101366.1 | - | 13 - 14 | 1454 | 40025823 | 64 | |
BC101367.2 | - | 13 - 14 | 1448 | 40025824 | 64 | |
BC101368.1 | - | 13 - 14 | 1451 | 40025828 | 64 | |
BC101369.1 | - | 13 - 14 | 1448 | 40025829 | 64 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC03000657.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0300008672.hg.1 | ENST00000352312.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008672.hg.1 | ENST00000352312.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008672.hg.1 | ENST00000352312.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008672.hg.1 | ENST00000352312.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008672.hg.1 | ENST00000352312.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008672.hg.1 | ENST00000352312.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008672.hg.1 | ENST00000352312.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008672.hg.1 | ENST00000352312.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008672.hg.1 | ENST00000352312.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008672.hg.1 | ENST00000352312.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008672.hg.1 | ENST00000352312.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008672.hg.1 | ENST00000352312.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008672.hg.1 | ENST00000352312.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008672.hg.1 | ENST00000352312.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008672.hg.1 | ENST00000352312.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008672.hg.1 | ENST00000352312.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008672.hg.1 | ENST00000352312.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008672.hg.1 | ENST00000352312.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008672.hg.1 | ENST00000352312.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008672.hg.1 | ENST00000352312.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008672.hg.1 | ENST00000352312.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008672.hg.1 | ENST00000352312.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008672.hg.1 | ENST00000352312.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008672.hg.1 | ENST00000352312.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008672.hg.1 | ENST00000503905.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008672.hg.1 | ENST00000503905.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008672.hg.1 | ENST00000503905.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008672.hg.1 | ENST00000505024.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008672.hg.1 | ENST00000505024.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008672.hg.1 | ENST00000505024.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008672.hg.1 | ENST00000505024.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008672.hg.1 | ENST00000505024.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008672.hg.1 | ENST00000505024.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008672.hg.1 | ENST00000505024.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008672.hg.1 | ENST00000505024.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008672.hg.1 | ENST00000505024.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008672.hg.1 | ENST00000505024.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008672.hg.1 | ENST00000505024.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008672.hg.1 | ENST00000505024.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008672.hg.1 | ENST00000505024.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008672.hg.1 | ENST00000505024.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008672.hg.1 | ENST00000505024.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008672.hg.1 | ENST00000505024.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008672.hg.1 | ENST00000505024.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008672.hg.1 | ENST00000505024.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008672.hg.1 | ENST00000505024.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008672.hg.1 | ENST00000505024.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008672.hg.1 | ENST00000505024.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008672.hg.1 | ENST00000506204.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008672.hg.1 | ENST00000506204.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008672.hg.1 | ENST00000506204.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008672.hg.1 | ENST00000506204.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008672.hg.1 | ENST00000506204.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008672.hg.1 | ENST00000506204.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008672.hg.1 | ENST00000507830.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008672.hg.1 | ENST00000507830.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008672.hg.1 | ENST00000507830.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008672.hg.1 | ENST00000507830.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008672.hg.1 | ENST00000508140.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008672.hg.1 | ENST00000508140.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008672.hg.1 | ENST00000508140.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008672.hg.1 | ENST00000508140.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008672.hg.1 | ENST00000508140.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008672.hg.1 | ENST00000508140.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008672.hg.1 | ENST00000508140.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008672.hg.1 | ENST00000508140.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008672.hg.1 | ENST00000510833.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008672.hg.1 | ENST00000510833.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008672.hg.1 | ENST00000510833.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008672.hg.1 | ENST00000510833.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008672.hg.1 | ENST00000510833.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008672.hg.1 | ENST00000510833.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008672.hg.1 | ENST00000510833.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008672.hg.1 | ENST00000510833.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008672.hg.1 | ENST00000533201.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008672.hg.1 | ENST00000533201.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300008672.hg.1 | NM_182628.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300008672.hg.1 | NM_182628.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300008672.hg.1 | NM_182628.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300008672.hg.1 | NM_182628.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300008672.hg.1 | NM_182628.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300008672.hg.1 | NM_182628.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300008672.hg.1 | NM_182628.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300008672.hg.1 | NM_182628.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300008672.hg.1 | NM_182628.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300008672.hg.1 | NM_182628.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300008672.hg.1 | NM_182628.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300008672.hg.1 | NM_182628.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300008672.hg.1 | NM_182628.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300008672.hg.1 | NM_182628.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300008672.hg.1 | NM_182628.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300008672.hg.1 | NM_182628.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300008672.hg.1 | NM_182628.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300008672.hg.1 | NM_182628.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300008672.hg.1 | NM_182628.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300008672.hg.1 | NM_182628.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300008672.hg.1 | NM_182628.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300008672.hg.1 | NM_182628.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300008672.hg.1 | NM_182628.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300008672.hg.1 | NM_182628.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR03011806.hg.1 | TC03000657.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR03011806.hg.1 | ENST00000352312.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR03011806.hg.1 | ENST00000393425.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR03011806.hg.1 | ENST00000505024.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR03011806.hg.1 | ENST00000506204.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR03011806.hg.1 | NM_182628.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300008672.hg.1 | ENST00000352312.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008672.hg.1 | ENST00000352312.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008672.hg.1 | ENST00000352312.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008672.hg.1 | ENST00000352312.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008672.hg.1 | ENST00000352312.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008672.hg.1 | ENST00000352312.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008672.hg.1 | ENST00000352312.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008672.hg.1 | ENST00000352312.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008672.hg.1 | ENST00000352312.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008672.hg.1 | ENST00000352312.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008672.hg.1 | ENST00000352312.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008672.hg.1 | ENST00000352312.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008672.hg.1 | ENST00000352312.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008672.hg.1 | ENST00000352312.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008672.hg.1 | ENST00000352312.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008672.hg.1 | ENST00000352312.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008672.hg.1 | ENST00000352312.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008672.hg.1 | ENST00000352312.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008672.hg.1 | ENST00000352312.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008672.hg.1 | ENST00000352312.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008672.hg.1 | ENST00000352312.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008672.hg.1 | ENST00000352312.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008672.hg.1 | ENST00000352312.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008672.hg.1 | ENST00000352312.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008672.hg.1 | ENST00000503905.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008672.hg.1 | ENST00000503905.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008672.hg.1 | ENST00000503905.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008672.hg.1 | ENST00000505024.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008672.hg.1 | ENST00000505024.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008672.hg.1 | ENST00000505024.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008672.hg.1 | ENST00000505024.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008672.hg.1 | ENST00000505024.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008672.hg.1 | ENST00000505024.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008672.hg.1 | ENST00000505024.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008672.hg.1 | ENST00000505024.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008672.hg.1 | ENST00000505024.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008672.hg.1 | ENST00000505024.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008672.hg.1 | ENST00000505024.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008672.hg.1 | ENST00000505024.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008672.hg.1 | ENST00000505024.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008672.hg.1 | ENST00000505024.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008672.hg.1 | ENST00000505024.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008672.hg.1 | ENST00000505024.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008672.hg.1 | ENST00000505024.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008672.hg.1 | ENST00000505024.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008672.hg.1 | ENST00000505024.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008672.hg.1 | ENST00000505024.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008672.hg.1 | ENST00000505024.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008672.hg.1 | ENST00000506204.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008672.hg.1 | ENST00000506204.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008672.hg.1 | ENST00000506204.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008672.hg.1 | ENST00000506204.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008672.hg.1 | ENST00000506204.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008672.hg.1 | ENST00000506204.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008672.hg.1 | ENST00000507830.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008672.hg.1 | ENST00000507830.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008672.hg.1 | ENST00000507830.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008672.hg.1 | ENST00000507830.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008672.hg.1 | ENST00000508140.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008672.hg.1 | ENST00000508140.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008672.hg.1 | ENST00000508140.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008672.hg.1 | ENST00000508140.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008672.hg.1 | ENST00000508140.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008672.hg.1 | ENST00000508140.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008672.hg.1 | ENST00000508140.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008672.hg.1 | ENST00000508140.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008672.hg.1 | ENST00000510833.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008672.hg.1 | ENST00000510833.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008672.hg.1 | ENST00000510833.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008672.hg.1 | ENST00000510833.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008672.hg.1 | ENST00000510833.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008672.hg.1 | ENST00000510833.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008672.hg.1 | ENST00000510833.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008672.hg.1 | ENST00000510833.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008672.hg.1 | ENST00000533201.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008672.hg.1 | ENST00000533201.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300008672.hg.1 | NM_182628.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300008672.hg.1 | NM_182628.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300008672.hg.1 | NM_182628.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300008672.hg.1 | NM_182628.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300008672.hg.1 | NM_182628.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300008672.hg.1 | NM_182628.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300008672.hg.1 | NM_182628.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300008672.hg.1 | NM_182628.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300008672.hg.1 | NM_182628.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300008672.hg.1 | NM_182628.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300008672.hg.1 | NM_182628.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300008672.hg.1 | NM_182628.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300008672.hg.1 | NM_182628.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300008672.hg.1 | NM_182628.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300008672.hg.1 | NM_182628.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300008672.hg.1 | NM_182628.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300008672.hg.1 | NM_182628.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300008672.hg.1 | NM_182628.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300008672.hg.1 | NM_182628.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300008672.hg.1 | NM_182628.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300008672.hg.1 | NM_182628.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300008672.hg.1 | NM_182628.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300008672.hg.1 | NM_182628.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300008672.hg.1 | NM_182628.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300161576.hg.1 | ENST00000352312.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300161576.hg.1 | ENST00000505024.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300161576.hg.1 | ENST00000506204.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300161576.hg.1 | NM_182628.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300161576.hg.1 | ENST00000352312.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300161576.hg.1 | ENST00000505024.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300161576.hg.1 | ENST00000506204.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300161576.hg.1 | NM_182628.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300161576.hg.1 | TC0300008672.hg.1 | Clariom D TC |
