Assay Details
Gene Symbol: PCSK6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_138319.3 | NP_612192.1 | 14 - 15 | 2352 | 77 | |
GenBank mRNA | D87994.1 | - | 14 - 15 | 2112 | 77 | |
D87995.1 | - | 14 - 15 | 2112 | 77 | ||
LN714797.1 | - | 2 - 3 | 210 | 77 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC15002009.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
