| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12018567.hg.1 |
ENST00000005386.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12018567.hg.1 |
ENST00000380650.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12018567.hg.1 |
ENST00000432584.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12018567.hg.1 |
NM_001146075.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12018567.hg.1 |
NM_001146076.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12018567.hg.1 |
NM_024604.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR12018567.hg.1 |
TC12001435.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR1200179555.hg.1 |
ENST00000005386.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200179555.hg.1 |
ENST00000380650.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200179555.hg.1 |
ENST00000432584.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200179555.hg.1 |
NM_001146075.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200179555.hg.1 |
NM_001146076.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200179555.hg.1 |
NM_024604.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200179555.hg.1 |
ENST00000005386.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200179555.hg.1 |
ENST00000380650.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200179555.hg.1 |
ENST00000432584.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1200179555.hg.1 |
NM_001146075.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200179555.hg.1 |
NM_001146076.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200179555.hg.1 |
NM_024604.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1200179555.hg.1 |
TC1200010550.hg.1 |
Clariom D TC |