Assay Details
| Gene Symbol: LVRN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173800.4 | NP_776161.3 | 19 - 20 | 2999 | 140 | |
| GenBank mRNA | AK075131.1 | - | 14 - 15 | 1534 | 140 | |
| AK291655.1 | - | 19 - 20 | 3013 | 140 | ||
| AY560010.1 | - | 19 - 20 | 2887 | 140 | ||
| BC060869.1 | - | 20 - 21 | 2926 | 30346049 | 140 | |
| BC109022.1 | - | 19 - 20 | 2863 | 40008501 | 140 | |
| BC109023.1 | - | 19 - 20 | 2863 | 40008502 | 140 | |
More Information
Set Membership: |
|

LVRN
Genomic Map