Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR12014460.hg.1 |
ENST00000280663.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12014460.hg.1 |
ENST00000382722.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12014460.hg.1 |
ENST00000456077.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12014460.hg.1 |
NM_172364.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12014460.hg.1 |
TC12001092.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1200171708.hg.1 |
ENST00000280663.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200171708.hg.1 |
ENST00000382722.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200171708.hg.1 |
ENST00000444595.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200171708.hg.1 |
ENST00000539048.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200171708.hg.1 |
ENST00000585708.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200171708.hg.1 |
ENST00000585732.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200171708.hg.1 |
ENST00000586184.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200171708.hg.1 |
ENST00000587995.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200171708.hg.1 |
ENST00000588077.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200171708.hg.1 |
NM_172364.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200171708.hg.1 |
ENST00000280663.12 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200171708.hg.1 |
ENST00000382722.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200171708.hg.1 |
ENST00000444595.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200171708.hg.1 |
ENST00000539048.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200171708.hg.1 |
ENST00000585708.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200171708.hg.1 |
ENST00000585732.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200171708.hg.1 |
ENST00000586184.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200171708.hg.1 |
ENST00000587995.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200171708.hg.1 |
ENST00000588077.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200171708.hg.1 |
NM_172364.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200171708.hg.1 |
TC1200009587.hg.1 |
Clariom D TC |