Assay Details
| Gene Symbol: PIPOX | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016518.2 | NP_057602.2 | 5 - 6 | 1126 | 60 | |
| XM_017024722.1 | XP_016880211.1 | 4 - 5 | 725 | 60 | ||
| GenBank mRNA | AF134593.1 | - | 5 - 6 | 843 | 60 | |
| AF136970.1 | - | 5 - 6 | 812 | 60 | ||
| AK022773.1 | - | 5 - 6 | 891 | 60 | ||
| AK027498.1 | - | 5 - 6 | 850 | 60 | ||
| BC008960.2 | - | 5 - 6 | 1126 | 2961289 | 60 | |
| BC027622.2 | - | 5 - 6 | 862 | 4797450 | 60 | |
| CR457155.1 | - | 5 - 6 | 800 | 60 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC17000337.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR17004142.hg.1 | ENST00000323372.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17004142.hg.1 | ENST00000484308.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17004142.hg.1 | NM_016518.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17004142.hg.1 | TC17000337.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007408.hg.1 | ENST00000323372.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007408.hg.1 | ENST00000323372.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007408.hg.1 | ENST00000323372.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007408.hg.1 | ENST00000323372.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007408.hg.1 | ENST00000323372.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007408.hg.1 | ENST00000323372.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007408.hg.1 | ENST00000323372.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007408.hg.1 | ENST00000323372.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007408.hg.1 | ENST00000323372.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007408.hg.1 | ENST00000323372.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007408.hg.1 | ENST00000323372.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007408.hg.1 | ENST00000323372.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007408.hg.1 | ENST00000323372.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007408.hg.1 | ENST00000323372.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007408.hg.1 | ENST00000323372.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007408.hg.1 | ENST00000419875.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007408.hg.1 | ENST00000419875.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007408.hg.1 | ENST00000419875.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007408.hg.1 | ENST00000419875.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007408.hg.1 | ENST00000419875.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007408.hg.1 | ENST00000466889.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007408.hg.1 | ENST00000466889.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007408.hg.1 | ENST00000466889.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007408.hg.1 | ENST00000466889.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007408.hg.1 | ENST00000466889.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007408.hg.1 | ENST00000466889.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007408.hg.1 | ENST00000466889.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007408.hg.1 | ENST00000469082.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007408.hg.1 | ENST00000469082.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007408.hg.1 | ENST00000469082.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007408.hg.1 | ENST00000469082.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007408.hg.1 | ENST00000469082.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007408.hg.1 | ENST00000469082.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007408.hg.1 | ENST00000484308.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007408.hg.1 | ENST00000484308.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007408.hg.1 | ENST00000484308.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007408.hg.1 | ENST00000484308.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007408.hg.1 | ENST00000484308.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007408.hg.1 | ENST00000484308.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007408.hg.1 | ENST00000484308.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007408.hg.1 | ENST00000484308.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007408.hg.1 | ENST00000577182.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007408.hg.1 | ENST00000577182.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007408.hg.1 | ENST00000577182.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007408.hg.1 | ENST00000577182.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007408.hg.1 | ENST00000578748.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007408.hg.1 | ENST00000578748.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007408.hg.1 | ENST00000578748.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007408.hg.1 | ENST00000580241.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007408.hg.1 | ENST00000580241.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007408.hg.1 | ENST00000580241.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007408.hg.1 | ENST00000580241.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007408.hg.1 | ENST00000580241.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007408.hg.1 | ENST00000580241.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007408.hg.1 | ENST00000580241.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007408.hg.1 | ENST00000580241.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007408.hg.1 | ENST00000580241.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007408.hg.1 | ENST00000580241.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007408.hg.1 | ENST00000580241.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007408.hg.1 | ENST00000580241.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007408.hg.1 | ENST00000580241.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007408.hg.1 | ENST00000580241.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007408.hg.1 | ENST00000580383.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007408.hg.1 | ENST00000580383.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007408.hg.1 | ENST00000580383.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007408.hg.1 | ENST00000580383.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007408.hg.1 | ENST00000580383.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007408.hg.1 | ENST00000583215.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007408.hg.1 | ENST00000583215.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007408.hg.1 | ENST00000583215.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007408.hg.1 | ENST00000583215.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007408.hg.1 | ENST00000583215.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007408.hg.1 | ENST00000583215.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007408.hg.1 | ENST00000583215.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007408.hg.1 | ENST00000583215.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007408.hg.1 | ENST00000583215.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007408.hg.1 | ENST00000583215.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007408.hg.1 | ENST00000583215.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007408.hg.1 | ENST00000583215.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007408.hg.1 | ENST00000583215.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007408.hg.1 | ENST00000583215.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007408.hg.1 | NM_016518.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007408.hg.1 | NM_016518.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007408.hg.1 | NM_016518.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007408.hg.1 | NM_016518.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007408.hg.1 | NM_016518.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007408.hg.1 | NM_016518.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007408.hg.1 | NM_016518.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007408.hg.1 | NM_016518.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007408.hg.1 | NM_016518.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007408.hg.1 | NM_016518.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007408.hg.1 | NM_016518.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007408.hg.1 | NM_016518.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007408.hg.1 | NM_016518.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007408.hg.1 | NM_016518.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007408.hg.1 | NM_016518.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700152810.hg.1 | ENST00000323372.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700152810.hg.1 | ENST00000484308.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700152810.hg.1 | ENST00000580241.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700152810.hg.1 | ENST00000583215.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700152810.hg.1 | NM_016518.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700152810.hg.1 | ENST00000323372.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700152810.hg.1 | ENST00000484308.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700152810.hg.1 | ENST00000580241.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700152810.hg.1 | ENST00000583215.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700152810.hg.1 | NM_016518.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700152810.hg.1 | TC1700007408.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1700007408.hg.1 | ENST00000323372.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007408.hg.1 | ENST00000323372.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007408.hg.1 | ENST00000323372.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007408.hg.1 | ENST00000323372.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007408.hg.1 | ENST00000323372.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007408.hg.1 | ENST00000323372.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007408.hg.1 | ENST00000323372.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007408.hg.1 | ENST00000323372.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007408.hg.1 | ENST00000323372.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007408.hg.1 | ENST00000323372.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007408.hg.1 | ENST00000323372.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007408.hg.1 | ENST00000323372.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007408.hg.1 | ENST00000323372.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007408.hg.1 | ENST00000323372.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007408.hg.1 | ENST00000323372.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007408.hg.1 | ENST00000419875.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007408.hg.1 | ENST00000419875.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007408.hg.1 | ENST00000419875.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007408.hg.1 | ENST00000419875.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007408.hg.1 | ENST00000419875.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007408.hg.1 | ENST00000466889.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007408.hg.1 | ENST00000466889.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007408.hg.1 | ENST00000466889.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007408.hg.1 | ENST00000466889.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007408.hg.1 | ENST00000466889.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007408.hg.1 | ENST00000466889.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007408.hg.1 | ENST00000466889.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007408.hg.1 | ENST00000469082.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007408.hg.1 | ENST00000469082.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007408.hg.1 | ENST00000469082.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007408.hg.1 | ENST00000469082.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007408.hg.1 | ENST00000469082.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007408.hg.1 | ENST00000469082.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007408.hg.1 | ENST00000484308.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007408.hg.1 | ENST00000484308.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007408.hg.1 | ENST00000484308.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007408.hg.1 | ENST00000484308.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007408.hg.1 | ENST00000484308.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007408.hg.1 | ENST00000484308.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007408.hg.1 | ENST00000484308.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007408.hg.1 | ENST00000484308.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007408.hg.1 | ENST00000577182.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007408.hg.1 | ENST00000577182.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007408.hg.1 | ENST00000577182.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007408.hg.1 | ENST00000577182.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007408.hg.1 | ENST00000578748.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007408.hg.1 | ENST00000578748.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007408.hg.1 | ENST00000578748.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007408.hg.1 | ENST00000580241.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007408.hg.1 | ENST00000580241.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007408.hg.1 | ENST00000580241.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007408.hg.1 | ENST00000580241.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007408.hg.1 | ENST00000580241.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007408.hg.1 | ENST00000580241.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007408.hg.1 | ENST00000580241.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007408.hg.1 | ENST00000580241.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007408.hg.1 | ENST00000580241.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007408.hg.1 | ENST00000580241.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007408.hg.1 | ENST00000580241.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007408.hg.1 | ENST00000580241.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007408.hg.1 | ENST00000580241.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007408.hg.1 | ENST00000580241.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007408.hg.1 | ENST00000580383.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007408.hg.1 | ENST00000580383.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007408.hg.1 | ENST00000580383.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007408.hg.1 | ENST00000580383.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007408.hg.1 | ENST00000580383.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007408.hg.1 | ENST00000583215.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007408.hg.1 | ENST00000583215.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007408.hg.1 | ENST00000583215.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007408.hg.1 | ENST00000583215.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007408.hg.1 | ENST00000583215.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007408.hg.1 | ENST00000583215.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007408.hg.1 | ENST00000583215.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007408.hg.1 | ENST00000583215.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007408.hg.1 | ENST00000583215.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007408.hg.1 | ENST00000583215.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007408.hg.1 | ENST00000583215.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007408.hg.1 | ENST00000583215.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007408.hg.1 | ENST00000583215.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007408.hg.1 | ENST00000583215.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007408.hg.1 | NM_016518.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007408.hg.1 | NM_016518.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007408.hg.1 | NM_016518.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007408.hg.1 | NM_016518.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007408.hg.1 | NM_016518.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007408.hg.1 | NM_016518.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007408.hg.1 | NM_016518.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007408.hg.1 | NM_016518.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007408.hg.1 | NM_016518.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007408.hg.1 | NM_016518.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007408.hg.1 | NM_016518.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007408.hg.1 | NM_016518.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007408.hg.1 | NM_016518.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007408.hg.1 | NM_016518.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700007408.hg.1 | NM_016518.2 | Refseq |

PIPOX
Genomic Map