Assay Details
| Gene Symbol: ARHGAP29 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_004815.3 | NP_004806.3 | 2 - 3 | 569 | 127 | |
| XM_011542438.2 | XP_011540740.1 | 2 - 3 | 444 | 127 | ||
| XM_011542439.2 | XP_011540741.1 | 2 - 3 | 289 | 127 | ||
| GenBank mRNA | AB208873.1 | - | 1 - 2 | 175 | 127 | |
| AK307733.1 | - | 2 - 3 | 398 | 127 | ||
| AK310491.1 | - | 2 - 3 | 289 | 127 | ||
| BC022483.1 | - | 2 - 3 | 451 | 127 | ||
| BC067839.1 | - | 2 - 3 | 564 | 127 | ||
| BC093741.1 | - | 1 - 2 | 242 | 7939586 | 127 | |
| BC093767.1 | - | 1 - 2 | 245 | 7939612 | 127 | |
| U90920.1 | - | 2 - 3 | 383 | 127 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01002884.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0100018454.hg.1 | ENST00000260526.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018454.hg.1 | ENST00000260526.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018454.hg.1 | ENST00000260526.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018454.hg.1 | ENST00000260526.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018454.hg.1 | ENST00000260526.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018454.hg.1 | ENST00000260526.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018454.hg.1 | ENST00000260526.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018454.hg.1 | ENST00000260526.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018454.hg.1 | ENST00000260526.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018454.hg.1 | ENST00000260526.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018454.hg.1 | ENST00000260526.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018454.hg.1 | ENST00000260526.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018454.hg.1 | ENST00000260526.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018454.hg.1 | ENST00000260526.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018454.hg.1 | ENST00000260526.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018454.hg.1 | ENST00000260526.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018454.hg.1 | ENST00000260526.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018454.hg.1 | ENST00000260526.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018454.hg.1 | ENST00000260526.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018454.hg.1 | ENST00000260526.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018454.hg.1 | ENST00000260526.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018454.hg.1 | ENST00000260526.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018454.hg.1 | ENST00000260526.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018454.hg.1 | ENST00000260526.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018454.hg.1 | ENST00000260526.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018454.hg.1 | ENST00000260526.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018454.hg.1 | ENST00000370217.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018454.hg.1 | ENST00000370217.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018454.hg.1 | ENST00000370217.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018454.hg.1 | ENST00000370217.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018454.hg.1 | ENST00000370217.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018454.hg.1 | ENST00000370217.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018454.hg.1 | ENST00000370217.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018454.hg.1 | ENST00000370217.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018454.hg.1 | ENST00000370217.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018454.hg.1 | ENST00000370217.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018454.hg.1 | ENST00000370217.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018454.hg.1 | ENST00000370217.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018454.hg.1 | ENST00000370217.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018454.hg.1 | ENST00000370217.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018454.hg.1 | ENST00000482481.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018454.hg.1 | ENST00000482481.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018454.hg.1 | ENST00000482481.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018454.hg.1 | ENST00000482481.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018454.hg.1 | ENST00000482481.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018454.hg.1 | ENST00000482481.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018454.hg.1 | ENST00000482481.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018454.hg.1 | ENST00000482481.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018454.hg.1 | ENST00000482481.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018454.hg.1 | ENST00000482481.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018454.hg.1 | ENST00000482481.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018454.hg.1 | ENST00000482481.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018454.hg.1 | NM_004815.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018454.hg.1 | NM_004815.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018454.hg.1 | NM_004815.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018454.hg.1 | NM_004815.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018454.hg.1 | NM_004815.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018454.hg.1 | NM_004815.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018454.hg.1 | NM_004815.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018454.hg.1 | NM_004815.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018454.hg.1 | NM_004815.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018454.hg.1 | NM_004815.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018454.hg.1 | NM_004815.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018454.hg.1 | NM_004815.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018454.hg.1 | NM_004815.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018454.hg.1 | NM_004815.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018454.hg.1 | NM_004815.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018454.hg.1 | NM_004815.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018454.hg.1 | NM_004815.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018454.hg.1 | NM_004815.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018454.hg.1 | NM_004815.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018454.hg.1 | NM_004815.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018454.hg.1 | NM_004815.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018454.hg.1 | NM_004815.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018454.hg.1 | NM_004815.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018454.hg.1 | NM_004815.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018454.hg.1 | NM_004815.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018454.hg.1 | NM_004815.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018454.hg.1 | NM_004815.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018454.hg.1 | NM_004815.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018454.hg.1 | NM_004815.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018454.hg.1 | NM_004815.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01045070.hg.1 | TC01002884.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01045070.hg.1 | ENST00000260526.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01045070.hg.1 | ENST00000370217.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01045070.hg.1 | NM_004815.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018454.hg.1 | ENST00000260526.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018454.hg.1 | ENST00000260526.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018454.hg.1 | ENST00000260526.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018454.hg.1 | ENST00000260526.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018454.hg.1 | ENST00000260526.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018454.hg.1 | ENST00000260526.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018454.hg.1 | ENST00000260526.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018454.hg.1 | ENST00000260526.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018454.hg.1 | ENST00000260526.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018454.hg.1 | ENST00000260526.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018454.hg.1 | ENST00000260526.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018454.hg.1 | ENST00000260526.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018454.hg.1 | ENST00000260526.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018454.hg.1 | ENST00000260526.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018454.hg.1 | ENST00000260526.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018454.hg.1 | ENST00000260526.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018454.hg.1 | ENST00000260526.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018454.hg.1 | ENST00000260526.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018454.hg.1 | ENST00000260526.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018454.hg.1 | ENST00000260526.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018454.hg.1 | ENST00000260526.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018454.hg.1 | ENST00000260526.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018454.hg.1 | ENST00000260526.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018454.hg.1 | ENST00000260526.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018454.hg.1 | ENST00000260526.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018454.hg.1 | ENST00000260526.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018454.hg.1 | ENST00000370217.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018454.hg.1 | ENST00000370217.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018454.hg.1 | ENST00000370217.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018454.hg.1 | ENST00000370217.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018454.hg.1 | ENST00000370217.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018454.hg.1 | ENST00000370217.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018454.hg.1 | ENST00000370217.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018454.hg.1 | ENST00000370217.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018454.hg.1 | ENST00000370217.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018454.hg.1 | ENST00000370217.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018454.hg.1 | ENST00000370217.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018454.hg.1 | ENST00000370217.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018454.hg.1 | ENST00000370217.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018454.hg.1 | ENST00000370217.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018454.hg.1 | ENST00000482481.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018454.hg.1 | ENST00000482481.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018454.hg.1 | ENST00000482481.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018454.hg.1 | ENST00000482481.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018454.hg.1 | ENST00000482481.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018454.hg.1 | ENST00000482481.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018454.hg.1 | ENST00000482481.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018454.hg.1 | ENST00000482481.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018454.hg.1 | ENST00000482481.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018454.hg.1 | ENST00000482481.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018454.hg.1 | ENST00000482481.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018454.hg.1 | ENST00000482481.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018454.hg.1 | NM_004815.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018454.hg.1 | NM_004815.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018454.hg.1 | NM_004815.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018454.hg.1 | NM_004815.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018454.hg.1 | NM_004815.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018454.hg.1 | NM_004815.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018454.hg.1 | NM_004815.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018454.hg.1 | NM_004815.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018454.hg.1 | NM_004815.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018454.hg.1 | NM_004815.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018454.hg.1 | NM_004815.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018454.hg.1 | NM_004815.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018454.hg.1 | NM_004815.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018454.hg.1 | NM_004815.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018454.hg.1 | NM_004815.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018454.hg.1 | NM_004815.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018454.hg.1 | NM_004815.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018454.hg.1 | NM_004815.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018454.hg.1 | NM_004815.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018454.hg.1 | NM_004815.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018454.hg.1 | NM_004815.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018454.hg.1 | NM_004815.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018454.hg.1 | NM_004815.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018454.hg.1 | NM_004815.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018454.hg.1 | NM_004815.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018454.hg.1 | NM_004815.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018454.hg.1 | NM_004815.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018454.hg.1 | NM_004815.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018454.hg.1 | NM_004815.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018454.hg.1 | NM_004815.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100242369.hg.1 | ENST00000260526.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100242369.hg.1 | ENST00000370217.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100242369.hg.1 | NM_004815.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100242369.hg.1 | ENST00000260526.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100242369.hg.1 | ENST00000370217.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100242369.hg.1 | NM_004815.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100242369.hg.1 | TC0100018454.hg.1 | Clariom D TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 232959 | - | - |

ARHGAP29
Genomic Map