Assay Details
| Gene Symbol: ARHGAP29 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_004815.3 | NP_004806.3 | 3 - 4 | 706 | 165 | |
| XM_006711048.3 | XP_006711111.1 | 2 - 3 | 340 | 165 | ||
| XM_011542438.2 | XP_011540740.1 | 3 - 4 | 581 | 165 | ||
| XM_011542439.2 | XP_011540741.1 | 3 - 4 | 426 | 165 | ||
| XM_011542440.2 | XP_011540742.1 | 2 - 3 | 300 | 165 | ||
| GenBank mRNA | AB208873.1 | - | 2 - 3 | 312 | 165 | |
| AK307733.1 | - | 3 - 4 | 535 | 165 | ||
| AK310491.1 | - | 3 - 4 | 426 | 165 | ||
| BC022483.1 | - | 3 - 4 | 588 | 165 | ||
| BC067839.1 | - | 3 - 4 | 701 | 165 | ||
| BC093741.1 | - | 2 - 3 | 379 | 7939586 | 165 | |
| BC093767.1 | - | 2 - 3 | 382 | 7939612 | 165 | |
| U90920.1 | - | 3 - 4 | 520 | 165 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01002884.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR01045069.hg.1 | TC01002884.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01045069.hg.1 | ENST00000260526.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01045069.hg.1 | ENST00000370217.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01045069.hg.1 | NM_004815.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018454.hg.1 | ENST00000260526.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018454.hg.1 | ENST00000260526.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018454.hg.1 | ENST00000260526.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018454.hg.1 | ENST00000260526.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018454.hg.1 | ENST00000260526.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018454.hg.1 | ENST00000260526.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018454.hg.1 | ENST00000260526.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018454.hg.1 | ENST00000260526.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018454.hg.1 | ENST00000260526.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018454.hg.1 | ENST00000260526.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018454.hg.1 | ENST00000260526.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018454.hg.1 | ENST00000260526.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018454.hg.1 | ENST00000260526.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018454.hg.1 | ENST00000260526.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018454.hg.1 | ENST00000260526.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018454.hg.1 | ENST00000260526.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018454.hg.1 | ENST00000260526.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018454.hg.1 | ENST00000260526.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018454.hg.1 | ENST00000260526.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018454.hg.1 | ENST00000260526.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018454.hg.1 | ENST00000260526.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018454.hg.1 | ENST00000260526.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018454.hg.1 | ENST00000260526.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018454.hg.1 | ENST00000260526.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018454.hg.1 | ENST00000260526.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018454.hg.1 | ENST00000260526.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018454.hg.1 | ENST00000370217.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018454.hg.1 | ENST00000370217.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018454.hg.1 | ENST00000370217.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018454.hg.1 | ENST00000370217.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018454.hg.1 | ENST00000370217.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018454.hg.1 | ENST00000370217.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018454.hg.1 | ENST00000370217.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018454.hg.1 | ENST00000370217.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018454.hg.1 | ENST00000370217.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018454.hg.1 | ENST00000370217.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018454.hg.1 | ENST00000370217.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018454.hg.1 | ENST00000370217.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018454.hg.1 | ENST00000370217.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018454.hg.1 | ENST00000370217.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018454.hg.1 | ENST00000482481.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018454.hg.1 | ENST00000482481.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018454.hg.1 | ENST00000482481.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018454.hg.1 | ENST00000482481.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018454.hg.1 | ENST00000482481.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018454.hg.1 | ENST00000482481.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018454.hg.1 | ENST00000482481.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018454.hg.1 | ENST00000482481.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018454.hg.1 | ENST00000482481.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018454.hg.1 | ENST00000482481.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018454.hg.1 | ENST00000482481.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018454.hg.1 | ENST00000482481.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018454.hg.1 | NM_004815.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018454.hg.1 | NM_004815.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018454.hg.1 | NM_004815.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018454.hg.1 | NM_004815.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018454.hg.1 | NM_004815.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018454.hg.1 | NM_004815.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018454.hg.1 | NM_004815.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018454.hg.1 | NM_004815.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018454.hg.1 | NM_004815.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018454.hg.1 | NM_004815.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018454.hg.1 | NM_004815.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018454.hg.1 | NM_004815.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018454.hg.1 | NM_004815.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018454.hg.1 | NM_004815.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018454.hg.1 | NM_004815.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018454.hg.1 | NM_004815.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018454.hg.1 | NM_004815.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018454.hg.1 | NM_004815.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018454.hg.1 | NM_004815.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018454.hg.1 | NM_004815.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018454.hg.1 | NM_004815.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018454.hg.1 | NM_004815.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018454.hg.1 | NM_004815.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018454.hg.1 | NM_004815.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018454.hg.1 | NM_004815.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018454.hg.1 | NM_004815.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018454.hg.1 | NM_004815.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018454.hg.1 | NM_004815.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018454.hg.1 | NM_004815.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018454.hg.1 | NM_004815.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100242367.hg.1 | ENST00000260526.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100242367.hg.1 | ENST00000370217.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100242367.hg.1 | NM_004815.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100242367.hg.1 | ENST00000260526.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100242367.hg.1 | ENST00000370217.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100242367.hg.1 | NM_004815.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100242367.hg.1 | TC0100018454.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0100018454.hg.1 | ENST00000260526.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018454.hg.1 | ENST00000260526.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018454.hg.1 | ENST00000260526.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018454.hg.1 | ENST00000260526.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018454.hg.1 | ENST00000260526.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018454.hg.1 | ENST00000260526.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018454.hg.1 | ENST00000260526.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018454.hg.1 | ENST00000260526.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018454.hg.1 | ENST00000260526.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018454.hg.1 | ENST00000260526.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018454.hg.1 | ENST00000260526.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018454.hg.1 | ENST00000260526.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018454.hg.1 | ENST00000260526.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018454.hg.1 | ENST00000260526.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018454.hg.1 | ENST00000260526.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018454.hg.1 | ENST00000260526.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018454.hg.1 | ENST00000260526.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018454.hg.1 | ENST00000260526.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018454.hg.1 | ENST00000260526.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018454.hg.1 | ENST00000260526.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018454.hg.1 | ENST00000260526.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018454.hg.1 | ENST00000260526.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018454.hg.1 | ENST00000260526.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018454.hg.1 | ENST00000260526.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018454.hg.1 | ENST00000260526.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018454.hg.1 | ENST00000260526.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018454.hg.1 | ENST00000370217.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018454.hg.1 | ENST00000370217.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018454.hg.1 | ENST00000370217.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018454.hg.1 | ENST00000370217.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018454.hg.1 | ENST00000370217.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018454.hg.1 | ENST00000370217.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018454.hg.1 | ENST00000370217.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018454.hg.1 | ENST00000370217.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018454.hg.1 | ENST00000370217.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018454.hg.1 | ENST00000370217.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018454.hg.1 | ENST00000370217.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018454.hg.1 | ENST00000370217.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018454.hg.1 | ENST00000370217.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018454.hg.1 | ENST00000370217.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018454.hg.1 | ENST00000482481.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018454.hg.1 | ENST00000482481.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018454.hg.1 | ENST00000482481.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018454.hg.1 | ENST00000482481.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018454.hg.1 | ENST00000482481.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018454.hg.1 | ENST00000482481.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018454.hg.1 | ENST00000482481.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018454.hg.1 | ENST00000482481.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018454.hg.1 | ENST00000482481.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018454.hg.1 | ENST00000482481.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018454.hg.1 | ENST00000482481.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018454.hg.1 | ENST00000482481.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018454.hg.1 | NM_004815.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018454.hg.1 | NM_004815.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018454.hg.1 | NM_004815.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018454.hg.1 | NM_004815.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018454.hg.1 | NM_004815.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018454.hg.1 | NM_004815.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018454.hg.1 | NM_004815.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018454.hg.1 | NM_004815.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018454.hg.1 | NM_004815.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018454.hg.1 | NM_004815.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018454.hg.1 | NM_004815.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018454.hg.1 | NM_004815.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018454.hg.1 | NM_004815.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018454.hg.1 | NM_004815.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018454.hg.1 | NM_004815.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018454.hg.1 | NM_004815.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018454.hg.1 | NM_004815.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018454.hg.1 | NM_004815.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018454.hg.1 | NM_004815.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018454.hg.1 | NM_004815.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018454.hg.1 | NM_004815.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018454.hg.1 | NM_004815.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018454.hg.1 | NM_004815.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018454.hg.1 | NM_004815.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018454.hg.1 | NM_004815.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018454.hg.1 | NM_004815.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018454.hg.1 | NM_004815.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018454.hg.1 | NM_004815.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018454.hg.1 | NM_004815.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018454.hg.1 | NM_004815.3 | Refseq |

ARHGAP29
Genomic Map