Assay Details
Gene Symbol: PTH2R | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001309516.1 | NP_001296445.1 | 12 - 13 | 1575 | 86 | |
NM_005048.3 | NP_005039.1 | 12 - 13 | 1471 | 86 | ||
GenBank mRNA | AK294181.1 | - | 12 - 13 | 1575 | 86 | |
AY497546.1 | - | 12 - 13 | 1257 | 86 | ||
BC036811.1 | - | 12 - 13 | 1470 | 5734330 | 86 | |
U25128.1 | - | 12 - 13 | 1399 | 86 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC02001236.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR02018964.hg.1 | TC02001236.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR02018964.hg.1 | ENST00000272847.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR02018964.hg.1 | ENST00000413482.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR02018964.hg.1 | NM_005048.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200010627.hg.1 | ENST00000272847.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010627.hg.1 | ENST00000272847.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010627.hg.1 | ENST00000272847.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010627.hg.1 | ENST00000272847.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010627.hg.1 | ENST00000272847.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010627.hg.1 | ENST00000272847.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010627.hg.1 | ENST00000272847.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010627.hg.1 | ENST00000272847.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010627.hg.1 | ENST00000272847.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010627.hg.1 | ENST00000272847.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010627.hg.1 | ENST00000272847.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010627.hg.1 | ENST00000272847.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010627.hg.1 | ENST00000272847.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010627.hg.1 | ENST00000272847.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010627.hg.1 | ENST00000272847.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010627.hg.1 | ENST00000272847.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010627.hg.1 | ENST00000272847.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010627.hg.1 | ENST00000419079.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010627.hg.1 | ENST00000419079.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010627.hg.1 | ENST00000419079.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010627.hg.1 | ENST00000419079.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010627.hg.1 | ENST00000419079.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010627.hg.1 | ENST00000419079.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010627.hg.1 | ENST00000419079.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010627.hg.1 | ENST00000487472.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010627.hg.1 | ENST00000487472.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010627.hg.1 | ENST00000487472.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010627.hg.1 | ENST00000487472.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010627.hg.1 | ENST00000487472.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010627.hg.1 | ENST00000487472.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010627.hg.1 | ENST00000617735.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010627.hg.1 | ENST00000617735.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010627.hg.1 | ENST00000617735.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010627.hg.1 | ENST00000617735.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010627.hg.1 | ENST00000617735.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010627.hg.1 | ENST00000617735.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010627.hg.1 | ENST00000617735.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010627.hg.1 | ENST00000617735.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010627.hg.1 | ENST00000617735.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010627.hg.1 | ENST00000617735.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010627.hg.1 | ENST00000617735.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010627.hg.1 | ENST00000617735.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010627.hg.1 | ENST00000617735.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010627.hg.1 | ENST00000617735.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010627.hg.1 | ENST00000617735.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010627.hg.1 | ENST00000617735.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200010627.hg.1 | NM_001309516.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200010627.hg.1 | NM_001309516.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200010627.hg.1 | NM_001309516.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200010627.hg.1 | NM_001309516.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200010627.hg.1 | NM_001309516.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200010627.hg.1 | NM_001309516.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200010627.hg.1 | NM_001309516.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200010627.hg.1 | NM_001309516.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200010627.hg.1 | NM_001309516.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200010627.hg.1 | NM_001309516.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200010627.hg.1 | NM_001309516.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200010627.hg.1 | NM_001309516.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200010627.hg.1 | NM_001309516.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200010627.hg.1 | NM_001309516.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200010627.hg.1 | NM_001309516.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200010627.hg.1 | NM_001309516.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200010627.hg.1 | NM_005048.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200010627.hg.1 | NM_005048.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200010627.hg.1 | NM_005048.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200010627.hg.1 | NM_005048.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200010627.hg.1 | NM_005048.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200010627.hg.1 | NM_005048.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200010627.hg.1 | NM_005048.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200010627.hg.1 | NM_005048.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200010627.hg.1 | NM_005048.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200010627.hg.1 | NM_005048.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200010627.hg.1 | NM_005048.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200010627.hg.1 | NM_005048.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200010627.hg.1 | NM_005048.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200010627.hg.1 | NM_005048.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200010627.hg.1 | NM_005048.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200010627.hg.1 | NM_005048.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200010627.hg.1 | NM_005048.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200010627.hg.1 | ENST00000272847.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200010627.hg.1 | ENST00000272847.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200010627.hg.1 | ENST00000272847.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200010627.hg.1 | ENST00000272847.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200010627.hg.1 | ENST00000272847.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200010627.hg.1 | ENST00000272847.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200010627.hg.1 | ENST00000272847.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200010627.hg.1 | ENST00000272847.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200010627.hg.1 | ENST00000272847.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200010627.hg.1 | ENST00000272847.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200010627.hg.1 | ENST00000272847.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200010627.hg.1 | ENST00000272847.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200010627.hg.1 | ENST00000272847.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200010627.hg.1 | ENST00000272847.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200010627.hg.1 | ENST00000272847.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200010627.hg.1 | ENST00000272847.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200010627.hg.1 | ENST00000272847.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200010627.hg.1 | ENST00000419079.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200010627.hg.1 | ENST00000419079.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200010627.hg.1 | ENST00000419079.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200010627.hg.1 | ENST00000419079.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200010627.hg.1 | ENST00000419079.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200010627.hg.1 | ENST00000419079.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200010627.hg.1 | ENST00000419079.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200010627.hg.1 | ENST00000487472.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200010627.hg.1 | ENST00000487472.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200010627.hg.1 | ENST00000487472.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200010627.hg.1 | ENST00000487472.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200010627.hg.1 | ENST00000487472.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200010627.hg.1 | ENST00000487472.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200010627.hg.1 | ENST00000617735.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200010627.hg.1 | ENST00000617735.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200010627.hg.1 | ENST00000617735.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200010627.hg.1 | ENST00000617735.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200010627.hg.1 | ENST00000617735.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200010627.hg.1 | ENST00000617735.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200010627.hg.1 | ENST00000617735.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200010627.hg.1 | ENST00000617735.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200010627.hg.1 | ENST00000617735.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200010627.hg.1 | ENST00000617735.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200010627.hg.1 | ENST00000617735.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200010627.hg.1 | ENST00000617735.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200010627.hg.1 | ENST00000617735.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200010627.hg.1 | ENST00000617735.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200010627.hg.1 | ENST00000617735.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200010627.hg.1 | ENST00000617735.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200010627.hg.1 | NM_001309516.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200010627.hg.1 | NM_001309516.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200010627.hg.1 | NM_001309516.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200010627.hg.1 | NM_001309516.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200010627.hg.1 | NM_001309516.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200010627.hg.1 | NM_001309516.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200010627.hg.1 | NM_001309516.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200010627.hg.1 | NM_001309516.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200010627.hg.1 | NM_001309516.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200010627.hg.1 | NM_001309516.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200010627.hg.1 | NM_001309516.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200010627.hg.1 | NM_001309516.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200010627.hg.1 | NM_001309516.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200010627.hg.1 | NM_001309516.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200010627.hg.1 | NM_001309516.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200010627.hg.1 | NM_001309516.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200010627.hg.1 | NM_005048.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200010627.hg.1 | NM_005048.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200010627.hg.1 | NM_005048.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200010627.hg.1 | NM_005048.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200010627.hg.1 | NM_005048.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200010627.hg.1 | NM_005048.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200010627.hg.1 | NM_005048.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200010627.hg.1 | NM_005048.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200010627.hg.1 | NM_005048.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200010627.hg.1 | NM_005048.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200010627.hg.1 | NM_005048.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200010627.hg.1 | NM_005048.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200010627.hg.1 | NM_005048.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200010627.hg.1 | NM_005048.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200010627.hg.1 | NM_005048.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200010627.hg.1 | NM_005048.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0200010627.hg.1 | NM_005048.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200175330.hg.1 | ENST00000272847.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200175330.hg.1 | ENST00000617735.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200175330.hg.1 | NM_001309516.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200175330.hg.1 | NM_005048.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200175330.hg.1 | ENST00000272847.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200175330.hg.1 | ENST00000617735.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200175330.hg.1 | NM_001309516.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200175330.hg.1 | NM_005048.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200175330.hg.1 | TC0200010627.hg.1 | Clariom D TC |
