Assay Details
| Gene Symbol: POLE4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019896.2 | NP_063949.2 | 1 - 2 | 246 | 77 | |
| GenBank mRNA | AI815728.1 | - | 1 - 2 | 273 | 77 | |
| BC031331.1 | - | 1 - 2 | 241 | 5443810 | 77 | |
| BU597500.1 | - | 253 | 77 | |||
| CA454961.1 | - | 1 - 2 | 188 | 77 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC02000470.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR02007385.hg.1 | TC02000470.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02007385.hg.1 | ENST00000459636.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02007385.hg.1 | ENST00000483063.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02007385.hg.1 | NM_019896.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02007383.hg.1 | TC02000470.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02007383.hg.1 | ENST00000483063.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02007383.hg.1 | NM_019896.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02007386.hg.1 | TC02000470.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02007386.hg.1 | ENST00000459636.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02007386.hg.1 | ENST00000483063.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02007386.hg.1 | NM_019896.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02007389.hg.1 | TC02000470.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02007389.hg.1 | ENST00000459636.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02007389.hg.1 | ENST00000483063.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02007389.hg.1 | NM_019896.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008103.hg.1 | ENST00000233699.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008103.hg.1 | ENST00000233699.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008103.hg.1 | ENST00000233699.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008103.hg.1 | ENST00000233699.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008103.hg.1 | ENST00000233699.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008103.hg.1 | ENST00000459636.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008103.hg.1 | ENST00000459636.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008103.hg.1 | ENST00000459636.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008103.hg.1 | ENST00000459636.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008103.hg.1 | ENST00000459636.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008103.hg.1 | ENST00000465242.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008103.hg.1 | ENST00000465242.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008103.hg.1 | ENST00000465242.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008103.hg.1 | ENST00000465242.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008103.hg.1 | ENST00000473023.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008103.hg.1 | ENST00000473023.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008103.hg.1 | ENST00000473023.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008103.hg.1 | ENST00000483063.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008103.hg.1 | ENST00000483063.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008103.hg.1 | ENST00000483063.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008103.hg.1 | ENST00000483063.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008103.hg.1 | ENST00000483063.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008103.hg.1 | ENST00000483063.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008103.hg.1 | ENST00000483063.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008103.hg.1 | ENST00000483063.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008103.hg.1 | ENST00000483063.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008103.hg.1 | ENST00000485527.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008103.hg.1 | ENST00000485527.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008103.hg.1 | ENST00000485527.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008103.hg.1 | ENST00000485527.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008103.hg.1 | ENST00000485527.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008103.hg.1 | NM_019896.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008103.hg.1 | NM_019896.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008103.hg.1 | NM_019896.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008103.hg.1 | NM_019896.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008103.hg.1 | NM_019896.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008103.hg.1 | NM_019896.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008103.hg.1 | NM_019896.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200156358.hg.1 | ENST00000459636.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200156358.hg.1 | ENST00000483063.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200156358.hg.1 | ENST00000485527.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200156358.hg.1 | NM_019896.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200156358.hg.1 | ENST00000459636.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200156358.hg.1 | ENST00000483063.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200156358.hg.1 | ENST00000485527.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200156358.hg.1 | NM_019896.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200156358.hg.1 | TC0200008103.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0200008103.hg.1 | ENST00000233699.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008103.hg.1 | ENST00000233699.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008103.hg.1 | ENST00000233699.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008103.hg.1 | ENST00000233699.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008103.hg.1 | ENST00000233699.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008103.hg.1 | ENST00000459636.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008103.hg.1 | ENST00000459636.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008103.hg.1 | ENST00000459636.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008103.hg.1 | ENST00000459636.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008103.hg.1 | ENST00000459636.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008103.hg.1 | ENST00000465242.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008103.hg.1 | ENST00000465242.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008103.hg.1 | ENST00000465242.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008103.hg.1 | ENST00000465242.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008103.hg.1 | ENST00000473023.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008103.hg.1 | ENST00000473023.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008103.hg.1 | ENST00000473023.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008103.hg.1 | ENST00000483063.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008103.hg.1 | ENST00000483063.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008103.hg.1 | ENST00000483063.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008103.hg.1 | ENST00000483063.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008103.hg.1 | ENST00000483063.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008103.hg.1 | ENST00000483063.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008103.hg.1 | ENST00000483063.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008103.hg.1 | ENST00000483063.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008103.hg.1 | ENST00000483063.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008103.hg.1 | ENST00000485527.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008103.hg.1 | ENST00000485527.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008103.hg.1 | ENST00000485527.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008103.hg.1 | ENST00000485527.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008103.hg.1 | ENST00000485527.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008103.hg.1 | NM_019896.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008103.hg.1 | NM_019896.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008103.hg.1 | NM_019896.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008103.hg.1 | NM_019896.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008103.hg.1 | NM_019896.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008103.hg.1 | NM_019896.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008103.hg.1 | NM_019896.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200156356.hg.1 | ENST00000483063.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200156356.hg.1 | NM_019896.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200156356.hg.1 | ENST00000483063.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200156356.hg.1 | NM_019896.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200156356.hg.1 | TC0200008103.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0200156359.hg.1 | ENST00000233699.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200156359.hg.1 | ENST00000459636.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200156359.hg.1 | ENST00000483063.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200156359.hg.1 | ENST00000485527.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200156359.hg.1 | NM_019896.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200156359.hg.1 | ENST00000233699.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200156359.hg.1 | ENST00000459636.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200156359.hg.1 | ENST00000483063.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200156359.hg.1 | ENST00000485527.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200156359.hg.1 | NM_019896.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200156359.hg.1 | TC0200008103.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0200156362.hg.1 | ENST00000233699.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200156362.hg.1 | ENST00000459636.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200156362.hg.1 | ENST00000465242.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200156362.hg.1 | ENST00000473023.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200156362.hg.1 | ENST00000483063.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200156362.hg.1 | ENST00000485527.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200156362.hg.1 | NM_019896.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200156362.hg.1 | ENST00000233699.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200156362.hg.1 | ENST00000459636.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200156362.hg.1 | ENST00000465242.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200156362.hg.1 | ENST00000473023.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200156362.hg.1 | ENST00000483063.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200156362.hg.1 | ENST00000485527.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200156362.hg.1 | NM_019896.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200156362.hg.1 | TC0200008103.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0200156355.hg.1 | ENST00000483063.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200156355.hg.1 | NM_019896.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200156355.hg.1 | ENST00000483063.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200156355.hg.1 | NM_019896.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200156355.hg.1 | TC0200008103.hg.1 | Clariom D TC |

POLE4
Genomic Map