Assay Details
Gene Symbol: GEMIN8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001042479.1 | NP_001035944.1 | 2 - 3 | 220 | 72 | |
XM_005274555.2 | XP_005274612.1 | 1 - 2 | 3173 | 72 | ||
GenBank mRNA | AK125062.1 | - | 1 - 2 | 2265 | 72 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001042479.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0X00009107.hg.1 | ENST00000332885.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009107.hg.1 | ENST00000332885.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009107.hg.1 | ENST00000332885.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009107.hg.1 | ENST00000332885.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009107.hg.1 | ENST00000332885.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009107.hg.1 | ENST00000380523.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009107.hg.1 | ENST00000380523.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009107.hg.1 | ENST00000380523.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009107.hg.1 | ENST00000380523.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009107.hg.1 | ENST00000380523.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009107.hg.1 | ENST00000380523.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009107.hg.1 | ENST00000380523.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009107.hg.1 | ENST00000380523.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009107.hg.1 | ENST00000380523.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009107.hg.1 | ENST00000380523.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009107.hg.1 | ENST00000398355.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009107.hg.1 | ENST00000398355.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009107.hg.1 | ENST00000398355.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009107.hg.1 | ENST00000398355.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009107.hg.1 | ENST00000398355.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009107.hg.1 | ENST00000398355.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009107.hg.1 | ENST00000398355.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009107.hg.1 | ENST00000460203.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009107.hg.1 | ENST00000460203.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009107.hg.1 | ENST00000460203.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009107.hg.1 | ENST00000477386.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009107.hg.1 | ENST00000477386.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009107.hg.1 | ENST00000477386.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009107.hg.1 | ENST00000477386.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009107.hg.1 | NM_001042479.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009107.hg.1 | NM_001042479.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009107.hg.1 | NM_001042479.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009107.hg.1 | NM_001042479.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009107.hg.1 | NM_001042479.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009107.hg.1 | NM_001042479.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009107.hg.1 | NM_001042479.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009107.hg.1 | NM_001042479.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009107.hg.1 | NM_001042479.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009107.hg.1 | NM_001042479.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009107.hg.1 | NM_001042480.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009107.hg.1 | NM_001042480.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009107.hg.1 | NM_001042480.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009107.hg.1 | NM_001042480.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009107.hg.1 | NM_001042480.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009107.hg.1 | NM_001042480.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009107.hg.1 | NM_001042480.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009107.hg.1 | NM_001042480.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009107.hg.1 | NM_001042480.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009107.hg.1 | NM_017856.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009107.hg.1 | NM_017856.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009107.hg.1 | NM_017856.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009107.hg.1 | NM_017856.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009107.hg.1 | NM_017856.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009107.hg.1 | NM_017856.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009107.hg.1 | NM_017856.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009107.hg.1 | NM_017856.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009107.hg.1 | NM_017856.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009107.hg.1 | NM_017856.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00009107.hg.1 | NM_017856.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00009107.hg.1 | ENST00000332885.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009107.hg.1 | ENST00000332885.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009107.hg.1 | ENST00000332885.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009107.hg.1 | ENST00000332885.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009107.hg.1 | ENST00000332885.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009107.hg.1 | ENST00000380523.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009107.hg.1 | ENST00000380523.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009107.hg.1 | ENST00000380523.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009107.hg.1 | ENST00000380523.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009107.hg.1 | ENST00000380523.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009107.hg.1 | ENST00000380523.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009107.hg.1 | ENST00000380523.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009107.hg.1 | ENST00000380523.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009107.hg.1 | ENST00000380523.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009107.hg.1 | ENST00000380523.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009107.hg.1 | ENST00000398355.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009107.hg.1 | ENST00000398355.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009107.hg.1 | ENST00000398355.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009107.hg.1 | ENST00000398355.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009107.hg.1 | ENST00000398355.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009107.hg.1 | ENST00000398355.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009107.hg.1 | ENST00000398355.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009107.hg.1 | ENST00000460203.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009107.hg.1 | ENST00000460203.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009107.hg.1 | ENST00000460203.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009107.hg.1 | ENST00000477386.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009107.hg.1 | ENST00000477386.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009107.hg.1 | ENST00000477386.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009107.hg.1 | ENST00000477386.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00009107.hg.1 | NM_001042479.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00009107.hg.1 | NM_001042479.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00009107.hg.1 | NM_001042479.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00009107.hg.1 | NM_001042479.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00009107.hg.1 | NM_001042479.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00009107.hg.1 | NM_001042479.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00009107.hg.1 | NM_001042479.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00009107.hg.1 | NM_001042479.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00009107.hg.1 | NM_001042479.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00009107.hg.1 | NM_001042479.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00009107.hg.1 | NM_001042480.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00009107.hg.1 | NM_001042480.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00009107.hg.1 | NM_001042480.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00009107.hg.1 | NM_001042480.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00009107.hg.1 | NM_001042480.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00009107.hg.1 | NM_001042480.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00009107.hg.1 | NM_001042480.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00009107.hg.1 | NM_001042480.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00009107.hg.1 | NM_001042480.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00009107.hg.1 | NM_017856.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00009107.hg.1 | NM_017856.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00009107.hg.1 | NM_017856.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00009107.hg.1 | NM_017856.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00009107.hg.1 | NM_017856.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00009107.hg.1 | NM_017856.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00009107.hg.1 | NM_017856.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00009107.hg.1 | NM_017856.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00009107.hg.1 | NM_017856.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00009107.hg.1 | NM_017856.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00009107.hg.1 | NM_017856.2 | Refseq |
