Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR11015303.hg.1 |
ENST00000339772.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11015303.hg.1 |
ENST00000389881.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11015303.hg.1 |
ENST00000529077.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11015303.hg.1 |
ENST00000535456.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11015303.hg.1 |
NM_138342.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11015303.hg.1 |
TC11001217.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1100172284.hg.1 |
ENST00000339772.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100172284.hg.1 |
ENST00000529077.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100172284.hg.1 |
ENST00000535456.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100172284.hg.1 |
NM_138342.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100172284.hg.1 |
ENST00000339772.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100172284.hg.1 |
ENST00000529077.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100172284.hg.1 |
ENST00000535456.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100172284.hg.1 |
NM_138342.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100172284.hg.1 |
TC1100009618.hg.1 |
Clariom D TC |