Assay Details
| Gene Symbol: FYB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001243093.1 | NP_001230022.1 | 13 - 14 | 2188 | 78 | |
| NM_001465.4 | NP_001456.3 | 13 - 14 | 2235 | 78 | ||
| XM_006714464.2 | XP_006714527.1 | 13 - 14 | 2220 | 78 | ||
| XM_006714465.2 | XP_006714528.1 | 14 - 15 | 2573 | 78 | ||
| XM_006714466.2 | XP_006714529.1 | 13 - 14 | 2349 | 78 | ||
| XM_011514008.2 | XP_011512310.1 | 15 - 16 | 2673 | 78 | ||
| XM_011514009.1 | XP_011512311.1 | 15 - 16 | 2594 | 78 | ||
| XM_011514010.1 | XP_011512312.1 | 14 - 15 | 2312 | 78 | ||
| GenBank mRNA | AF198052.1 | - | 13 - 14 | 2072 | 78 | |
| AK290403.1 | - | 13 - 14 | 2230 | 78 | ||
| AK297077.1 | - | 13 - 14 | 2188 | 78 | ||
| BC015933.1 | - | 13 - 14 | 2164 | 78 | ||
| BC117449.1 | - | 13 - 14 | 2134 | 40126000 | 78 | |
| BC143645.1 | - | 13 - 14 | 2134 | 9052159 | 78 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC05001295.hg.1
Set Membership: |
|

FYB
Genomic Map