Assay Details
Gene Symbol: THSD7B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001316349.1 | NP_001303278.1 | 16 - 17 | 3434 | 64 | |
XM_017005049.1 | XP_016860538.1 | 10 - 11 | 1911 | 64 | ||
GenBank mRNA | AB051466.1 | - | 15 - 16 | 3064 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0200009464.hg.1 | ENST00000272643.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009464.hg.1 | ENST00000272643.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009464.hg.1 | ENST00000272643.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009464.hg.1 | ENST00000272643.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009464.hg.1 | ENST00000272643.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009464.hg.1 | ENST00000272643.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009464.hg.1 | ENST00000272643.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009464.hg.1 | ENST00000272643.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009464.hg.1 | ENST00000272643.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009464.hg.1 | ENST00000272643.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009464.hg.1 | ENST00000272643.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009464.hg.1 | ENST00000272643.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009464.hg.1 | ENST00000272643.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009464.hg.1 | ENST00000272643.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009464.hg.1 | ENST00000272643.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009464.hg.1 | ENST00000272643.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009464.hg.1 | ENST00000272643.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009464.hg.1 | ENST00000272643.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009464.hg.1 | ENST00000272643.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009464.hg.1 | ENST00000272643.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009464.hg.1 | ENST00000272643.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009464.hg.1 | ENST00000272643.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009464.hg.1 | ENST00000272643.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009464.hg.1 | ENST00000272643.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009464.hg.1 | ENST00000272643.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009464.hg.1 | ENST00000272643.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009464.hg.1 | ENST00000272643.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009464.hg.1 | ENST00000272643.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009464.hg.1 | ENST00000409968.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009464.hg.1 | ENST00000409968.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009464.hg.1 | ENST00000409968.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009464.hg.1 | ENST00000409968.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009464.hg.1 | ENST00000409968.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009464.hg.1 | ENST00000409968.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009464.hg.1 | ENST00000409968.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009464.hg.1 | ENST00000409968.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009464.hg.1 | ENST00000409968.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009464.hg.1 | ENST00000409968.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009464.hg.1 | ENST00000409968.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009464.hg.1 | ENST00000409968.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009464.hg.1 | ENST00000409968.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009464.hg.1 | ENST00000409968.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009464.hg.1 | ENST00000409968.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009464.hg.1 | ENST00000409968.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009464.hg.1 | ENST00000409968.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009464.hg.1 | ENST00000409968.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009464.hg.1 | ENST00000409968.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009464.hg.1 | ENST00000409968.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009464.hg.1 | ENST00000409968.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009464.hg.1 | ENST00000409968.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009464.hg.1 | ENST00000409968.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009464.hg.1 | ENST00000409968.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009464.hg.1 | ENST00000409968.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009464.hg.1 | ENST00000409968.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009464.hg.1 | ENST00000409968.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009464.hg.1 | ENST00000409968.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009464.hg.1 | ENST00000409968.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009464.hg.1 | ENST00000409968.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009464.hg.1 | ENST00000413152.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009464.hg.1 | ENST00000413152.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009464.hg.1 | ENST00000413152.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009464.hg.1 | ENST00000413152.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009464.hg.1 | ENST00000413152.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009464.hg.1 | ENST00000413152.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009464.hg.1 | ENST00000413152.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009464.hg.1 | ENST00000413152.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009464.hg.1 | ENST00000413152.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009464.hg.1 | ENST00000413152.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009464.hg.1 | ENST00000413152.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009464.hg.1 | ENST00000413152.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009464.hg.1 | ENST00000413152.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009464.hg.1 | ENST00000413152.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009464.hg.1 | ENST00000413152.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009464.hg.1 | ENST00000413152.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009464.hg.1 | ENST00000413152.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009464.hg.1 | ENST00000413152.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009464.hg.1 | ENST00000413152.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009464.hg.1 | ENST00000413152.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009464.hg.1 | ENST00000413152.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009464.hg.1 | ENST00000413152.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009464.hg.1 | ENST00000413152.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009464.hg.1 | ENST00000413152.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009464.hg.1 | ENST00000413152.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009464.hg.1 | ENST00000413152.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009464.hg.1 | ENST00000413152.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009464.hg.1 | ENST00000413152.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009464.hg.1 | ENST00000472720.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009464.hg.1 | ENST00000472720.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009464.hg.1 | ENST00000472720.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009464.hg.1 | ENST00000472720.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009464.hg.1 | ENST00000472720.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009464.hg.1 | ENST00000480352.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009464.hg.1 | ENST00000480352.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009464.hg.1 | ENST00000480352.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009464.hg.1 | ENST00000485379.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009464.hg.1 | ENST00000485379.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200009464.hg.1 | ENST00000485379.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009464.hg.1 | ENST00000272643.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009464.hg.1 | ENST00000272643.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009464.hg.1 | ENST00000272643.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009464.hg.1 | ENST00000272643.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009464.hg.1 | ENST00000272643.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009464.hg.1 | ENST00000272643.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009464.hg.1 | ENST00000272643.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009464.hg.1 | ENST00000272643.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009464.hg.1 | ENST00000272643.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009464.hg.1 | ENST00000272643.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009464.hg.1 | ENST00000272643.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009464.hg.1 | ENST00000272643.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009464.hg.1 | ENST00000272643.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009464.hg.1 | ENST00000272643.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009464.hg.1 | ENST00000272643.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009464.hg.1 | ENST00000272643.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009464.hg.1 | ENST00000272643.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009464.hg.1 | ENST00000272643.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009464.hg.1 | ENST00000272643.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009464.hg.1 | ENST00000272643.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009464.hg.1 | ENST00000272643.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009464.hg.1 | ENST00000272643.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009464.hg.1 | ENST00000272643.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009464.hg.1 | ENST00000272643.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009464.hg.1 | ENST00000272643.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009464.hg.1 | ENST00000272643.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009464.hg.1 | ENST00000272643.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009464.hg.1 | ENST00000272643.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009464.hg.1 | ENST00000409968.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009464.hg.1 | ENST00000409968.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009464.hg.1 | ENST00000409968.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009464.hg.1 | ENST00000409968.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009464.hg.1 | ENST00000409968.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009464.hg.1 | ENST00000409968.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009464.hg.1 | ENST00000409968.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009464.hg.1 | ENST00000409968.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009464.hg.1 | ENST00000409968.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009464.hg.1 | ENST00000409968.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009464.hg.1 | ENST00000409968.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009464.hg.1 | ENST00000409968.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009464.hg.1 | ENST00000409968.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009464.hg.1 | ENST00000409968.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009464.hg.1 | ENST00000409968.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009464.hg.1 | ENST00000409968.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009464.hg.1 | ENST00000409968.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009464.hg.1 | ENST00000409968.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009464.hg.1 | ENST00000409968.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009464.hg.1 | ENST00000409968.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009464.hg.1 | ENST00000409968.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009464.hg.1 | ENST00000409968.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009464.hg.1 | ENST00000409968.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009464.hg.1 | ENST00000409968.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009464.hg.1 | ENST00000409968.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009464.hg.1 | ENST00000409968.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009464.hg.1 | ENST00000409968.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009464.hg.1 | ENST00000409968.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009464.hg.1 | ENST00000409968.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009464.hg.1 | ENST00000409968.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009464.hg.1 | ENST00000413152.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009464.hg.1 | ENST00000413152.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009464.hg.1 | ENST00000413152.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009464.hg.1 | ENST00000413152.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009464.hg.1 | ENST00000413152.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009464.hg.1 | ENST00000413152.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009464.hg.1 | ENST00000413152.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009464.hg.1 | ENST00000413152.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009464.hg.1 | ENST00000413152.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009464.hg.1 | ENST00000413152.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009464.hg.1 | ENST00000413152.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009464.hg.1 | ENST00000413152.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009464.hg.1 | ENST00000413152.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009464.hg.1 | ENST00000413152.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009464.hg.1 | ENST00000413152.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009464.hg.1 | ENST00000413152.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009464.hg.1 | ENST00000413152.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009464.hg.1 | ENST00000413152.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009464.hg.1 | ENST00000413152.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009464.hg.1 | ENST00000413152.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009464.hg.1 | ENST00000413152.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009464.hg.1 | ENST00000413152.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009464.hg.1 | ENST00000413152.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009464.hg.1 | ENST00000413152.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009464.hg.1 | ENST00000413152.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009464.hg.1 | ENST00000413152.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009464.hg.1 | ENST00000413152.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009464.hg.1 | ENST00000413152.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009464.hg.1 | ENST00000472720.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009464.hg.1 | ENST00000472720.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009464.hg.1 | ENST00000472720.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009464.hg.1 | ENST00000472720.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009464.hg.1 | ENST00000472720.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009464.hg.1 | ENST00000480352.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009464.hg.1 | ENST00000480352.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009464.hg.1 | ENST00000480352.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009464.hg.1 | ENST00000485379.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009464.hg.1 | ENST00000485379.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200009464.hg.1 | ENST00000485379.1 | Ensembl |
