Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR02002996.hg.1 |
TC02000210.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02002996.hg.1 |
ENST00000315285.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02002996.hg.1 |
ENST00000345662.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02002996.hg.1 |
NM_014946.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02002996.hg.1 |
NM_199436.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200149436.hg.1 |
ENST00000315285.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200149436.hg.1 |
ENST00000345662.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200149436.hg.1 |
ENST00000615843.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200149436.hg.1 |
ENST00000621856.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200149436.hg.1 |
NM_014946.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200149436.hg.1 |
NM_199436.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200149436.hg.1 |
ENST00000315285.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200149436.hg.1 |
ENST00000345662.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200149436.hg.1 |
ENST00000615843.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200149436.hg.1 |
ENST00000621856.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200149436.hg.1 |
NM_014946.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200149436.hg.1 |
NM_199436.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200149436.hg.1 |
TC0200007174.hg.1 |
Clariom D TC |