Assay Details
| Gene Symbol: CNTFR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001842.4 | NP_001833.1 | 1 - 2 | 187 | 137 | |
| GenBank mRNA | BC001492.2 | - | 1 - 2 | 144 | 3510004 | 137 |
| BI829871.1 | - | 1 - 2 | 189 | 137 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC001492.2, NM_001842.4 transcripts.
Set Membership: |
|

CNTFR
Genomic Map