Assay Details
Gene Symbol: COX7A2L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001319038.1 | NP_001305967.1 | 2 - 3 | 287 | 99 | |
GenBank mRNA | CA336587.1 | - | 2 - 3 | 251 | 99 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
