Assay Details
Gene Symbol: TMX4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021156.3 | NP_066979.2 | 2 - 3 | 510 | 115 | |
GenBank mRNA | AK074838.1 | - | 2 - 3 | 442 | 115 | |
AK075404.1 | - | 2 - 3 | 489 | 115 | ||
AY359004.1 | - | 2 - 3 | 427 | 115 | ||
BC033787.1 | - | 2 - 3 | 321 | 5165952 | 115 | |
BC044777.1 | - | 2 - 3 | 510 | 115 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC20000611.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC2000008345.hg.1 | ENST00000246024.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008345.hg.1 | ENST00000246024.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008345.hg.1 | ENST00000246024.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008345.hg.1 | ENST00000246024.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008345.hg.1 | ENST00000246024.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008345.hg.1 | ENST00000246024.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008345.hg.1 | ENST00000246024.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008345.hg.1 | ENST00000246024.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008345.hg.1 | ENST00000246024.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008345.hg.1 | ENST00000246024.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008345.hg.1 | ENST00000246024.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008345.hg.1 | ENST00000246024.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008345.hg.1 | ENST00000246024.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008345.hg.1 | ENST00000246024.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008345.hg.1 | ENST00000462384.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008345.hg.1 | ENST00000462384.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008345.hg.1 | ENST00000462384.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008345.hg.1 | ENST00000462384.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008345.hg.1 | ENST00000462384.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008345.hg.1 | ENST00000462384.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008345.hg.1 | ENST00000462384.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008345.hg.1 | ENST00000462384.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008345.hg.1 | ENST00000462384.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008345.hg.1 | ENST00000527925.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008345.hg.1 | ENST00000527925.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008345.hg.1 | ENST00000527925.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008345.hg.1 | ENST00000527925.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008345.hg.1 | ENST00000527925.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008345.hg.1 | ENST00000527925.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008345.hg.1 | ENST00000527925.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008345.hg.1 | ENST00000527925.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008345.hg.1 | ENST00000527925.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008345.hg.1 | ENST00000530935.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008345.hg.1 | ENST00000530935.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008345.hg.1 | ENST00000530935.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008345.hg.1 | ENST00000530935.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008345.hg.1 | ENST00000530935.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008345.hg.1 | ENST00000530935.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008345.hg.1 | ENST00000530935.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008345.hg.1 | NM_021156.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000008345.hg.1 | NM_021156.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000008345.hg.1 | NM_021156.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000008345.hg.1 | NM_021156.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000008345.hg.1 | NM_021156.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000008345.hg.1 | NM_021156.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000008345.hg.1 | NM_021156.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000008345.hg.1 | NM_021156.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000008345.hg.1 | NM_021156.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000008345.hg.1 | NM_021156.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000008345.hg.1 | NM_021156.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000008345.hg.1 | NM_021156.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000008345.hg.1 | NM_021156.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000008345.hg.1 | NM_021156.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR20008831.hg.1 | ENST00000246024.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR20008831.hg.1 | ENST00000530935.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR20008831.hg.1 | NM_021156.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR20008831.hg.1 | TC20000611.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | PSR2000158826.hg.1 | ENST00000246024.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000158826.hg.1 | ENST00000462384.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000158826.hg.1 | ENST00000530935.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000158826.hg.1 | NM_021156.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR2000158826.hg.1 | TC2000008345.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR2000158826.hg.1 | ENST00000246024.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000158826.hg.1 | ENST00000462384.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000158826.hg.1 | ENST00000530935.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000158826.hg.1 | NM_021156.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2000008345.hg.1 | ENST00000246024.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008345.hg.1 | ENST00000246024.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008345.hg.1 | ENST00000246024.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008345.hg.1 | ENST00000246024.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008345.hg.1 | ENST00000246024.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008345.hg.1 | ENST00000246024.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008345.hg.1 | ENST00000246024.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008345.hg.1 | ENST00000246024.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008345.hg.1 | ENST00000246024.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008345.hg.1 | ENST00000246024.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008345.hg.1 | ENST00000246024.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008345.hg.1 | ENST00000246024.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008345.hg.1 | ENST00000246024.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008345.hg.1 | ENST00000246024.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008345.hg.1 | ENST00000462384.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008345.hg.1 | ENST00000462384.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008345.hg.1 | ENST00000462384.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008345.hg.1 | ENST00000462384.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008345.hg.1 | ENST00000462384.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008345.hg.1 | ENST00000462384.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008345.hg.1 | ENST00000462384.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008345.hg.1 | ENST00000462384.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008345.hg.1 | ENST00000462384.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008345.hg.1 | ENST00000527925.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008345.hg.1 | ENST00000527925.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008345.hg.1 | ENST00000527925.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008345.hg.1 | ENST00000527925.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008345.hg.1 | ENST00000527925.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008345.hg.1 | ENST00000527925.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008345.hg.1 | ENST00000527925.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008345.hg.1 | ENST00000527925.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008345.hg.1 | ENST00000527925.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008345.hg.1 | ENST00000530935.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008345.hg.1 | ENST00000530935.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008345.hg.1 | ENST00000530935.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008345.hg.1 | ENST00000530935.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008345.hg.1 | ENST00000530935.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008345.hg.1 | ENST00000530935.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008345.hg.1 | ENST00000530935.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008345.hg.1 | NM_021156.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2000008345.hg.1 | NM_021156.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2000008345.hg.1 | NM_021156.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2000008345.hg.1 | NM_021156.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2000008345.hg.1 | NM_021156.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2000008345.hg.1 | NM_021156.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2000008345.hg.1 | NM_021156.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2000008345.hg.1 | NM_021156.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2000008345.hg.1 | NM_021156.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2000008345.hg.1 | NM_021156.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2000008345.hg.1 | NM_021156.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2000008345.hg.1 | NM_021156.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2000008345.hg.1 | NM_021156.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2000008345.hg.1 | NM_021156.3 | Refseq |
