Assay Details
Gene Symbol: TMX4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021156.3 | NP_066979.2 | 3 - 4 | 556 | 60 | |
GenBank mRNA | AK075404.1 | - | 3 - 4 | 535 | 60 | |
AK226005.1 | - | 2 - 3 | 90 | 60 | ||
AL118494.1 | - | 2 - 3 | 81 | 60 | ||
AY359004.1 | - | 3 - 4 | 473 | 60 | ||
BC033787.1 | - | 3 - 4 | 367 | 5165952 | 60 | |
BC044777.1 | - | 3 - 4 | 556 | 60 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC20000611.hg
Additional Information:
This assay is in the 5'UTR region of the AL118494.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC2000008345.hg.1 | ENST00000246024.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008345.hg.1 | ENST00000246024.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008345.hg.1 | ENST00000246024.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008345.hg.1 | ENST00000246024.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008345.hg.1 | ENST00000246024.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008345.hg.1 | ENST00000246024.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008345.hg.1 | ENST00000246024.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008345.hg.1 | ENST00000246024.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008345.hg.1 | ENST00000246024.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008345.hg.1 | ENST00000246024.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008345.hg.1 | ENST00000246024.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008345.hg.1 | ENST00000246024.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008345.hg.1 | ENST00000246024.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008345.hg.1 | ENST00000246024.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008345.hg.1 | ENST00000462384.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008345.hg.1 | ENST00000462384.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008345.hg.1 | ENST00000462384.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008345.hg.1 | ENST00000462384.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008345.hg.1 | ENST00000462384.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008345.hg.1 | ENST00000462384.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008345.hg.1 | ENST00000462384.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008345.hg.1 | ENST00000462384.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008345.hg.1 | ENST00000462384.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008345.hg.1 | ENST00000527925.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008345.hg.1 | ENST00000527925.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008345.hg.1 | ENST00000527925.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008345.hg.1 | ENST00000527925.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008345.hg.1 | ENST00000527925.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008345.hg.1 | ENST00000527925.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008345.hg.1 | ENST00000527925.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008345.hg.1 | ENST00000527925.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008345.hg.1 | ENST00000527925.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008345.hg.1 | ENST00000530935.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008345.hg.1 | ENST00000530935.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008345.hg.1 | ENST00000530935.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008345.hg.1 | ENST00000530935.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008345.hg.1 | ENST00000530935.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008345.hg.1 | ENST00000530935.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008345.hg.1 | ENST00000530935.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000008345.hg.1 | NM_021156.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000008345.hg.1 | NM_021156.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000008345.hg.1 | NM_021156.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000008345.hg.1 | NM_021156.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000008345.hg.1 | NM_021156.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000008345.hg.1 | NM_021156.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000008345.hg.1 | NM_021156.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000008345.hg.1 | NM_021156.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000008345.hg.1 | NM_021156.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000008345.hg.1 | NM_021156.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000008345.hg.1 | NM_021156.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000008345.hg.1 | NM_021156.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000008345.hg.1 | NM_021156.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000008345.hg.1 | NM_021156.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR20008830.hg.1 | ENST00000246024.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR20008830.hg.1 | ENST00000530935.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR20008830.hg.1 | NM_021156.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR20008830.hg.1 | TC20000611.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | TC2000008345.hg.1 | ENST00000246024.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008345.hg.1 | ENST00000246024.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008345.hg.1 | ENST00000246024.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008345.hg.1 | ENST00000246024.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008345.hg.1 | ENST00000246024.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008345.hg.1 | ENST00000246024.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008345.hg.1 | ENST00000246024.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008345.hg.1 | ENST00000246024.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008345.hg.1 | ENST00000246024.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008345.hg.1 | ENST00000246024.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008345.hg.1 | ENST00000246024.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008345.hg.1 | ENST00000246024.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008345.hg.1 | ENST00000246024.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008345.hg.1 | ENST00000246024.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008345.hg.1 | ENST00000462384.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008345.hg.1 | ENST00000462384.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008345.hg.1 | ENST00000462384.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008345.hg.1 | ENST00000462384.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008345.hg.1 | ENST00000462384.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008345.hg.1 | ENST00000462384.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008345.hg.1 | ENST00000462384.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008345.hg.1 | ENST00000462384.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008345.hg.1 | ENST00000462384.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008345.hg.1 | ENST00000527925.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008345.hg.1 | ENST00000527925.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008345.hg.1 | ENST00000527925.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008345.hg.1 | ENST00000527925.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008345.hg.1 | ENST00000527925.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008345.hg.1 | ENST00000527925.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008345.hg.1 | ENST00000527925.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008345.hg.1 | ENST00000527925.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008345.hg.1 | ENST00000527925.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008345.hg.1 | ENST00000530935.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008345.hg.1 | ENST00000530935.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008345.hg.1 | ENST00000530935.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008345.hg.1 | ENST00000530935.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008345.hg.1 | ENST00000530935.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008345.hg.1 | ENST00000530935.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008345.hg.1 | ENST00000530935.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000008345.hg.1 | NM_021156.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2000008345.hg.1 | NM_021156.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2000008345.hg.1 | NM_021156.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2000008345.hg.1 | NM_021156.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2000008345.hg.1 | NM_021156.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2000008345.hg.1 | NM_021156.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2000008345.hg.1 | NM_021156.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2000008345.hg.1 | NM_021156.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2000008345.hg.1 | NM_021156.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2000008345.hg.1 | NM_021156.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2000008345.hg.1 | NM_021156.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2000008345.hg.1 | NM_021156.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2000008345.hg.1 | NM_021156.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2000008345.hg.1 | NM_021156.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR2000158824.hg.1 | ENST00000246024.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000158824.hg.1 | ENST00000462384.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000158824.hg.1 | ENST00000527925.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000158824.hg.1 | ENST00000530935.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000158824.hg.1 | NM_021156.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR2000158824.hg.1 | TC2000008345.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR2000158824.hg.1 | ENST00000246024.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000158824.hg.1 | ENST00000462384.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000158824.hg.1 | ENST00000527925.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000158824.hg.1 | ENST00000530935.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000158824.hg.1 | NM_021156.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR2000158825.hg.1 | ENST00000246024.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000158825.hg.1 | ENST00000462384.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000158825.hg.1 | ENST00000530935.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000158825.hg.1 | NM_021156.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR2000158825.hg.1 | TC2000008345.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR2000158825.hg.1 | ENST00000246024.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000158825.hg.1 | ENST00000462384.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000158825.hg.1 | ENST00000530935.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2000158825.hg.1 | NM_021156.3 | Refseq |
