Assay Details
| Gene Symbol: GNB5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016194.3 | NP_057278.2 | 2 - 3 | 195 | 72 | |
| GenBank mRNA | AF300650.1 | - | 1 - 2 | 130 | 72 | |
| AW967075.1 | - | 2 - 3 | 195 | 72 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC15001453.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
protein folding
signal transduction
G-protein coupled receptor signaling pathway
positive regulation of GTPase activity
negative regulation of voltage-gated calcium channel activity
GTPase activity
signal transducer activity
GTPase activator activity
protein binding
G-protein gamma-subunit binding
chaperone binding
protein folding
signal transduction
G-protein coupled receptor signaling pathway
positive regulation of GTPase activity
negative regulation of voltage-gated calcium channel activity
GTPase activity
signal transducer activity
GTPase activator activity
protein binding
G-protein gamma-subunit binding
chaperone binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1500009452.hg.1 | ENST00000261837.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009452.hg.1 | ENST00000261837.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009452.hg.1 | ENST00000261837.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009452.hg.1 | ENST00000261837.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009452.hg.1 | ENST00000261837.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009452.hg.1 | ENST00000261837.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009452.hg.1 | ENST00000261837.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009452.hg.1 | ENST00000261837.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009452.hg.1 | ENST00000261837.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009452.hg.1 | ENST00000261837.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009452.hg.1 | ENST00000261837.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009452.hg.1 | ENST00000261837.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009452.hg.1 | ENST00000261837.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009452.hg.1 | ENST00000261837.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009452.hg.1 | ENST00000261837.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009452.hg.1 | ENST00000261837.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009452.hg.1 | ENST00000261837.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009452.hg.1 | ENST00000261837.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009452.hg.1 | ENST00000261837.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009452.hg.1 | ENST00000261837.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009452.hg.1 | ENST00000261837.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009452.hg.1 | ENST00000261837.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009452.hg.1 | ENST00000358784.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009452.hg.1 | ENST00000358784.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009452.hg.1 | ENST00000358784.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009452.hg.1 | ENST00000358784.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009452.hg.1 | ENST00000358784.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009452.hg.1 | ENST00000358784.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009452.hg.1 | ENST00000358784.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009452.hg.1 | ENST00000358784.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009452.hg.1 | ENST00000358784.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009452.hg.1 | ENST00000358784.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009452.hg.1 | ENST00000358784.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009452.hg.1 | ENST00000358784.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009452.hg.1 | ENST00000358784.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009452.hg.1 | ENST00000358784.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009452.hg.1 | ENST00000358784.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009452.hg.1 | ENST00000358784.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009452.hg.1 | ENST00000396335.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009452.hg.1 | ENST00000396335.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009452.hg.1 | ENST00000396335.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009452.hg.1 | ENST00000396335.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009452.hg.1 | ENST00000396335.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009452.hg.1 | ENST00000396335.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009452.hg.1 | ENST00000396335.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009452.hg.1 | ENST00000396335.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009452.hg.1 | ENST00000396335.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009452.hg.1 | ENST00000396335.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009452.hg.1 | ENST00000396335.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009452.hg.1 | ENST00000396335.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009452.hg.1 | ENST00000396335.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009452.hg.1 | ENST00000557936.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009452.hg.1 | ENST00000557936.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009452.hg.1 | ENST00000557936.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009452.hg.1 | ENST00000557936.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009452.hg.1 | ENST00000557936.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009452.hg.1 | ENST00000557936.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009452.hg.1 | ENST00000557936.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009452.hg.1 | ENST00000557936.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009452.hg.1 | ENST00000557936.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009452.hg.1 | ENST00000557936.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009452.hg.1 | ENST00000558122.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009452.hg.1 | ENST00000558122.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009452.hg.1 | ENST00000558519.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009452.hg.1 | ENST00000558519.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009452.hg.1 | ENST00000558519.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009452.hg.1 | ENST00000558519.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009452.hg.1 | ENST00000558519.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009452.hg.1 | ENST00000558519.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009452.hg.1 | ENST00000558519.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009452.hg.1 | ENST00000558519.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009452.hg.1 | ENST00000558519.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009452.hg.1 | ENST00000559348.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009452.hg.1 | ENST00000559348.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009452.hg.1 | ENST00000559348.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009452.hg.1 | ENST00000559348.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009452.hg.1 | ENST00000559348.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009452.hg.1 | ENST00000559348.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009452.hg.1 | ENST00000559348.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009452.hg.1 | ENST00000559348.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009452.hg.1 | ENST00000559348.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009452.hg.1 | ENST00000559348.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009452.hg.1 | ENST00000559541.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009452.hg.1 | ENST00000559541.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009452.hg.1 | ENST00000559541.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009452.hg.1 | ENST00000559541.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009452.hg.1 | ENST00000559541.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009452.hg.1 | ENST00000559541.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009452.hg.1 | ENST00000559541.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009452.hg.1 | ENST00000559541.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009452.hg.1 | ENST00000560075.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009452.hg.1 | ENST00000560075.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009452.hg.1 | ENST00000560075.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009452.hg.1 | ENST00000560075.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009452.hg.1 | ENST00000560075.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009452.hg.1 | ENST00000560075.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009452.hg.1 | ENST00000560085.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009452.hg.1 | ENST00000560085.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009452.hg.1 | ENST00000560085.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009452.hg.1 | ENST00000560085.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009452.hg.1 | ENST00000560085.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009452.hg.1 | ENST00000560116.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009452.hg.1 | ENST00000560116.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009452.hg.1 | ENST00000560116.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009452.hg.1 | ENST00000560116.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009452.hg.1 | ENST00000561313.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009452.hg.1 | ENST00000561313.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009452.hg.1 | ENST00000561313.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009452.hg.1 | ENST00000561313.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009452.hg.1 | ENST00000561313.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009452.hg.1 | ENST00000561313.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009452.hg.1 | ENST00000561313.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009452.hg.1 | NM_006578.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009452.hg.1 | NM_006578.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009452.hg.1 | NM_006578.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009452.hg.1 | NM_006578.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009452.hg.1 | NM_006578.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009452.hg.1 | NM_006578.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009452.hg.1 | NM_006578.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009452.hg.1 | NM_006578.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009452.hg.1 | NM_006578.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009452.hg.1 | NM_006578.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009452.hg.1 | NM_006578.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009452.hg.1 | NM_006578.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009452.hg.1 | NM_006578.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009452.hg.1 | NM_006578.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009452.hg.1 | NM_006578.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009452.hg.1 | NM_006578.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009452.hg.1 | NM_006578.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009452.hg.1 | NM_016194.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009452.hg.1 | NM_016194.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009452.hg.1 | NM_016194.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009452.hg.1 | NM_016194.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009452.hg.1 | NM_016194.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009452.hg.1 | NM_016194.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009452.hg.1 | NM_016194.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009452.hg.1 | NM_016194.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009452.hg.1 | NM_016194.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009452.hg.1 | NM_016194.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009452.hg.1 | NM_016194.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009452.hg.1 | NM_016194.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009452.hg.1 | NM_016194.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009452.hg.1 | NM_016194.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009452.hg.1 | NM_016194.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009452.hg.1 | NM_016194.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009452.hg.1 | NM_016194.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009452.hg.1 | NM_016194.3 | Refseq |

GNB5
Genomic Map