Assay Details
| Gene Symbol: KAT6A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001305878.1 | NP_001292807.1 | 12 - 13 | 2446 | 64 | |
| NM_006766.4 | NP_006757.2 | 12 - 13 | 2446 | 64 | ||
| XM_011544656.2 | XP_011542958.1 | 14 - 15 | 2672 | 64 | ||
| XM_011544657.2 | XP_011542959.1 | 13 - 14 | 2539 | 64 | ||
| XM_011544658.2 | XP_011542960.1 | 14 - 15 | 2628 | 64 | ||
| XM_011544659.2 | XP_011542961.1 | 14 - 15 | 2651 | 64 | ||
| XM_017013863.1 | XP_016869352.1 | 13 - 14 | 2540 | 64 | ||
| XM_017013864.1 | XP_016869353.1 | 13 - 14 | 2496 | 64 | ||
| GenBank mRNA | AK128323.1 | - | 5 - 6 | 1828 | 64 | |
| BC142659.1 | - | 12 - 13 | 2393 | 64 | ||
| BC142959.1 | - | 12 - 13 | 2407 | 64 | ||
| U47742.1 | - | 12 - 13 | 2388 | 64 | ||
More Information
Set Membership: |
|

KAT6A
Genomic Map