Assay Details
| Gene Symbol: ERCC4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_005236.2 | NP_005227.1 | 3 - 4 | 588 | 151 | |
| XM_011522424.2 | XP_011520726.1 | 4 - 5 | 745 | 151 | ||
| XM_017023043.1 | XP_016878532.1 | 3 - 4 | 554 | 151 | ||
| GenBank mRNA | AK289726.1 | - | 3 - 4 | 587 | 151 | |
| AK308341.1 | - | 3 - 4 | 554 | 151 | ||
| BC020741.1 | - | 3 - 4 | 559 | 151 | ||
| BC142631.1 | - | 3 - 4 | 588 | 151 | ||
| U64315.1 | - | 3 - 4 | 561 | 151 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC16000170.hg.1
Additional Information:
This assay is in the 5'UTR region of the XM_017023043.1 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
resolution of meiotic recombination intermediates
telomere maintenance
double-strand break repair via homologous recombination
DNA repair
transcription-coupled nucleotide-excision repair
nucleotide-excision repair
nucleotide-excision repair, preincision complex stabilization
nucleotide-excision repair, DNA incision, 3'-to lesion
nucleotide-excision repair, DNA incision, 5'-to lesion
response to UV
UV protection
negative regulation of telomere maintenance
nucleotide-excision repair, DNA incision
cellular response to UV
interstrand cross-link repair
global genome nucleotide-excision repair
nucleotide-excision repair involved in interstrand cross-link repair
single-stranded DNA endodeoxyribonuclease activity
TFIID-class transcription factor binding
damaged DNA binding
single-stranded DNA binding
endodeoxyribonuclease activity
protein binding
protein C-terminus binding
structure-specific DNA binding
protein N-terminus binding
resolution of meiotic recombination intermediates
telomere maintenance
double-strand break repair via homologous recombination
DNA repair
transcription-coupled nucleotide-excision repair
nucleotide-excision repair
nucleotide-excision repair, preincision complex stabilization
nucleotide-excision repair, DNA incision, 3'-to lesion
nucleotide-excision repair, DNA incision, 5'-to lesion
response to UV
UV protection
negative regulation of telomere maintenance
nucleotide-excision repair, DNA incision
cellular response to UV
interstrand cross-link repair
global genome nucleotide-excision repair
nucleotide-excision repair involved in interstrand cross-link repair
single-stranded DNA endodeoxyribonuclease activity
TFIID-class transcription factor binding
damaged DNA binding
single-stranded DNA binding
endodeoxyribonuclease activity
protein binding
protein C-terminus binding
structure-specific DNA binding
protein N-terminus binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR16002572.hg.1 | ENST00000311895.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16002572.hg.1 | ENST00000389138.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16002572.hg.1 | ENST00000439007.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16002572.hg.1 | NM_005236.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16002572.hg.1 | TC16000170.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006976.hg.1 | ENST00000311895.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006976.hg.1 | ENST00000311895.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006976.hg.1 | ENST00000311895.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006976.hg.1 | ENST00000311895.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006976.hg.1 | ENST00000311895.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006976.hg.1 | ENST00000311895.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006976.hg.1 | ENST00000311895.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006976.hg.1 | ENST00000311895.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006976.hg.1 | ENST00000311895.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006976.hg.1 | ENST00000311895.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006976.hg.1 | ENST00000311895.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006976.hg.1 | ENST00000311895.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006976.hg.1 | ENST00000311895.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006976.hg.1 | ENST00000311895.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006976.hg.1 | ENST00000311895.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006976.hg.1 | ENST00000311895.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006976.hg.1 | ENST00000311895.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006976.hg.1 | ENST00000311895.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006976.hg.1 | ENST00000389138.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006976.hg.1 | ENST00000389138.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006976.hg.1 | ENST00000389138.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006976.hg.1 | ENST00000389138.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006976.hg.1 | ENST00000389138.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006976.hg.1 | ENST00000389138.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006976.hg.1 | ENST00000389138.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006976.hg.1 | ENST00000389138.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006976.hg.1 | ENST00000389138.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006976.hg.1 | ENST00000389138.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006976.hg.1 | ENST00000389138.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006976.hg.1 | ENST00000389138.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006976.hg.1 | ENST00000462862.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006976.hg.1 | ENST00000462862.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006976.hg.1 | ENST00000462862.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006976.hg.1 | ENST00000462862.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006976.hg.1 | ENST00000573018.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006976.hg.1 | ENST00000573018.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006976.hg.1 | ENST00000573018.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006976.hg.1 | ENST00000574194.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006976.hg.1 | ENST00000574194.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006976.hg.1 | ENST00000574194.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006976.hg.1 | ENST00000574194.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006976.hg.1 | ENST00000574781.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006976.hg.1 | ENST00000574781.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006976.hg.1 | ENST00000574781.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006976.hg.1 | ENST00000574781.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006976.hg.1 | ENST00000574781.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006976.hg.1 | ENST00000575156.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006976.hg.1 | ENST00000575156.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006976.hg.1 | ENST00000575156.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006976.hg.1 | ENST00000575156.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006976.hg.1 | ENST00000575156.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006976.hg.1 | ENST00000575156.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006976.hg.1 | ENST00000575156.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006976.hg.1 | ENST00000575156.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006976.hg.1 | ENST00000575156.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006976.hg.1 | ENST00000575156.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006976.hg.1 | ENST00000575156.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006976.hg.1 | ENST00000576348.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006976.hg.1 | ENST00000576348.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006976.hg.1 | ENST00000576348.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006976.hg.1 | ENST00000576348.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006976.hg.1 | NM_005236.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006976.hg.1 | NM_005236.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006976.hg.1 | NM_005236.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006976.hg.1 | NM_005236.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006976.hg.1 | NM_005236.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006976.hg.1 | NM_005236.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006976.hg.1 | NM_005236.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006976.hg.1 | NM_005236.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006976.hg.1 | NM_005236.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006976.hg.1 | NM_005236.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006976.hg.1 | NM_005236.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006976.hg.1 | NM_005236.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006976.hg.1 | NM_005236.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006976.hg.1 | NM_005236.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006976.hg.1 | NM_005236.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006976.hg.1 | NM_005236.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006976.hg.1 | NM_005236.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600006976.hg.1 | NM_005236.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600149401.hg.1 | ENST00000311895.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600149401.hg.1 | ENST00000574781.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600149401.hg.1 | ENST00000575156.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600149401.hg.1 | NM_005236.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600149401.hg.1 | ENST00000311895.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600149401.hg.1 | ENST00000574781.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600149401.hg.1 | ENST00000575156.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600149401.hg.1 | NM_005236.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600149401.hg.1 | TC1600006976.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1600149402.hg.1 | ENST00000311895.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600149402.hg.1 | ENST00000574194.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600149402.hg.1 | ENST00000574781.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600149402.hg.1 | ENST00000575156.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600149402.hg.1 | NM_005236.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600149402.hg.1 | ENST00000311895.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600149402.hg.1 | ENST00000574194.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600149402.hg.1 | ENST00000574781.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600149402.hg.1 | ENST00000575156.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600149402.hg.1 | NM_005236.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600149402.hg.1 | TC1600006976.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1600006976.hg.1 | ENST00000311895.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006976.hg.1 | ENST00000311895.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006976.hg.1 | ENST00000311895.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006976.hg.1 | ENST00000311895.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006976.hg.1 | ENST00000311895.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006976.hg.1 | ENST00000311895.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006976.hg.1 | ENST00000311895.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006976.hg.1 | ENST00000311895.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006976.hg.1 | ENST00000311895.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006976.hg.1 | ENST00000311895.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006976.hg.1 | ENST00000311895.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006976.hg.1 | ENST00000311895.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006976.hg.1 | ENST00000311895.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006976.hg.1 | ENST00000311895.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006976.hg.1 | ENST00000311895.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006976.hg.1 | ENST00000311895.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006976.hg.1 | ENST00000311895.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006976.hg.1 | ENST00000311895.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006976.hg.1 | ENST00000389138.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006976.hg.1 | ENST00000389138.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006976.hg.1 | ENST00000389138.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006976.hg.1 | ENST00000389138.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006976.hg.1 | ENST00000389138.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006976.hg.1 | ENST00000389138.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006976.hg.1 | ENST00000389138.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006976.hg.1 | ENST00000389138.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006976.hg.1 | ENST00000389138.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006976.hg.1 | ENST00000389138.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006976.hg.1 | ENST00000389138.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006976.hg.1 | ENST00000389138.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006976.hg.1 | ENST00000462862.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006976.hg.1 | ENST00000462862.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006976.hg.1 | ENST00000462862.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006976.hg.1 | ENST00000462862.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006976.hg.1 | ENST00000573018.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006976.hg.1 | ENST00000573018.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006976.hg.1 | ENST00000573018.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006976.hg.1 | ENST00000574194.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006976.hg.1 | ENST00000574194.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006976.hg.1 | ENST00000574194.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006976.hg.1 | ENST00000574194.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006976.hg.1 | ENST00000574781.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006976.hg.1 | ENST00000574781.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006976.hg.1 | ENST00000574781.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006976.hg.1 | ENST00000574781.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006976.hg.1 | ENST00000574781.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006976.hg.1 | ENST00000575156.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006976.hg.1 | ENST00000575156.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006976.hg.1 | ENST00000575156.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006976.hg.1 | ENST00000575156.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006976.hg.1 | ENST00000575156.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006976.hg.1 | ENST00000575156.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006976.hg.1 | ENST00000575156.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006976.hg.1 | ENST00000575156.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006976.hg.1 | ENST00000575156.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006976.hg.1 | ENST00000575156.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006976.hg.1 | ENST00000575156.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006976.hg.1 | ENST00000576348.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006976.hg.1 | ENST00000576348.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006976.hg.1 | ENST00000576348.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006976.hg.1 | ENST00000576348.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600006976.hg.1 | NM_005236.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600006976.hg.1 | NM_005236.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600006976.hg.1 | NM_005236.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600006976.hg.1 | NM_005236.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600006976.hg.1 | NM_005236.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600006976.hg.1 | NM_005236.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600006976.hg.1 | NM_005236.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600006976.hg.1 | NM_005236.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600006976.hg.1 | NM_005236.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600006976.hg.1 | NM_005236.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600006976.hg.1 | NM_005236.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600006976.hg.1 | NM_005236.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600006976.hg.1 | NM_005236.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600006976.hg.1 | NM_005236.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600006976.hg.1 | NM_005236.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600006976.hg.1 | NM_005236.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600006976.hg.1 | NM_005236.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600006976.hg.1 | NM_005236.2 | Refseq |

ERCC4
Genomic Map