Assay Details
| Gene Symbol: HNRNPK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001318186.1 | NP_001305115.1 | 2 - 3 | 205 | 98 | |
| NM_001318187.1 | NP_001305116.1 | 2 - 3 | 205 | 98 | ||
| NM_001318188.1 | NP_001305117.1 | 2 - 3 | 205 | 98 | ||
| NM_002140.4 | NP_002131.2 | 2 - 3 | 366 | 98 | ||
| NM_031262.3 | NP_112552.1 | 2 - 3 | 366 | 98 | ||
| NM_031263.3 | NP_112553.1 | 2 - 3 | 205 | 98 | ||
| XM_005251960.2 | XP_005252017.1 | 2 - 3 | 184 | 98 | ||
| XM_005251963.3 | XP_005252020.1 | 2 - 3 | 218 | 98 | ||
| XM_005251965.2 | XP_005252022.1 | 2 - 3 | 176 | 98 | ||
| XM_011518616.1 | XP_011516918.1 | 2 - 3 | 218 | 98 | ||
| XM_017014668.1 | XP_016870157.1 | 2 - 3 | 184 | 98 | ||
| XM_017014669.1 | XP_016870158.1 | 2 - 3 | 176 | 98 | ||
| GenBank mRNA | AB209562.1 | - | 2 - 3 | 134 | 96 | |
| AK096385.1 | - | 2 - 3 | 142 | 98 | ||
| AK291336.1 | - | 2 - 3 | 188 | 98 | ||
| AK294067.1 | - | 2 - 3 | 178 | 98 | ||
| AK300744.1 | - | 2 - 3 | 148 | 98 | ||
| BC000355.2 | - | 1 - 2 | 101 | 2964383 | 98 | |
| BC014980.1 | - | 2 - 3 | 172 | 4906241 | 98 | |
| BC025321.1 | - | 2 - 3 | 124 | 98 | ||
| DA394419.1 | - | 2 - 3 | 203 | 98 | ||
| DC318847.1 | - | 2 - 3 | 205 | 98 | ||
| DC407458.1 | - | 2 - 3 | 238 | 98 | ||
| DC428989.1 | - | 2 - 3 | 366 | 98 | ||
| S74678.1 | - | 1 - 2 | 86 | 98 | ||
| X72727.1 | - | 2 - 3 | 188 | 98 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK096385.1, AK291336.1, AK294067.1, AK300744.1, BC000355.2, BC014980.1, NM_001318186.1, NM_001318187.1, NM_001318188.1, NM_002140.4, NM_031262.3, NM_031263.3, S74678.1, X72727.1, XM_005251960.2, XM_005251963.3, XM_005251965.2, XM_011518616.1, XM_017014668.1, XM_017014669.1 transcripts.
Set Membership: |
|

HNRNPK
Genomic Map