Assay Details
| Gene Symbol: HNRNPK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001318186.1 | NP_001305115.1 | 12 - 13 | 1253 | 94 | |
| NM_001318187.1 | NP_001305116.1 | 12 - 13 | 1253 | 94 | ||
| NM_001318188.1 | NP_001305117.1 | 13 - 14 | 1325 | 94 | ||
| NM_002140.4 | NP_002131.2 | 13 - 14 | 1486 | 94 | ||
| NM_031262.3 | NP_112552.1 | 13 - 14 | 1486 | 94 | ||
| NM_031263.3 | NP_112553.1 | 13 - 14 | 1325 | 94 | ||
| XM_005251960.2 | XP_005252017.1 | 13 - 14 | 1304 | 94 | ||
| XM_005251963.3 | XP_005252020.1 | 12 - 13 | 1266 | 94 | ||
| XM_005251965.2 | XP_005252022.1 | 12 - 13 | 1224 | 94 | ||
| XM_011518616.1 | XP_011516918.1 | 12 - 13 | 1266 | 94 | ||
| XM_017014668.1 | XP_016870157.1 | 13 - 14 | 1304 | 94 | ||
| XM_017014669.1 | XP_016870158.1 | 12 - 13 | 1224 | 94 | ||
| GenBank mRNA | AB209562.1 | - | 12 - 13 | 1182 | 94 | |
| AB451263.1 | - | 11 - 12 | 1099 | 94 | ||
| AB451390.1 | - | 11 - 12 | 1099 | 94 | ||
| AK096385.1 | - | 12 - 13 | 1111 | 94 | ||
| AK123117.1 | - | 7 - 8 | 2051 | 94 | ||
| AK291336.1 | - | 13 - 14 | 1308 | 94 | ||
| AK294067.1 | - | 12 - 13 | 1283 | 94 | ||
| AK300744.1 | - | 12 - 13 | 1196 | 94 | ||
| AK307962.1 | - | 14 - 15 | 1146 | 94 | ||
| AY911506.1 | - | 12 - 13 | 1162 | 94 | ||
| BC000355.2 | - | 12 - 13 | 1221 | 2964383 | 94 | |
| BC014980.1 | - | 13 - 14 | 1292 | 4906241 | 94 | |
| BC025321.1 | - | 13 - 14 | 1244 | 94 | ||
| CR456771.1 | - | 11 - 12 | 1099 | 94 | ||
| S74678.1 | - | 12 - 13 | 1206 | 94 | ||
| X72727.1 | - | 13 - 14 | 1308 | 94 | ||
More Information
Set Membership: |
|

HNRNPK
Genomic Map