Assay Details
| Gene Symbol: KIF2A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001243953.1 | NP_001230882.1 | 5 - 6 | 712 | 115 | |
| GenBank mRNA | AK302270.1 | - | 5 - 6 | 406 | 115 | |
| AK307880.1 | - | 5 - 6 | 422 | 115 | ||
| AK315866.1 | - | 5 - 6 | 396 | 115 | ||
More Information
Set Membership: |
|

KIF2A
Genomic Map