Assay Details
| Gene Symbol: DGUOK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001318861.1 | NP_001305790.1 | 1 - 2 | 225 | 111 | |
| NM_001318862.1 | NP_001305791.1 | 1 - 2 | 225 | 111 | ||
| NR_134897.1 | 1 - 2 | 225 | 111 | |||
| GenBank mRNA | BG575800.1 | - | 1 - 2 | 130 | 111 | |
| CN311954.1 | - | 1 - 2 | 182 | 111 | ||
| CN311963.1 | - | 1 - 2 | 154 | 111 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001318861.1, NM_001318862.1 transcripts.
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
protein phosphorylation
guanosine metabolic process
deoxyribonucleoside monophosphate biosynthetic process
nucleotide biosynthetic process
negative regulation of neuron projection development
purine-containing compound salvage
dGTP metabolic process
purine deoxyribonucleoside metabolic process
deoxyguanosine kinase activity
ATP binding
nucleoside kinase activity
protein phosphorylation
guanosine metabolic process
deoxyribonucleoside monophosphate biosynthetic process
nucleotide biosynthetic process
negative regulation of neuron projection development
purine-containing compound salvage
dGTP metabolic process
purine deoxyribonucleoside metabolic process
deoxyguanosine kinase activity
ATP binding
nucleoside kinase activity
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0200008063.hg.1 | ENST00000264093.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008063.hg.1 | ENST00000264093.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008063.hg.1 | ENST00000264093.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008063.hg.1 | ENST00000264093.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008063.hg.1 | ENST00000264093.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008063.hg.1 | ENST00000264093.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008063.hg.1 | ENST00000264093.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008063.hg.1 | ENST00000264093.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008063.hg.1 | ENST00000264093.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008063.hg.1 | ENST00000264093.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008063.hg.1 | ENST00000264093.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008063.hg.1 | ENST00000348222.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008063.hg.1 | ENST00000348222.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008063.hg.1 | ENST00000348222.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008063.hg.1 | ENST00000348222.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008063.hg.1 | ENST00000348222.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008063.hg.1 | ENST00000348222.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008063.hg.1 | ENST00000348222.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008063.hg.1 | ENST00000348222.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008063.hg.1 | ENST00000418996.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008063.hg.1 | ENST00000418996.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008063.hg.1 | ENST00000418996.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008063.hg.1 | ENST00000418996.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008063.hg.1 | ENST00000418996.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008063.hg.1 | ENST00000418996.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008063.hg.1 | ENST00000462551.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008063.hg.1 | ENST00000462551.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008063.hg.1 | ENST00000462551.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008063.hg.1 | ENST00000462551.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008063.hg.1 | ENST00000462685.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008063.hg.1 | ENST00000462685.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008063.hg.1 | ENST00000462685.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008063.hg.1 | ENST00000462685.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008063.hg.1 | ENST00000462685.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008063.hg.1 | ENST00000462685.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008063.hg.1 | ENST00000462685.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008063.hg.1 | ENST00000489796.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008063.hg.1 | ENST00000489796.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008063.hg.1 | ENST00000489796.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008063.hg.1 | ENST00000489796.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008063.hg.1 | ENST00000489796.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008063.hg.1 | ENST00000489796.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008063.hg.1 | ENST00000493055.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008063.hg.1 | ENST00000493055.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008063.hg.1 | ENST00000493055.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008063.hg.1 | ENST00000493055.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008063.hg.1 | ENST00000629438.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008063.hg.1 | ENST00000629438.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008063.hg.1 | ENST00000629438.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008063.hg.1 | ENST00000629438.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008063.hg.1 | ENST00000629438.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008063.hg.1 | ENST00000629438.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008063.hg.1 | ENST00000629438.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008063.hg.1 | ENST00000629438.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008063.hg.1 | ENST00000629438.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008063.hg.1 | ENST00000629438.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008063.hg.1 | NM_080916.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008063.hg.1 | NM_080916.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008063.hg.1 | NM_080916.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008063.hg.1 | NM_080916.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008063.hg.1 | NM_080916.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008063.hg.1 | NM_080916.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008063.hg.1 | NM_080916.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008063.hg.1 | NM_080916.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008063.hg.1 | NM_080916.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008063.hg.1 | NM_080916.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008063.hg.1 | NM_080916.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008063.hg.1 | NM_080918.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008063.hg.1 | NM_080918.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008063.hg.1 | NM_080918.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008063.hg.1 | NM_080918.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008063.hg.1 | NM_080918.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008063.hg.1 | NM_080918.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008063.hg.1 | NM_080918.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008063.hg.1 | NM_080918.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008063.hg.1 | ENST00000264093.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008063.hg.1 | ENST00000264093.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008063.hg.1 | ENST00000264093.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008063.hg.1 | ENST00000264093.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008063.hg.1 | ENST00000264093.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008063.hg.1 | ENST00000264093.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008063.hg.1 | ENST00000264093.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008063.hg.1 | ENST00000264093.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008063.hg.1 | ENST00000264093.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008063.hg.1 | ENST00000264093.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008063.hg.1 | ENST00000264093.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008063.hg.1 | ENST00000348222.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008063.hg.1 | ENST00000348222.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008063.hg.1 | ENST00000348222.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008063.hg.1 | ENST00000348222.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008063.hg.1 | ENST00000348222.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008063.hg.1 | ENST00000348222.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008063.hg.1 | ENST00000348222.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008063.hg.1 | ENST00000348222.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008063.hg.1 | ENST00000418996.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008063.hg.1 | ENST00000418996.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008063.hg.1 | ENST00000418996.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008063.hg.1 | ENST00000418996.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008063.hg.1 | ENST00000418996.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008063.hg.1 | ENST00000418996.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008063.hg.1 | ENST00000462551.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008063.hg.1 | ENST00000462551.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008063.hg.1 | ENST00000462551.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008063.hg.1 | ENST00000462551.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008063.hg.1 | ENST00000462685.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008063.hg.1 | ENST00000462685.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008063.hg.1 | ENST00000462685.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008063.hg.1 | ENST00000462685.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008063.hg.1 | ENST00000462685.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008063.hg.1 | ENST00000462685.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008063.hg.1 | ENST00000462685.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008063.hg.1 | ENST00000489796.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008063.hg.1 | ENST00000489796.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008063.hg.1 | ENST00000489796.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008063.hg.1 | ENST00000489796.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008063.hg.1 | ENST00000489796.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008063.hg.1 | ENST00000489796.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008063.hg.1 | ENST00000493055.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008063.hg.1 | ENST00000493055.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008063.hg.1 | ENST00000493055.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008063.hg.1 | ENST00000493055.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008063.hg.1 | ENST00000629438.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008063.hg.1 | ENST00000629438.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008063.hg.1 | ENST00000629438.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008063.hg.1 | ENST00000629438.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008063.hg.1 | ENST00000629438.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008063.hg.1 | ENST00000629438.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008063.hg.1 | ENST00000629438.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008063.hg.1 | ENST00000629438.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008063.hg.1 | ENST00000629438.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008063.hg.1 | ENST00000629438.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008063.hg.1 | NM_080916.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008063.hg.1 | NM_080916.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008063.hg.1 | NM_080916.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008063.hg.1 | NM_080916.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008063.hg.1 | NM_080916.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008063.hg.1 | NM_080916.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008063.hg.1 | NM_080916.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008063.hg.1 | NM_080916.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008063.hg.1 | NM_080916.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008063.hg.1 | NM_080916.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008063.hg.1 | NM_080916.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008063.hg.1 | NM_080918.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008063.hg.1 | NM_080918.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008063.hg.1 | NM_080918.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008063.hg.1 | NM_080918.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008063.hg.1 | NM_080918.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008063.hg.1 | NM_080918.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008063.hg.1 | NM_080918.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008063.hg.1 | NM_080918.2 | Refseq |

DGUOK
Genomic Map