Assay Details
| Gene Symbol: DGUOK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001318861.1 | NP_001305790.1 | 2 - 3 | 315 | 106 | |
| NM_001318862.1 | NP_001305791.1 | 2 - 3 | 315 | 106 | ||
| NR_134897.1 | 2 - 3 | 315 | 106 | |||
| GenBank mRNA | BG575800.1 | - | 2 - 3 | 220 | 106 | |
| CN311954.1 | - | 2 - 3 | 272 | 106 | ||
| CN311963.1 | - | 2 - 3 | 244 | 106 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001318861.1, NM_001318862.1 transcripts.
Set Membership: |
|

DGUOK
Genomic Map