Assay Details
Gene Symbol: TCF7L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031283.2 | NP_112573.1 | 2 - 3 | 535 | 119 | |
XM_006712109.2 | XP_006712172.1 | 2 - 3 | 325 | 119 | ||
GenBank mRNA | AB031046.1 | - | 2 - 3 | 384 | 119 | |
BC058894.1 | - | 2 - 3 | 544 | 6141641 | 119 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC02000502.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
chromatin organization
transcription, DNA-templated
regulation of transcription, DNA-templated
regulation of transcription from RNA polymerase II promoter
regulation of Wnt signaling pathway
canonical Wnt signaling pathway
beta-catenin-TCF complex assembly
DNA binding
transcription factor activity, sequence-specific DNA binding
beta-catenin binding
sequence-specific DNA binding
transcription regulatory region DNA binding
chromatin organization
transcription, DNA-templated
regulation of transcription, DNA-templated
regulation of transcription from RNA polymerase II promoter
regulation of Wnt signaling pathway
canonical Wnt signaling pathway
beta-catenin-TCF complex assembly
DNA binding
transcription factor activity, sequence-specific DNA binding
beta-catenin binding
sequence-specific DNA binding
transcription regulatory region DNA binding
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR02007698.hg.1 | TC02000502.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR02007698.hg.1 | ENST00000282111.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR02007698.hg.1 | ENST00000494519.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR02007698.hg.1 | NM_031283.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR02007697.hg.1 | TC02000502.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR02007697.hg.1 | ENST00000282111.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR02007697.hg.1 | NM_031283.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200008236.hg.1 | ENST00000282111.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008236.hg.1 | ENST00000282111.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008236.hg.1 | ENST00000282111.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008236.hg.1 | ENST00000282111.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008236.hg.1 | ENST00000282111.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008236.hg.1 | ENST00000282111.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008236.hg.1 | ENST00000282111.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008236.hg.1 | ENST00000282111.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008236.hg.1 | ENST00000282111.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008236.hg.1 | ENST00000282111.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008236.hg.1 | ENST00000282111.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008236.hg.1 | ENST00000282111.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008236.hg.1 | ENST00000282111.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008236.hg.1 | ENST00000282111.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008236.hg.1 | ENST00000282111.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008236.hg.1 | ENST00000282111.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008236.hg.1 | ENST00000282111.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008236.hg.1 | ENST00000442813.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008236.hg.1 | ENST00000442813.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008236.hg.1 | ENST00000442813.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008236.hg.1 | ENST00000442813.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008236.hg.1 | ENST00000442813.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008236.hg.1 | ENST00000490744.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008236.hg.1 | ENST00000490744.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008236.hg.1 | ENST00000490744.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008236.hg.1 | ENST00000490744.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008236.hg.1 | ENST00000494519.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008236.hg.1 | ENST00000494519.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008236.hg.1 | NM_031283.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200008236.hg.1 | NM_031283.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200008236.hg.1 | NM_031283.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200008236.hg.1 | NM_031283.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200008236.hg.1 | NM_031283.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200008236.hg.1 | NM_031283.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200008236.hg.1 | NM_031283.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200008236.hg.1 | NM_031283.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200008236.hg.1 | NM_031283.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200008236.hg.1 | NM_031283.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200008236.hg.1 | NM_031283.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200008236.hg.1 | NM_031283.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200008236.hg.1 | NM_031283.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200008236.hg.1 | NM_031283.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200008236.hg.1 | NM_031283.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200008236.hg.1 | NM_031283.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200008236.hg.1 | ENST00000282111.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008236.hg.1 | ENST00000282111.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008236.hg.1 | ENST00000282111.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008236.hg.1 | ENST00000282111.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008236.hg.1 | ENST00000282111.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008236.hg.1 | ENST00000282111.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008236.hg.1 | ENST00000282111.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008236.hg.1 | ENST00000282111.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008236.hg.1 | ENST00000282111.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008236.hg.1 | ENST00000282111.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008236.hg.1 | ENST00000282111.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008236.hg.1 | ENST00000282111.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008236.hg.1 | ENST00000282111.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008236.hg.1 | ENST00000282111.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008236.hg.1 | ENST00000282111.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008236.hg.1 | ENST00000282111.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008236.hg.1 | ENST00000282111.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008236.hg.1 | ENST00000442813.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008236.hg.1 | ENST00000442813.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008236.hg.1 | ENST00000442813.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008236.hg.1 | ENST00000442813.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008236.hg.1 | ENST00000442813.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008236.hg.1 | ENST00000490744.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008236.hg.1 | ENST00000490744.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008236.hg.1 | ENST00000490744.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008236.hg.1 | ENST00000490744.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008236.hg.1 | ENST00000494519.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008236.hg.1 | ENST00000494519.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008236.hg.1 | NM_031283.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200008236.hg.1 | NM_031283.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200008236.hg.1 | NM_031283.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200008236.hg.1 | NM_031283.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200008236.hg.1 | NM_031283.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200008236.hg.1 | NM_031283.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200008236.hg.1 | NM_031283.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200008236.hg.1 | NM_031283.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200008236.hg.1 | NM_031283.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200008236.hg.1 | NM_031283.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200008236.hg.1 | NM_031283.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200008236.hg.1 | NM_031283.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200008236.hg.1 | NM_031283.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200008236.hg.1 | NM_031283.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200008236.hg.1 | NM_031283.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0200008236.hg.1 | NM_031283.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200157007.hg.1 | ENST00000282111.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200157007.hg.1 | ENST00000494519.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200157007.hg.1 | NM_031283.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200157007.hg.1 | ENST00000282111.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200157007.hg.1 | ENST00000494519.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200157007.hg.1 | NM_031283.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200157007.hg.1 | TC0200008236.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0200157006.hg.1 | ENST00000282111.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200157006.hg.1 | NM_031283.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200157006.hg.1 | ENST00000282111.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200157006.hg.1 | NM_031283.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200157006.hg.1 | TC0200008236.hg.1 | Clariom D TC |
