Assay Details
| Gene Symbol: LPCAT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_024830.3 | NP_079106.3 | 11 - 12 | 1296 | 82 | |
| XM_005248373.3 | XP_005248430.1 | 12 - 13 | 1204 | 82 | ||
| XM_011514132.1 | XP_011512434.1 | 11 - 12 | 1305 | 82 | ||
| XM_011514134.1 | XP_011512436.1 | 11 - 12 | 1071 | 82 | ||
| XM_017009879.1 | XP_016865368.1 | 5 - 6 | 607 | 82 | ||
| GenBank mRNA | AB244719.2 | - | 11 - 12 | 1179 | 82 | |
| AF530061.1 | - | 2 - 3 | 288 | 82 | ||
| AK022499.1 | - | 6 - 7 | 475 | 82 | ||
| AK022505.1 | - | 6 - 7 | 475 | 82 | ||
| AK090444.1 | - | 9 - 10 | 775 | 82 | ||
| BC020166.1 | - | 9 - 10 | 762 | 82 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC05001110.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
phosphatidic acid biosynthetic process
phospholipid biosynthetic process
triglyceride biosynthetic process
phosphatidylglycerol acyl-chain remodeling
phosphatidylcholine acyl-chain remodeling
surfactant homeostasis
positive regulation of protein catabolic process
retina development in camera-type eye
negative regulation of phosphatidylcholine biosynthetic process
1-acylglycerol-3-phosphate O-acyltransferase activity
calcium ion binding
2-acylglycerol-3-phosphate O-acyltransferase activity
1-alkenylglycerophosphocholine O-acyltransferase activity
1-acylglycerophosphocholine O-acyltransferase activity
1-alkylglycerophosphocholine O-acyltransferase activity
1-alkylglycerophosphocholine O-acetyltransferase activity
phosphatidic acid biosynthetic process
phospholipid biosynthetic process
triglyceride biosynthetic process
phosphatidylglycerol acyl-chain remodeling
phosphatidylcholine acyl-chain remodeling
surfactant homeostasis
positive regulation of protein catabolic process
retina development in camera-type eye
negative regulation of phosphatidylcholine biosynthetic process
1-acylglycerol-3-phosphate O-acyltransferase activity
calcium ion binding
2-acylglycerol-3-phosphate O-acyltransferase activity
1-alkenylglycerophosphocholine O-acyltransferase activity
1-acylglycerophosphocholine O-acyltransferase activity
1-alkylglycerophosphocholine O-acyltransferase activity
1-alkylglycerophosphocholine O-acetyltransferase activity
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0500009863.hg.1 | ENST00000283415.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009863.hg.1 | ENST00000283415.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009863.hg.1 | ENST00000283415.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009863.hg.1 | ENST00000283415.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009863.hg.1 | ENST00000283415.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009863.hg.1 | ENST00000283415.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009863.hg.1 | ENST00000283415.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009863.hg.1 | ENST00000283415.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009863.hg.1 | ENST00000283415.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009863.hg.1 | ENST00000283415.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009863.hg.1 | ENST00000283415.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009863.hg.1 | ENST00000283415.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009863.hg.1 | ENST00000283415.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009863.hg.1 | ENST00000283415.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009863.hg.1 | ENST00000283415.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009863.hg.1 | ENST00000283415.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009863.hg.1 | ENST00000283415.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009863.hg.1 | ENST00000283415.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009863.hg.1 | ENST00000283415.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009863.hg.1 | ENST00000475622.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009863.hg.1 | ENST00000475622.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009863.hg.1 | ENST00000475622.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009863.hg.1 | ENST00000475622.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009863.hg.1 | ENST00000475622.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009863.hg.1 | ENST00000475622.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009863.hg.1 | ENST00000475622.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009863.hg.1 | ENST00000475622.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009863.hg.1 | ENST00000475622.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009863.hg.1 | ENST00000475622.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009863.hg.1 | ENST00000475622.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009863.hg.1 | ENST00000475622.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009863.hg.1 | ENST00000475622.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009863.hg.1 | ENST00000475622.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009863.hg.1 | ENST00000475622.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009863.hg.1 | ENST00000475622.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009863.hg.1 | ENST00000475622.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009863.hg.1 | ENST00000475622.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009863.hg.1 | ENST00000475622.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009863.hg.1 | ENST00000475622.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009863.hg.1 | ENST00000475622.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009863.hg.1 | ENST00000503252.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009863.hg.1 | ENST00000503252.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009863.hg.1 | ENST00000503252.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009863.hg.1 | ENST00000503252.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009863.hg.1 | ENST00000503252.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009863.hg.1 | ENST00000507282.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009863.hg.1 | ENST00000507282.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009863.hg.1 | ENST00000507282.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009863.hg.1 | ENST00000507282.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009863.hg.1 | ENST00000513757.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009863.hg.1 | ENST00000513757.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009863.hg.1 | ENST00000513757.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009863.hg.1 | ENST00000513757.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009863.hg.1 | ENST00000513757.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009863.hg.1 | ENST00000513757.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009863.hg.1 | ENST00000513757.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009863.hg.1 | ENST00000514484.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009863.hg.1 | ENST00000514484.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009863.hg.1 | ENST00000514484.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009863.hg.1 | ENST00000514484.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009863.hg.1 | ENST00000514484.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009863.hg.1 | ENST00000514484.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009863.hg.1 | ENST00000514484.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009863.hg.1 | ENST00000514484.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009863.hg.1 | NM_024830.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009863.hg.1 | NM_024830.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009863.hg.1 | NM_024830.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009863.hg.1 | NM_024830.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009863.hg.1 | NM_024830.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009863.hg.1 | NM_024830.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009863.hg.1 | NM_024830.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009863.hg.1 | NM_024830.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009863.hg.1 | NM_024830.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009863.hg.1 | NM_024830.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009863.hg.1 | NM_024830.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009863.hg.1 | NM_024830.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009863.hg.1 | NM_024830.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009863.hg.1 | NM_024830.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009863.hg.1 | NM_024830.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009863.hg.1 | NM_024830.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009863.hg.1 | NM_024830.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009863.hg.1 | NM_024830.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500009863.hg.1 | NM_024830.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR05016181.hg.1 | TC05001110.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR05016181.hg.1 | ENST00000283415.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05016181.hg.1 | ENST00000503252.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05016181.hg.1 | NM_024830.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR05016183.hg.1 | TC05001110.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR05016183.hg.1 | ENST00000283415.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05016183.hg.1 | NM_024830.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500169646.hg.1 | ENST00000283415.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500169646.hg.1 | ENST00000475622.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500169646.hg.1 | NM_024830.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500169646.hg.1 | ENST00000283415.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500169646.hg.1 | ENST00000475622.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500169646.hg.1 | NM_024830.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500169646.hg.1 | TC0500009863.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0500169644.hg.1 | ENST00000283415.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500169644.hg.1 | ENST00000475622.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500169644.hg.1 | ENST00000503252.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500169644.hg.1 | NM_024830.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500169644.hg.1 | ENST00000283415.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500169644.hg.1 | ENST00000475622.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500169644.hg.1 | ENST00000503252.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500169644.hg.1 | NM_024830.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500169644.hg.1 | TC0500009863.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0500009863.hg.1 | ENST00000283415.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009863.hg.1 | ENST00000283415.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009863.hg.1 | ENST00000283415.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009863.hg.1 | ENST00000283415.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009863.hg.1 | ENST00000283415.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009863.hg.1 | ENST00000283415.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009863.hg.1 | ENST00000283415.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009863.hg.1 | ENST00000283415.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009863.hg.1 | ENST00000283415.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009863.hg.1 | ENST00000283415.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009863.hg.1 | ENST00000283415.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009863.hg.1 | ENST00000283415.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009863.hg.1 | ENST00000283415.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009863.hg.1 | ENST00000283415.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009863.hg.1 | ENST00000283415.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009863.hg.1 | ENST00000283415.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009863.hg.1 | ENST00000283415.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009863.hg.1 | ENST00000283415.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009863.hg.1 | ENST00000283415.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009863.hg.1 | ENST00000475622.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009863.hg.1 | ENST00000475622.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009863.hg.1 | ENST00000475622.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009863.hg.1 | ENST00000475622.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009863.hg.1 | ENST00000475622.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009863.hg.1 | ENST00000475622.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009863.hg.1 | ENST00000475622.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009863.hg.1 | ENST00000475622.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009863.hg.1 | ENST00000475622.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009863.hg.1 | ENST00000475622.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009863.hg.1 | ENST00000475622.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009863.hg.1 | ENST00000475622.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009863.hg.1 | ENST00000475622.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009863.hg.1 | ENST00000475622.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009863.hg.1 | ENST00000475622.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009863.hg.1 | ENST00000475622.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009863.hg.1 | ENST00000475622.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009863.hg.1 | ENST00000475622.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009863.hg.1 | ENST00000475622.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009863.hg.1 | ENST00000475622.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009863.hg.1 | ENST00000475622.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009863.hg.1 | ENST00000503252.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009863.hg.1 | ENST00000503252.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009863.hg.1 | ENST00000503252.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009863.hg.1 | ENST00000503252.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009863.hg.1 | ENST00000503252.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009863.hg.1 | ENST00000507282.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009863.hg.1 | ENST00000507282.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009863.hg.1 | ENST00000507282.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009863.hg.1 | ENST00000507282.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009863.hg.1 | ENST00000513757.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009863.hg.1 | ENST00000513757.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009863.hg.1 | ENST00000513757.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009863.hg.1 | ENST00000513757.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009863.hg.1 | ENST00000513757.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009863.hg.1 | ENST00000513757.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009863.hg.1 | ENST00000513757.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009863.hg.1 | ENST00000514484.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009863.hg.1 | ENST00000514484.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009863.hg.1 | ENST00000514484.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009863.hg.1 | ENST00000514484.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009863.hg.1 | ENST00000514484.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009863.hg.1 | ENST00000514484.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009863.hg.1 | ENST00000514484.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009863.hg.1 | ENST00000514484.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500009863.hg.1 | NM_024830.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009863.hg.1 | NM_024830.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009863.hg.1 | NM_024830.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009863.hg.1 | NM_024830.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009863.hg.1 | NM_024830.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009863.hg.1 | NM_024830.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009863.hg.1 | NM_024830.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009863.hg.1 | NM_024830.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009863.hg.1 | NM_024830.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009863.hg.1 | NM_024830.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009863.hg.1 | NM_024830.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009863.hg.1 | NM_024830.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009863.hg.1 | NM_024830.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009863.hg.1 | NM_024830.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009863.hg.1 | NM_024830.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009863.hg.1 | NM_024830.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009863.hg.1 | NM_024830.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009863.hg.1 | NM_024830.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500009863.hg.1 | NM_024830.4 | Refseq |

LPCAT1
Genomic Map