Assay Details
| Gene Symbol: ARHGEF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_011510136.2 | XP_011508438.2 | 2 - 3 | 536 | 104 | |
| XM_011510137.2 | XP_011508439.2 | 2 - 3 | 536 | 104 | ||
| XM_011510139.2 | XP_011508441.1 | 2 - 3 | 536 | 104 | ||
| XM_017002794.1 | XP_016858283.1 | 2 - 3 | 536 | 104 | ||
| GenBank mRNA | AB014551.3 | - | 2 - 3 | 195 | 103 | |
| AK304548.1 | - | 1 - 2 | 261 | 104 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AB014551.3, AK304548.1 transcripts.
Set Membership: |
|

ARHGEF2
Genomic Map