Assay Details
| Gene Symbol: RYR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_000540.2 | NP_000531.2 | 82 - 83 | 11723 | 91 | |
| NM_001042723.1 | NP_001036188.1 | 81 - 82 | 11708 | 91 | ||
| XM_011527205.2 | XP_011525507.1 | 82 - 83 | 11673 | 91 | ||
| GenBank mRNA | A20359.1 | - | 81 - 82 | 11680 | 91 | |
| J05200.1 | - | 81 - 82 | 11680 | 91 | ||
More Information
Set Membership: |
|

RYR1
Genomic Map