Assay Details
Gene Symbol: PLEKHG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022835.2 | NP_073746.2 | 15 - 16 | 1815 | 63 | |
XM_005259163.2 | XP_005259220.1 | 15 - 16 | 1636 | 63 | ||
XM_006723334.2 | XP_006723397.1 | 15 - 16 | 2166 | 63 | ||
XM_011527232.2 | XP_011525534.1 | 15 - 16 | 1742 | 63 | ||
XM_017027151.1 | XP_016882640.1 | 14 - 15 | 2298 | 63 | ||
GenBank mRNA | AK024429.1 | - | 15 - 16 | 1815 | 63 | |
AK026111.1 | - | 15 - 16 | 1624 | 63 | ||
AK302476.1 | - | 15 - 16 | 1439 | 63 | ||
BC013426.1 | - | 5 - 6 | 306 | 63 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
