Assay Details
| Gene Symbol: IL17RA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_014339.6 | NP_055154.3 | 10 - 11 | 1079 | 64 | |
| GenBank mRNA | BC011624.2 | - | 10 - 11 | 1023 | 4053746 | 64 |
| CT841520.1 | - | 10 - 11 | 1002 | 64 | ||
| JF305973.1 | - | 10 - 11 | 946 | 64 | ||
| U58917.1 | - | 10 - 11 | 978 | 64 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC22000019.hg.1
Set Membership: |
|

IL17RA
Genomic Map