Assay Details
| Gene Symbol: IL17RA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_014339.6 | NP_055154.3 | 9 - 10 | 1070 | 75 | |
| GenBank mRNA | BC011624.2 | - | 9 - 10 | 1014 | 4053746 | 75 |
| CT841520.1 | - | 9 - 10 | 993 | 75 | ||
| JF305973.1 | - | 9 - 10 | 937 | 75 | ||
| U58917.1 | - | 9 - 10 | 969 | 75 | ||
More Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
cell surface receptor signaling pathway
cytokine-mediated signaling pathway
positive regulation of interleukin-23 production
defense response to fungus
granulocyte chemotaxis
fibroblast activation
positive regulation of cytokine production involved in inflammatory response
positive regulation of interleukin-5 secretion
positive regulation of interleukin-13 secretion
receptor binding
protein binding
interleukin-17 receptor activity
cell surface receptor signaling pathway
cytokine-mediated signaling pathway
positive regulation of interleukin-23 production
defense response to fungus
granulocyte chemotaxis
fibroblast activation
positive regulation of cytokine production involved in inflammatory response
positive regulation of interleukin-5 secretion
positive regulation of interleukin-13 secretion
receptor binding
protein binding
interleukin-17 receptor activity
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC2200006501.hg.1 | ENST00000319363.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006501.hg.1 | ENST00000319363.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006501.hg.1 | ENST00000319363.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006501.hg.1 | ENST00000319363.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006501.hg.1 | ENST00000319363.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006501.hg.1 | ENST00000319363.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006501.hg.1 | ENST00000319363.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006501.hg.1 | ENST00000319363.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006501.hg.1 | ENST00000319363.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006501.hg.1 | ENST00000319363.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006501.hg.1 | ENST00000319363.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006501.hg.1 | ENST00000319363.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006501.hg.1 | ENST00000319363.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006501.hg.1 | ENST00000319363.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006501.hg.1 | ENST00000319363.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006501.hg.1 | ENST00000319363.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006501.hg.1 | ENST00000319363.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006501.hg.1 | ENST00000319363.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006501.hg.1 | ENST00000319363.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006501.hg.1 | ENST00000319363.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006501.hg.1 | ENST00000319363.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006501.hg.1 | ENST00000319363.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006501.hg.1 | ENST00000459971.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006501.hg.1 | ENST00000459971.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006501.hg.1 | ENST00000459971.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006501.hg.1 | ENST00000459971.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006501.hg.1 | ENST00000477874.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006501.hg.1 | ENST00000477874.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006501.hg.1 | ENST00000477874.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006501.hg.1 | ENST00000477874.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006501.hg.1 | ENST00000477874.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006501.hg.1 | ENST00000477874.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006501.hg.1 | ENST00000477874.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006501.hg.1 | ENST00000612619.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006501.hg.1 | ENST00000612619.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006501.hg.1 | ENST00000612619.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006501.hg.1 | ENST00000612619.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006501.hg.1 | ENST00000612619.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006501.hg.1 | ENST00000612619.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006501.hg.1 | ENST00000612619.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006501.hg.1 | ENST00000612619.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006501.hg.1 | ENST00000612619.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006501.hg.1 | ENST00000612619.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006501.hg.1 | ENST00000612619.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006501.hg.1 | ENST00000612619.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006501.hg.1 | ENST00000612619.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006501.hg.1 | ENST00000612619.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006501.hg.1 | ENST00000612619.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006501.hg.1 | ENST00000612619.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006501.hg.1 | ENST00000612619.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006501.hg.1 | ENST00000612619.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006501.hg.1 | ENST00000612619.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006501.hg.1 | ENST00000612619.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006501.hg.1 | ENST00000612619.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006501.hg.1 | NM_001289905.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006501.hg.1 | NM_001289905.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006501.hg.1 | NM_001289905.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006501.hg.1 | NM_001289905.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006501.hg.1 | NM_001289905.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006501.hg.1 | NM_001289905.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006501.hg.1 | NM_001289905.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006501.hg.1 | NM_001289905.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006501.hg.1 | NM_001289905.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006501.hg.1 | NM_001289905.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006501.hg.1 | NM_001289905.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006501.hg.1 | NM_001289905.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006501.hg.1 | NM_001289905.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006501.hg.1 | NM_001289905.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006501.hg.1 | NM_001289905.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006501.hg.1 | NM_001289905.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006501.hg.1 | NM_001289905.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006501.hg.1 | NM_001289905.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006501.hg.1 | NM_001289905.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006501.hg.1 | NM_001289905.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006501.hg.1 | NM_001289905.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006501.hg.1 | NM_014339.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006501.hg.1 | NM_014339.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006501.hg.1 | NM_014339.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006501.hg.1 | NM_014339.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006501.hg.1 | NM_014339.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006501.hg.1 | NM_014339.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006501.hg.1 | NM_014339.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006501.hg.1 | NM_014339.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006501.hg.1 | NM_014339.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006501.hg.1 | NM_014339.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006501.hg.1 | NM_014339.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006501.hg.1 | NM_014339.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006501.hg.1 | NM_014339.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006501.hg.1 | NM_014339.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006501.hg.1 | NM_014339.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006501.hg.1 | NM_014339.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006501.hg.1 | NM_014339.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006501.hg.1 | NM_014339.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006501.hg.1 | NM_014339.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006501.hg.1 | NM_014339.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006501.hg.1 | NM_014339.6 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200006501.hg.1 | NM_014339.6 | Refseq |

IL17RA
Genomic Map