Assay Details
Gene Symbol: TRANK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_014831.2 | NP_055646.2 | 18 - 19 | 5502 | 83 | |
XM_006713432.3 | XP_006713495.1 | 19 - 20 | 5670 | 83 | ||
XM_011534289.2 | XP_011532591.1 | 19 - 20 | 6431 | 83 | ||
XM_017007570.1 | XP_016863059.1 | 18 - 19 | 5302 | 83 | ||
XM_017007571.1 | XP_016863060.1 | 19 - 20 | 5563 | 83 | ||
XM_017007572.1 | XP_016863061.1 | 19 - 20 | 5465 | 83 | ||
GenBank mRNA | AB002340.3 | - | 8 - 9 | 3880 | 83 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC03001283.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR03022004.hg.1 | TC03001283.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR03022004.hg.1 | ENST00000301807.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR03022004.hg.1 | ENST00000428977.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR03022004.hg.1 | ENST00000429976.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR03022004.hg.1 | NM_014831.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300010714.hg.1 | ENST00000429976.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300010714.hg.1 | ENST00000429976.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300010714.hg.1 | ENST00000429976.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300010714.hg.1 | ENST00000429976.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300010714.hg.1 | ENST00000429976.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300010714.hg.1 | ENST00000429976.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300010714.hg.1 | ENST00000429976.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300010714.hg.1 | ENST00000429976.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300010714.hg.1 | ENST00000429976.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300010714.hg.1 | ENST00000429976.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300010714.hg.1 | ENST00000429976.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300010714.hg.1 | ENST00000429976.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300010714.hg.1 | ENST00000429976.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300010714.hg.1 | ENST00000429976.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300010714.hg.1 | ENST00000429976.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300010714.hg.1 | ENST00000429976.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300010714.hg.1 | ENST00000429976.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300010714.hg.1 | ENST00000429976.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300010714.hg.1 | ENST00000429976.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300010714.hg.1 | ENST00000429976.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300010714.hg.1 | ENST00000429976.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300010714.hg.1 | ENST00000429976.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300010714.hg.1 | ENST00000429976.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300010714.hg.1 | ENST00000429976.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300010714.hg.1 | ENST00000429976.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300010714.hg.1 | ENST00000429976.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300010714.hg.1 | ENST00000429976.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300010714.hg.1 | ENST00000429976.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300010714.hg.1 | ENST00000463764.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300010714.hg.1 | ENST00000463764.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300010714.hg.1 | ENST00000463764.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300010714.hg.1 | ENST00000463764.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300010714.hg.1 | ENST00000463764.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300010714.hg.1 | ENST00000463984.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300010714.hg.1 | ENST00000463984.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300010714.hg.1 | ENST00000463984.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300010714.hg.1 | ENST00000463984.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300010714.hg.1 | NM_014831.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300010714.hg.1 | NM_014831.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300010714.hg.1 | NM_014831.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300010714.hg.1 | NM_014831.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300010714.hg.1 | NM_014831.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300010714.hg.1 | NM_014831.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300010714.hg.1 | NM_014831.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300010714.hg.1 | NM_014831.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300010714.hg.1 | NM_014831.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300010714.hg.1 | NM_014831.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300010714.hg.1 | NM_014831.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300010714.hg.1 | NM_014831.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300010714.hg.1 | NM_014831.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300010714.hg.1 | NM_014831.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300010714.hg.1 | NM_014831.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300010714.hg.1 | NM_014831.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300010714.hg.1 | NM_014831.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300010714.hg.1 | NM_014831.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300010714.hg.1 | NM_014831.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300010714.hg.1 | NM_014831.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300010714.hg.1 | NM_014831.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300010714.hg.1 | NM_014831.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300010714.hg.1 | NM_014831.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300010714.hg.1 | NM_014831.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300010714.hg.1 | NM_014831.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300010714.hg.1 | NM_014831.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300010714.hg.1 | NM_014831.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300010714.hg.1 | NM_014831.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300177924.hg.1 | ENST00000429976.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300177924.hg.1 | NM_014831.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300177924.hg.1 | ENST00000429976.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300177924.hg.1 | NM_014831.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300177924.hg.1 | TC0300010714.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0300010714.hg.1 | ENST00000429976.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300010714.hg.1 | ENST00000429976.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300010714.hg.1 | ENST00000429976.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300010714.hg.1 | ENST00000429976.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300010714.hg.1 | ENST00000429976.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300010714.hg.1 | ENST00000429976.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300010714.hg.1 | ENST00000429976.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300010714.hg.1 | ENST00000429976.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300010714.hg.1 | ENST00000429976.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300010714.hg.1 | ENST00000429976.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300010714.hg.1 | ENST00000429976.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300010714.hg.1 | ENST00000429976.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300010714.hg.1 | ENST00000429976.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300010714.hg.1 | ENST00000429976.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300010714.hg.1 | ENST00000429976.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300010714.hg.1 | ENST00000429976.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300010714.hg.1 | ENST00000429976.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300010714.hg.1 | ENST00000429976.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300010714.hg.1 | ENST00000429976.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300010714.hg.1 | ENST00000429976.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300010714.hg.1 | ENST00000429976.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300010714.hg.1 | ENST00000429976.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300010714.hg.1 | ENST00000429976.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300010714.hg.1 | ENST00000429976.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300010714.hg.1 | ENST00000429976.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300010714.hg.1 | ENST00000429976.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300010714.hg.1 | ENST00000429976.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300010714.hg.1 | ENST00000429976.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300010714.hg.1 | ENST00000463764.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300010714.hg.1 | ENST00000463764.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300010714.hg.1 | ENST00000463764.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300010714.hg.1 | ENST00000463764.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300010714.hg.1 | ENST00000463764.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300010714.hg.1 | ENST00000463984.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300010714.hg.1 | ENST00000463984.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300010714.hg.1 | ENST00000463984.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300010714.hg.1 | ENST00000463984.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300010714.hg.1 | NM_014831.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300010714.hg.1 | NM_014831.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300010714.hg.1 | NM_014831.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300010714.hg.1 | NM_014831.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300010714.hg.1 | NM_014831.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300010714.hg.1 | NM_014831.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300010714.hg.1 | NM_014831.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300010714.hg.1 | NM_014831.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300010714.hg.1 | NM_014831.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300010714.hg.1 | NM_014831.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300010714.hg.1 | NM_014831.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300010714.hg.1 | NM_014831.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300010714.hg.1 | NM_014831.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300010714.hg.1 | NM_014831.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300010714.hg.1 | NM_014831.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300010714.hg.1 | NM_014831.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300010714.hg.1 | NM_014831.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300010714.hg.1 | NM_014831.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300010714.hg.1 | NM_014831.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300010714.hg.1 | NM_014831.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300010714.hg.1 | NM_014831.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300010714.hg.1 | NM_014831.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300010714.hg.1 | NM_014831.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300010714.hg.1 | NM_014831.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300010714.hg.1 | NM_014831.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300010714.hg.1 | NM_014831.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300010714.hg.1 | NM_014831.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0300010714.hg.1 | NM_014831.2 | Refseq |
