Assay Details
Gene Symbol: LOC101927181 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK024457.1 | - | 1 | 4306 | 76 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | carbohydrate (chondroitin 4) sulfotransferase 12 | CHST12 |
C4S-2, C4ST-2, C4ST2 |
-11.0kb | Sense | Chr. 7 - 2403489 - 2434607 on NCBI Build GRCh38 |
Downstream | galectin-related inter-fiber protein | GRIFIN | 24.3kb | Anti-sense | Chr. 7 - 2473576 - 2476855 on NCBI Build GRCh38 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Agilent
Affymetrix
ABI
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Human Whole Genome ChIP Tiling Array | A_17_P25480425 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P05266876 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P05266905 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P15901708 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P05266895 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P05266878 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P05266884 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P05266873 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P05266872 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P05266904 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P05266891 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P05266881 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P05266885 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P05266866 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P05266896 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P05266875 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P25480385 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P05266861 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P05266869 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P05266899 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P25480435 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P05266870 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P25480381 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P05266902 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P05266857 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P05266867 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P25480383 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P05266906 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P05266887 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P05266898 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P05266892 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P05266865 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P25480430 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P25480398 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P05266888 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P25480386 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P05266894 | - | - |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 561200 | - | - |
Agilent SurePrint G3 Human CNV Microarray 2x400K | 114994 | - | - |
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_44k_32_P227615 | - | - |
Agilent Whole Human Genome Oligo Microarray 4x44K | 25344 | - | - |
Whole Human Genome Oligo Microarray G4112A | A_32_P227615 | - | - |
Agilent Human Genome 244K CGH Microarray (Alpha Test) | A_16_P01644792 | - | - |
Agilent Whole Human Genome Microarray 4x44K G4112F | A_32_P227615 | - | - |
Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 | 22897 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P01644792 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P01644804 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P01644802 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P17826423 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P37905241 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P01644789 | - | - |
Agilent Human 1B Microarray G4111A | A_32_P805094 | - | - |
Agilent SurePrint G3 Human CGH Microarray 2x400K | A_16_P01644804 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Gene 1.0 ST Array | 8131142 | - | - |
Affymetrix Human Gene 1.0 ST Array | 8131141 | - | - |
Affymetrix Human Gene 1.1 ST Array | 8131140 | - | - |
Human Genome U133 Plus 2.0 Array | 216455_PM_at | - | - |
Human Genome U133 Plus 2.0 Array | 216455_at | - | - |
Human Genome U133 Plus 2.0 Array | 216460_PM_at | - | - |
Human Genome U133 Plus 2.0 Array | 216460_at | - | - |
Human Genome U133A 2.0 Array | 216455_at | - | - |
Human Genome U133A 2.0 Array | 216460_at | - | - |
Human Genome U133A 2.0 Array | 216460_PM_at | - | - |
Human Genome U133A 2.0 Array | 216455_PM_at | - | - |
AffymetrixPathwork Diagnostics Pathchip | pw517426 | - | - |
AffymetrixPathwork Diagnostics Pathchip | pw522073 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 216455_at4 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 216460_at8 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 216460_at9 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 216455_at11 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 216460_at3 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 216460_at4 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 216455_at2 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 216460_at1 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 216460_at11 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 216455_at6 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 216455_at8 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 216460_at2 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 216460_at5 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 216460_at10 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 216455_at3 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 216455_at5 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 216460_at6 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 216460_at7 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 216455_at10 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 216455_at7 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 216455_at9 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 216455_at1 | - | - |
Human Genome U133 Set | 216455_PM_at | - | - |
Human Genome U133 Set | 216455_at | - | - |
Human Genome U133 Set | 216460_PM_at | - | - |
Human Genome U133 Set | 216460_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Human Genome Survey Microarray Version 2.0 | 229825 | - | - |
