Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015991.hg.1 |
ENST00000361813.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015991.hg.1 |
ENST00000361813.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015991.hg.1 |
ENST00000361813.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015991.hg.1 |
ENST00000361813.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015991.hg.1 |
ENST00000361813.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015991.hg.1 |
ENST00000361813.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015991.hg.1 |
ENST00000361813.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015991.hg.1 |
ENST00000361813.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015991.hg.1 |
ENST00000361813.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015991.hg.1 |
ENST00000361813.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015991.hg.1 |
ENST00000361813.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015991.hg.1 |
ENST00000361813.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015991.hg.1 |
ENST00000361813.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015991.hg.1 |
ENST00000361813.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015991.hg.1 |
ENST00000361813.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015991.hg.1 |
ENST00000361813.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015991.hg.1 |
ENST00000361813.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015991.hg.1 |
ENST00000361813.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015991.hg.1 |
ENST00000361813.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015991.hg.1 |
ENST00000361813.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015991.hg.1 |
ENST00000361813.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015991.hg.1 |
ENST00000361813.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015991.hg.1 |
ENST00000361813.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015991.hg.1 |
ENST00000361813.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015991.hg.1 |
ENST00000361813.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015991.hg.1 |
ENST00000361813.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015991.hg.1 |
ENST00000361813.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015991.hg.1 |
ENST00000361813.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015991.hg.1 |
ENST00000361813.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015991.hg.1 |
ENST00000361813.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015991.hg.1 |
ENST00000468993.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015991.hg.1 |
ENST00000468993.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015991.hg.1 |
ENST00000468993.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015991.hg.1 |
ENST00000473643.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015991.hg.1 |
ENST00000473643.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015991.hg.1 |
ENST00000473643.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015991.hg.1 |
ENST00000473643.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015991.hg.1 |
ENST00000476954.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015991.hg.1 |
ENST00000476954.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015991.hg.1 |
ENST00000476954.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015991.hg.1 |
ENST00000476954.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015991.hg.1 |
ENST00000476954.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015991.hg.1 |
ENST00000476954.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015991.hg.1 |
ENST00000489907.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015991.hg.1 |
ENST00000489907.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015991.hg.1 |
ENST00000489907.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015991.hg.1 |
ENST00000489907.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015991.hg.1 |
NM_015327.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015991.hg.1 |
NM_015327.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015991.hg.1 |
NM_015327.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015991.hg.1 |
NM_015327.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015991.hg.1 |
NM_015327.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015991.hg.1 |
NM_015327.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015991.hg.1 |
NM_015327.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015991.hg.1 |
NM_015327.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015991.hg.1 |
NM_015327.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015991.hg.1 |
NM_015327.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015991.hg.1 |
NM_015327.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015991.hg.1 |
NM_015327.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015991.hg.1 |
NM_015327.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015991.hg.1 |
NM_015327.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015991.hg.1 |
NM_015327.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015991.hg.1 |
NM_015327.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015991.hg.1 |
NM_015327.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015991.hg.1 |
NM_015327.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015991.hg.1 |
NM_015327.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015991.hg.1 |
NM_015327.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015991.hg.1 |
NM_015327.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015991.hg.1 |
NM_015327.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015991.hg.1 |
NM_015327.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015991.hg.1 |
NM_015327.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015991.hg.1 |
NM_015327.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015991.hg.1 |
NM_015327.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015991.hg.1 |
NM_015327.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015991.hg.1 |
NM_015327.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015991.hg.1 |
NM_015327.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100015991.hg.1 |
NM_015327.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01051966.hg.1 |
TC01003334.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01051966.hg.1 |
ENST00000361813.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01051966.hg.1 |
ENST00000368267.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01051966.hg.1 |
NM_015327.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015991.hg.1 |
ENST00000361813.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015991.hg.1 |
ENST00000361813.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015991.hg.1 |
ENST00000361813.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015991.hg.1 |
ENST00000361813.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015991.hg.1 |
ENST00000361813.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015991.hg.1 |
ENST00000361813.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015991.hg.1 |
ENST00000361813.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015991.hg.1 |
ENST00000361813.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015991.hg.1 |
ENST00000361813.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015991.hg.1 |
ENST00000361813.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015991.hg.1 |
ENST00000361813.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015991.hg.1 |
ENST00000361813.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015991.hg.1 |
ENST00000361813.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015991.hg.1 |
ENST00000361813.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015991.hg.1 |
ENST00000361813.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015991.hg.1 |
ENST00000361813.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015991.hg.1 |
ENST00000361813.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015991.hg.1 |
ENST00000361813.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015991.hg.1 |
ENST00000361813.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015991.hg.1 |
ENST00000361813.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015991.hg.1 |
ENST00000361813.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015991.hg.1 |
ENST00000361813.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015991.hg.1 |
ENST00000361813.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015991.hg.1 |
ENST00000361813.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015991.hg.1 |
ENST00000361813.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015991.hg.1 |
ENST00000361813.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015991.hg.1 |
ENST00000361813.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015991.hg.1 |
ENST00000361813.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015991.hg.1 |
ENST00000361813.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015991.hg.1 |
ENST00000361813.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015991.hg.1 |
ENST00000468993.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015991.hg.1 |
ENST00000468993.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015991.hg.1 |
ENST00000468993.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015991.hg.1 |
ENST00000473643.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015991.hg.1 |
ENST00000473643.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015991.hg.1 |
ENST00000473643.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015991.hg.1 |
ENST00000473643.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015991.hg.1 |
ENST00000476954.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015991.hg.1 |
ENST00000476954.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015991.hg.1 |
ENST00000476954.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015991.hg.1 |
ENST00000476954.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015991.hg.1 |
ENST00000476954.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015991.hg.1 |
ENST00000476954.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015991.hg.1 |
ENST00000489907.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015991.hg.1 |
ENST00000489907.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015991.hg.1 |
ENST00000489907.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015991.hg.1 |
ENST00000489907.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100015991.hg.1 |
NM_015327.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015991.hg.1 |
NM_015327.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015991.hg.1 |
NM_015327.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015991.hg.1 |
NM_015327.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015991.hg.1 |
NM_015327.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015991.hg.1 |
NM_015327.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015991.hg.1 |
NM_015327.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015991.hg.1 |
NM_015327.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015991.hg.1 |
NM_015327.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015991.hg.1 |
NM_015327.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015991.hg.1 |
NM_015327.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015991.hg.1 |
NM_015327.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015991.hg.1 |
NM_015327.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015991.hg.1 |
NM_015327.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015991.hg.1 |
NM_015327.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015991.hg.1 |
NM_015327.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015991.hg.1 |
NM_015327.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015991.hg.1 |
NM_015327.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015991.hg.1 |
NM_015327.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015991.hg.1 |
NM_015327.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015991.hg.1 |
NM_015327.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015991.hg.1 |
NM_015327.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015991.hg.1 |
NM_015327.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015991.hg.1 |
NM_015327.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015991.hg.1 |
NM_015327.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015991.hg.1 |
NM_015327.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015991.hg.1 |
NM_015327.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015991.hg.1 |
NM_015327.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015991.hg.1 |
NM_015327.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100015991.hg.1 |
NM_015327.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100221117.hg.1 |
ENST00000361813.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100221117.hg.1 |
NM_015327.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100221117.hg.1 |
ENST00000361813.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100221117.hg.1 |
NM_015327.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100221117.hg.1 |
TC0100015991.hg.1 |
Clariom D TC |