Assay Details
Gene Symbol: LOC284454 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_036515.1 | 1 | 1721 | 77 | ||
GenBank mRNA | AL832183.1 | - | 1 | 1717 | 77 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
ABI
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 708184 | - | - |
Agilent SurePrint G3 Human CNV Microarray 2x400K | 102606 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3461633 | - | - |
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_44k_24_P734060 | - | - |
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_44k_32_P444104 | - | - |
Agilent Whole Human Genome Oligo Microarray 4x44K | 45084 | - | - |
Agilent Whole Human Genome Oligo Microarray 4x44K | 1004 | - | - |
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 46565 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3461633 | - | - |
SurePrint G3 Human GE Microarray G4851A | A_33_P3461633 | - | - |
Whole Human Genome Oligo Microarray G4112A | A_24_P734060 | - | - |
Whole Human Genome Oligo Microarray G4112A | A_32_P444104 | - | - |
Agilent Whole Human Genome Microarray 4x44K G4112F | A_24_P734060 | - | - |
Agilent Whole Human Genome Microarray 4x44K G4112F | A_32_P444104 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P03425977 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_18_P12990723 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P03425975 | - | - |
Agilent Human 1B Microarray G4111A | A_32_P444104 | - | - |
Agilent SurePrint G3 Human CGH Microarray 2x400K | A_16_P03425975 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix HT HG-U133+ PM Array Plate | 235317_PM_at | - | - |
Affymetrix HT HG-U133+ PM Array Plate | 1555847_PM_a_at | - | - |
Affymetrix Human Transcriptome Array 2.0 | TC1900008427.hg.1 | ENST00000599924.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008427.hg.1 | ENST00000599924.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1900008427.hg.1 | ENST00000599924.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008427.hg.1 | ENST00000599924.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008427.hg.1 | ENST00000599924.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1900008427.hg.1 | ENST00000599924.1 | Ensembl |
Human Genome U133 Plus 2.0 Array | 1555847_a_at | - | - |
Human Genome U133 Plus 2.0 Array | 235317_at | - | - |
Human Genome U133 Plus 2.0 Array | 1555847_PM_a_at | - | - |
Human Genome U133 Plus 2.0 Array | 235317_PM_at | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1555847_a_at1 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1555847_a_at2 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1555847_a_at11 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 235317_at9 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 235317_at7 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 235317_at3 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 235317_at1 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1555847_a_at10 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1555847_a_at5 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 235317_at4 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1555847_a_at7 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1555847_a_at6 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 235317_at8 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1555847_a_at3 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 235317_at5 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1555847_a_at9 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1555847_a_at4 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 1555847_a_at8 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 235317_at10 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 235317_at6 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 235317_at11 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 235317_at2 | - | - |
Human Genome U95 Set | 86762_i_at | - | - |
Human Genome U95 Set | 86762_PM_i_at | - | - |
Human Genome U133 Set | 235317_at | - | - |
Human Genome U133 Set | 235317_PM_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A011120 | - | - |
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A011120 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Human Genome Survey Microarray Version 2.0 | 234758 | - | - |
