Assay Details
| Gene Symbol: APPBP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001282476.1 | NP_001269405.1 | 2 - 3 | 597 | 82 | |
| NM_006380.3 | NP_006371.2 | 3 - 4 | 686 | 82 | ||
| XM_011524198.2 | XP_011522500.1 | 4 - 5 | 413 | 82 | ||
| GenBank mRNA | AF017782.1 | - | 3 - 4 | 662 | 82 | |
| AK292227.1 | - | 3 - 4 | 453 | 82 | ||
| AK307666.1 | - | 2 - 3 | 491 | 82 | ||
| BC018121.1 | - | 3 - 4 | 589 | 3916891 | 82 | |
| D86981.1 | - | 3 - 4 | 665 | 82 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC17001764.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | ENST00000083182.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | ENST00000083182.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | ENST00000083182.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | ENST00000083182.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | ENST00000083182.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | ENST00000083182.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | ENST00000083182.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | ENST00000083182.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | ENST00000083182.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | ENST00000083182.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | ENST00000083182.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | ENST00000083182.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | ENST00000083182.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | ENST00000083182.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | ENST00000083182.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | ENST00000083182.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | ENST00000083182.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | ENST00000083182.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | ENST00000083182.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | ENST00000083182.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | ENST00000585368.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | ENST00000585368.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | ENST00000585368.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | ENST00000585368.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | ENST00000588668.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | ENST00000588668.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | ENST00000588668.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | ENST00000588668.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | ENST00000588668.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | ENST00000589341.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | ENST00000589341.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | ENST00000589341.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | ENST00000589341.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | ENST00000589341.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | ENST00000589341.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | ENST00000589341.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | ENST00000589341.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | ENST00000589341.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | ENST00000589341.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | ENST00000589341.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | ENST00000589341.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | ENST00000589341.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | ENST00000589341.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | ENST00000589341.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | ENST00000590244.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | ENST00000590244.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | ENST00000590244.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | ENST00000590244.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | ENST00000590244.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | ENST00000590244.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | ENST00000590244.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | ENST00000590244.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | ENST00000592995.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | ENST00000592995.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | ENST00000592995.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | ENST00000592995.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | ENST00000592995.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | ENST00000592995.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | ENST00000592995.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | ENST00000592995.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | NM_001282476.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | NM_001282476.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | NM_001282476.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | NM_001282476.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | NM_001282476.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | NM_001282476.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | NM_001282476.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | NM_001282476.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | NM_001282476.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | NM_001282476.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | NM_001282476.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | NM_001282476.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | NM_001282476.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | NM_001282476.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | NM_001282476.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | NM_001282476.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | NM_001282476.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | NM_001282476.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | NM_001282476.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | NM_006380.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | NM_006380.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | NM_006380.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | NM_006380.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | NM_006380.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | NM_006380.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | NM_006380.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | NM_006380.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | NM_006380.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | NM_006380.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | NM_006380.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | NM_006380.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | NM_006380.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | NM_006380.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | NM_006380.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | NM_006380.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | NM_006380.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | NM_006380.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | NM_006380.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | NM_006380.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17023554.hg.1 | ENST00000083182.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17023554.hg.1 | NM_006380.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17023554.hg.1 | TC17001764.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | ENST00000083182.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | ENST00000083182.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | ENST00000083182.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | ENST00000083182.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | ENST00000083182.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | ENST00000083182.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | ENST00000083182.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | ENST00000083182.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | ENST00000083182.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | ENST00000083182.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | ENST00000083182.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | ENST00000083182.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | ENST00000083182.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | ENST00000083182.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | ENST00000083182.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | ENST00000083182.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | ENST00000083182.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | ENST00000083182.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | ENST00000083182.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | ENST00000083182.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | ENST00000585368.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | ENST00000585368.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | ENST00000585368.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | ENST00000585368.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | ENST00000588668.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | ENST00000588668.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | ENST00000588668.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | ENST00000588668.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | ENST00000588668.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | ENST00000589341.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | ENST00000589341.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | ENST00000589341.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | ENST00000589341.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | ENST00000589341.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | ENST00000589341.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | ENST00000589341.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | ENST00000589341.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | ENST00000589341.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | ENST00000589341.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | ENST00000589341.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | ENST00000589341.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | ENST00000589341.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | ENST00000589341.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | ENST00000589341.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | ENST00000590244.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | ENST00000590244.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | ENST00000590244.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | ENST00000590244.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | ENST00000590244.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | ENST00000590244.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | ENST00000590244.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | ENST00000590244.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | ENST00000592995.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | ENST00000592995.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | ENST00000592995.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | ENST00000592995.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | ENST00000592995.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | ENST00000592995.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | ENST00000592995.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | ENST00000592995.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | NM_001282476.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | NM_001282476.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | NM_001282476.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | NM_001282476.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | NM_001282476.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | NM_001282476.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | NM_001282476.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | NM_001282476.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | NM_001282476.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | NM_001282476.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | NM_001282476.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | NM_001282476.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | NM_001282476.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | NM_001282476.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | NM_001282476.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | NM_001282476.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | NM_001282476.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | NM_001282476.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | NM_001282476.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | NM_006380.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | NM_006380.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | NM_006380.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | NM_006380.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | NM_006380.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | NM_006380.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | NM_006380.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | NM_006380.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | NM_006380.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | NM_006380.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | NM_006380.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | NM_006380.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | NM_006380.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | NM_006380.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | NM_006380.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | NM_006380.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | NM_006380.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | NM_006380.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | NM_006380.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | NM_006380.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700191258.hg.1 | ENST00000083182.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700191258.hg.1 | ENST00000588668.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700191258.hg.1 | ENST00000589341.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700191258.hg.1 | ENST00000590244.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700191258.hg.1 | NM_001282476.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700191258.hg.1 | NM_006380.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700191258.hg.1 | ENST00000083182.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700191258.hg.1 | ENST00000588668.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700191258.hg.1 | ENST00000589341.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700191258.hg.1 | ENST00000590244.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700191258.hg.1 | NM_001282476.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700191258.hg.1 | NM_006380.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700191258.hg.1 | TC1700011319.hg.1 | Clariom D TC |

APPBP2
Genomic Map