Assay Details
Gene Symbol: APPBP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001282476.1 | NP_001269405.1 | 3 - 4 | 721 | 65 | |
NM_006380.3 | NP_006371.2 | 4 - 5 | 810 | 65 | ||
XM_011524198.2 | XP_011522500.1 | 5 - 6 | 537 | 65 | ||
GenBank mRNA | AF017782.1 | - | 4 - 5 | 786 | 65 | |
AK292227.1 | - | 4 - 5 | 577 | 65 | ||
AK307666.1 | - | 3 - 4 | 615 | 65 | ||
BC018121.1 | - | 4 - 5 | 713 | 3916891 | 65 | |
D86981.1 | - | 4 - 5 | 789 | 65 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC17001764.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | ENST00000083182.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | ENST00000083182.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | ENST00000083182.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | ENST00000083182.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | ENST00000083182.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | ENST00000083182.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | ENST00000083182.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | ENST00000083182.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | ENST00000083182.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | ENST00000083182.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | ENST00000083182.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | ENST00000083182.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | ENST00000083182.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | ENST00000083182.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | ENST00000083182.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | ENST00000083182.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | ENST00000083182.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | ENST00000083182.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | ENST00000083182.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | ENST00000083182.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | ENST00000585368.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | ENST00000585368.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | ENST00000585368.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | ENST00000585368.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | ENST00000588668.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | ENST00000588668.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | ENST00000588668.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | ENST00000588668.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | ENST00000588668.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | ENST00000589341.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | ENST00000589341.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | ENST00000589341.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | ENST00000589341.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | ENST00000589341.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | ENST00000589341.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | ENST00000589341.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | ENST00000589341.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | ENST00000589341.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | ENST00000589341.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | ENST00000589341.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | ENST00000589341.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | ENST00000589341.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | ENST00000589341.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | ENST00000589341.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | ENST00000590244.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | ENST00000590244.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | ENST00000590244.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | ENST00000590244.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | ENST00000590244.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | ENST00000590244.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | ENST00000590244.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | ENST00000590244.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | ENST00000592995.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | ENST00000592995.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | ENST00000592995.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | ENST00000592995.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | ENST00000592995.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | ENST00000592995.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | ENST00000592995.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | ENST00000592995.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | NM_001282476.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | NM_001282476.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | NM_001282476.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | NM_001282476.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | NM_001282476.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | NM_001282476.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | NM_001282476.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | NM_001282476.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | NM_001282476.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | NM_001282476.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | NM_001282476.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | NM_001282476.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | NM_001282476.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | NM_001282476.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | NM_001282476.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | NM_001282476.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | NM_001282476.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | NM_001282476.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | NM_001282476.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | NM_006380.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | NM_006380.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | NM_006380.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | NM_006380.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | NM_006380.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | NM_006380.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | NM_006380.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | NM_006380.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | NM_006380.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | NM_006380.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | NM_006380.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | NM_006380.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | NM_006380.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | NM_006380.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | NM_006380.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | NM_006380.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | NM_006380.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | NM_006380.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | NM_006380.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011319.hg.1 | NM_006380.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR17023553.hg.1 | ENST00000083182.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR17023553.hg.1 | NM_006380.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR17023553.hg.1 | TC17001764.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | ENST00000083182.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | ENST00000083182.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | ENST00000083182.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | ENST00000083182.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | ENST00000083182.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | ENST00000083182.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | ENST00000083182.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | ENST00000083182.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | ENST00000083182.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | ENST00000083182.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | ENST00000083182.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | ENST00000083182.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | ENST00000083182.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | ENST00000083182.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | ENST00000083182.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | ENST00000083182.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | ENST00000083182.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | ENST00000083182.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | ENST00000083182.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | ENST00000083182.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | ENST00000585368.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | ENST00000585368.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | ENST00000585368.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | ENST00000585368.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | ENST00000588668.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | ENST00000588668.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | ENST00000588668.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | ENST00000588668.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | ENST00000588668.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | ENST00000589341.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | ENST00000589341.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | ENST00000589341.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | ENST00000589341.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | ENST00000589341.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | ENST00000589341.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | ENST00000589341.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | ENST00000589341.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | ENST00000589341.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | ENST00000589341.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | ENST00000589341.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | ENST00000589341.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | ENST00000589341.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | ENST00000589341.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | ENST00000589341.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | ENST00000590244.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | ENST00000590244.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | ENST00000590244.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | ENST00000590244.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | ENST00000590244.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | ENST00000590244.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | ENST00000590244.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | ENST00000590244.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | ENST00000592995.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | ENST00000592995.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | ENST00000592995.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | ENST00000592995.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | ENST00000592995.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | ENST00000592995.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | ENST00000592995.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | ENST00000592995.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | NM_001282476.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | NM_001282476.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | NM_001282476.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | NM_001282476.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | NM_001282476.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | NM_001282476.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | NM_001282476.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | NM_001282476.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | NM_001282476.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | NM_001282476.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | NM_001282476.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | NM_001282476.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | NM_001282476.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | NM_001282476.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | NM_001282476.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | NM_001282476.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | NM_001282476.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | NM_001282476.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | NM_001282476.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | NM_006380.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | NM_006380.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | NM_006380.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | NM_006380.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | NM_006380.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | NM_006380.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | NM_006380.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | NM_006380.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | NM_006380.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | NM_006380.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | NM_006380.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | NM_006380.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | NM_006380.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | NM_006380.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | NM_006380.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | NM_006380.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | NM_006380.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | NM_006380.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | NM_006380.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1700011319.hg.1 | NM_006380.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700191254.hg.1 | ENST00000083182.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700191254.hg.1 | ENST00000589341.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700191254.hg.1 | ENST00000590244.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700191254.hg.1 | NM_001282476.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700191254.hg.1 | NM_006380.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700191254.hg.1 | ENST00000083182.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700191254.hg.1 | ENST00000589341.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700191254.hg.1 | ENST00000590244.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700191254.hg.1 | NM_001282476.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700191254.hg.1 | NM_006380.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700191254.hg.1 | TC1700011319.hg.1 | Clariom D TC |
